Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 154280504-154281007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX36
Entrez Gene ID
170506
Gene Name
DEAH-box helicase 36
Gene Aliases
DDX36, G4R1, MLEL1, RHAU
UniGene
Hs.446270
Gene Chromosome Location
Chr.3: 154275668-154324497 on Build GRCh38
Transcripts
NM_001114397.1 NM_020865.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4391670 c.2358G>C p.Q786H Chr.3: 154280839-154280839 on Build GRCh38
1661442 c.2564A>T p.K855I Chr.3: 154280582-154280582 on Build GRCh38
6428117 c.2498G>A p.R833Q Chr.3: 154280606-154280606 on Build GRCh38
7133468 c.2416G>A p.E806K Chr.3: 154280781-154280781 on Build GRCh38
4337390 c.2518_2519insA p.R840fs*19 Chr.3: 154280585-154280586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371002609 Chr.3: 154280673-154280673 on Build GRCh38
rs769719815 Chr.3: 154280696-154280696 on Build GRCh38
rs755988740 Chr.3: 154280895-154280895 on Build GRCh38
rs756689810 Chr.3: 154280669-154280669 on Build GRCh38
rs755083794 Chr.3: 154280688-154280688 on Build GRCh38
rs375116031 Chr.3: 154280914-154280914 on Build GRCh38
rs112993756 Chr.3: 154280676-154280676 on Build GRCh38
rs752006898 Chr.3: 154280906-154280907 on Build GRCh38
rs767053229 Chr.3: 154280716-154280717 on Build GRCh38
rs761963036 Chr.3: 154280556-154280558 on Build GRCh38
rs146446831 Chr.3: 154280853-154280853 on Build GRCh38
rs1035914470 Chr.3: 154280587-154280587 on Build GRCh38
rs780981635 Chr.3: 154280681-154280681 on Build GRCh38
rs376754975 Chr.3: 154280906-154280906 on Build GRCh38
rs761583165 Chr.3: 154280552-154280552 on Build GRCh38
rs773428560 Chr.3: 154280711-154280712 on Build GRCh38
rs745739630 Chr.3: 154280535-154280539 on Build GRCh38
rs531134804 Chr.3: 154280948-154280948 on Build GRCh38
rs769359215 Chr.3: 154280614-154280614 on Build GRCh38
rs956741079 Chr.3: 154280673-154280674 on Build GRCh38
rs762931017 Chr.3: 154280643-154280643 on Build GRCh38
rs199890757 Chr.3: 154280579-154280579 on Build GRCh38
rs979242085 Chr.3: 154280721-154280721 on Build GRCh38
rs778970022 Chr.3: 154280674-154280674 on Build GRCh38
rs1020645509 Chr.3: 154280694-154280694 on Build GRCh38
rs761178422 Chr.3: 154280744-154280744 on Build GRCh38
rs891026092 Chr.3: 154280924-154280924 on Build GRCh38
rs577053711 Chr.3: 154280720-154280720 on Build GRCh38
rs754409173 Chr.3: 154280803-154280803 on Build GRCh38
rs201549631 Chr.3: 154280757-154280757 on Build GRCh38
rs758626024 Chr.3: 154280875-154280875 on Build GRCh38
rs530250010 Chr.3: 154280843-154280843 on Build GRCh38
rs766272317 Chr.3: 154280941-154280941 on Build GRCh38
rs752478425 Chr.3: 154280886-154280886 on Build GRCh38
rs780323433 Chr.3: 154280884-154280884 on Build GRCh38
rs764725753 Chr.3: 154280672-154280672 on Build GRCh38
rs748107331 Chr.3: 154280548-154280548 on Build GRCh38
rs115358645 Chr.3: 154280830-154280830 on Build GRCh38
rs770517946 Chr.3: 154280722-154280722 on Build GRCh38
rs372394653 Chr.3: 154280729-154280729 on Build GRCh38
rs772493680 Chr.3: 154280735-154280735 on Build GRCh38
rs563741813 Chr.3: 154280945-154280945 on Build GRCh38
rs763070791 Chr.3: 154280715-154280716 on Build GRCh38
rs554210711 Chr.3: 154280549-154280549 on Build GRCh38
rs943987851 Chr.3: 154280928-154280928 on Build GRCh38
rs770099570 Chr.3: 154280695-154280699 on Build GRCh38
rs766398981 Chr.3: 154280639-154280639 on Build GRCh38
rs775988489 Chr.3: 154280736-154280736 on Build GRCh38
rs765938551 Chr.3: 154280559-154280559 on Build GRCh38
rs768012758 Chr.3: 154280625-154280625 on Build GRCh38
rs759272615 Chr.3: 154280733-154280733 on Build GRCh38
rs112204498 Chr.3: 154280663-154280663 on Build GRCh38
rs771729616 Chr.3: 154280535-154280537 on Build GRCh38
rs774105156 Chr.3: 154280560-154280560 on Build GRCh38
rs749282482 Chr.3: 154280908-154280908 on Build GRCh38
rs527309363 Chr.3: 154280599-154280599 on Build GRCh38
rs991636221 Chr.3: 154280641-154280641 on Build GRCh38
rs762595349 Chr.3: 154280550-154280550 on Build GRCh38
rs891383913 Chr.3: 154280932-154280932 on Build GRCh38
rs1000868093 Chr.3: 154280584-154280585 on Build GRCh38
rs368441104 Chr.3: 154280620-154280620 on Build GRCh38
rs899978752 Chr.3: 154280964-154280964 on Build GRCh38
rs947785930 Chr.3: 154280711-154280711 on Build GRCh38
rs777828802 Chr.3: 154280713-154280713 on Build GRCh38
rs776957093 Chr.3: 154280538-154280538 on Build GRCh38

View Full Product Details