Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 155763571-155764118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf33
Entrez Gene ID
285315
Gene Name
chromosome 3 open reading frame 33
Gene Aliases
AC3-33
UniGene
Hs.350846
Gene Chromosome Location
Chr.3: 155762612-155806287 on Build GRCh38
Transcripts
NM_001308229.1 NM_173657.2 XM_005247379.4 XM_011512710.2 XM_011512711.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7322134 c.747T>G p.D249E Chr.3: 155763655-155763655 on Build GRCh38
1617109 c.358C>A p.L120M Chr.3: 155763897-155763897 on Build GRCh38
6301625 c.655T>A p.W219R Chr.3: 155763600-155763600 on Build GRCh38
1632927 c.493G>A p.G165R Chr.3: 155763762-155763762 on Build GRCh38
5697130 c.656G>T p.W219L Chr.3: 155763599-155763599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761276267 Chr.3: 155763684-155763684 on Build GRCh38
rs780436730 Chr.3: 155763789-155763789 on Build GRCh38
rs771659485 Chr.3: 155763855-155763855 on Build GRCh38
rs762429849 Chr.3: 155763736-155763736 on Build GRCh38
rs1010494344 Chr.3: 155763981-155763981 on Build GRCh38
rs769797856 Chr.3: 155763662-155763662 on Build GRCh38
rs1049443950 Chr.3: 155763964-155763964 on Build GRCh38
rs113890115 Chr.3: 155763651-155763651 on Build GRCh38
rs764823659 Chr.3: 155763762-155763764 on Build GRCh38
rs764618620 Chr.3: 155763733-155763733 on Build GRCh38
rs748793393 Chr.3: 155763666-155763666 on Build GRCh38
rs1041819349 Chr.3: 155763602-155763602 on Build GRCh38
rs999541816 Chr.3: 155764066-155764066 on Build GRCh38
rs758679808 Chr.3: 155763775-155763775 on Build GRCh38
rs749859513 Chr.3: 155763765-155763767 on Build GRCh38
rs575233706 Chr.3: 155763782-155763782 on Build GRCh38
rs115731681 Chr.3: 155763904-155763904 on Build GRCh38
rs1024755082 Chr.3: 155763772-155763772 on Build GRCh38
rs763138200 Chr.3: 155763947-155763947 on Build GRCh38
rs768256976 Chr.3: 155763609-155763609 on Build GRCh38
rs977450790 Chr.3: 155763998-155763998 on Build GRCh38
rs764518767 Chr.3: 155763625-155763625 on Build GRCh38
rs576856906 Chr.3: 155763606-155763606 on Build GRCh38
rs766658152 Chr.3: 155763961-155763961 on Build GRCh38
rs376049300 Chr.3: 155763658-155763658 on Build GRCh38
rs903428940 Chr.3: 155764030-155764030 on Build GRCh38
rs933299338 Chr.3: 155764049-155764049 on Build GRCh38
rs571937562 Chr.3: 155763694-155763694 on Build GRCh38
rs750597545 Chr.3: 155763774-155763774 on Build GRCh38
rs770945064 Chr.3: 155763836-155763836 on Build GRCh38
rs984130638 Chr.3: 155763796-155763796 on Build GRCh38
rs773359213 Chr.3: 155763926-155763926 on Build GRCh38
rs7653384 Chr.3: 155763820-155763820 on Build GRCh38
rs1010546435 Chr.3: 155763626-155763626 on Build GRCh38
rs559220741 Chr.3: 155763706-155763706 on Build GRCh38
rs757310431 Chr.3: 155763637-155763637 on Build GRCh38
rs901831860 Chr.3: 155763628-155763628 on Build GRCh38
rs751861179 Chr.3: 155763968-155763968 on Build GRCh38
rs753885223 Chr.3: 155763629-155763629 on Build GRCh38
rs201584610 Chr.3: 155763665-155763665 on Build GRCh38
rs893570004 Chr.3: 155763972-155763972 on Build GRCh38
rs766362355 Chr.3: 155763917-155763917 on Build GRCh38
rs770619794 Chr.3: 155763743-155763743 on Build GRCh38
rs759161457 Chr.3: 155763693-155763693 on Build GRCh38
rs775769963 Chr.3: 155763703-155763703 on Build GRCh38
rs776314192 Chr.3: 155763673-155763674 on Build GRCh38
rs755894625 Chr.3: 155763819-155763819 on Build GRCh38
rs527782923 Chr.3: 155764016-155764016 on Build GRCh38
rs942267332 Chr.3: 155764073-155764073 on Build GRCh38
rs745444960 Chr.3: 155763845-155763845 on Build GRCh38
rs889162578 Chr.3: 155764072-155764072 on Build GRCh38
rs770372441 Chr.3: 155763674-155763674 on Build GRCh38
rs923368757 Chr.3: 155764018-155764018 on Build GRCh38
rs778772192 Chr.3: 155763781-155763781 on Build GRCh38
rs761170768 Chr.3: 155763621-155763621 on Build GRCh38
rs772069197 Chr.3: 155763858-155763858 on Build GRCh38
rs749161316 Chr.3: 155763830-155763830 on Build GRCh38
rs752474590 Chr.3: 155763792-155763792 on Build GRCh38
rs761011278 Chr.3: 155763595-155763595 on Build GRCh38
rs1042449535 Chr.3: 155764025-155764025 on Build GRCh38
rs773963276 Chr.3: 155763684-155763684 on Build GRCh38
rs779145276 Chr.3: 155763639-155763639 on Build GRCh38
rs926529641 Chr.3: 155763882-155763882 on Build GRCh38
rs374247931 Chr.3: 155763915-155763915 on Build GRCh38
rs993232711 Chr.3: 155763756-155763756 on Build GRCh38
rs753103866 Chr.3: 155763608-155763608 on Build GRCh38
rs147770210 Chr.3: 155763903-155763903 on Build GRCh38
rs267599663 Chr.3: 155763895-155763895 on Build GRCh38
rs751458858 Chr.3: 155763931-155763933 on Build GRCh38
rs765199403 Chr.3: 155763753-155763753 on Build GRCh38
rs747337825 Chr.3: 155763879-155763879 on Build GRCh38
rs775150071 Chr.3: 155763870-155763870 on Build GRCh38
rs952664076 Chr.3: 155763773-155763773 on Build GRCh38
rs1050860468 Chr.3: 155764051-155764051 on Build GRCh38
rs778285413 Chr.3: 155763843-155763843 on Build GRCh38
rs750650694 Chr.3: 155763648-155763648 on Build GRCh38
rs762988845 Chr.3: 155763769-155763769 on Build GRCh38

View Full Product Details