Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 155806009-155806550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf33
Entrez Gene ID
285315
Gene Name
chromosome 3 open reading frame 33
Gene Aliases
AC3-33
UniGene
Hs.350846
Gene Chromosome Location
Chr.3: 155762612-155806287 on Build GRCh38
Transcripts
NM_001308229.1 NM_173657.2 XM_011512710.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6397500 c.113G>A p.R38Q Chr.3: 155806140-155806140 on Build GRCh38
6420034 c.88G>A p.A30T Chr.3: 155806165-155806165 on Build GRCh38
6408827 c.67G>A p.V23M Chr.3: 155806186-155806186 on Build GRCh38
3392155 c.62A>G p.N21S Chr.3: 155806191-155806191 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs951847116 Chr.3: 155806229-155806229 on Build GRCh38
rs1014743161 Chr.3: 155806453-155806453 on Build GRCh38
rs752761755 Chr.3: 155806301-155806301 on Build GRCh38
rs1054521697 Chr.3: 155806245-155806245 on Build GRCh38
rs529870341 Chr.3: 155806430-155806430 on Build GRCh38
rs1055045558 Chr.3: 155806483-155806483 on Build GRCh38
rs564190185 Chr.3: 155806238-155806238 on Build GRCh38
rs113650032 Chr.3: 155806356-155806356 on Build GRCh38
rs932145975 Chr.3: 155806468-155806468 on Build GRCh38
rs1034186752 Chr.3: 155806143-155806143 on Build GRCh38
rs982427897 Chr.3: 155806367-155806367 on Build GRCh38
rs569643608 Chr.3: 155806191-155806191 on Build GRCh38
rs12486921 Chr.3: 155806061-155806061 on Build GRCh38
rs900189230 Chr.3: 155806168-155806168 on Build GRCh38
rs540406524 Chr.3: 155806284-155806284 on Build GRCh38
rs936132575 Chr.3: 155806333-155806333 on Build GRCh38
rs866618851 Chr.3: 155806145-155806145 on Build GRCh38
rs1018597731 Chr.3: 155806129-155806129 on Build GRCh38
rs114877878 Chr.3: 155806226-155806226 on Build GRCh38
rs982449322 Chr.3: 155806224-155806224 on Build GRCh38
rs750541775 Chr.3: 155806044-155806044 on Build GRCh38
rs762707842 Chr.3: 155806205-155806205 on Build GRCh38
rs932154681 Chr.3: 155806241-155806241 on Build GRCh38
rs912273169 Chr.3: 155806379-155806379 on Build GRCh38
rs944452207 Chr.3: 155806351-155806351 on Build GRCh38
rs1010671357 Chr.3: 155806048-155806048 on Build GRCh38
rs948734789 Chr.3: 155806282-155806282 on Build GRCh38
rs891986802 Chr.3: 155806345-155806345 on Build GRCh38
rs1020276681 Chr.3: 155806247-155806247 on Build GRCh38
rs753828818 Chr.3: 155806462-155806462 on Build GRCh38
rs973880269 Chr.3: 155806180-155806180 on Build GRCh38
rs891563536 Chr.3: 155806088-155806088 on Build GRCh38
rs1040564294 Chr.3: 155806353-155806353 on Build GRCh38
rs756818755 Chr.3: 155806472-155806472 on Build GRCh38
rs974333781 Chr.3: 155806303-155806303 on Build GRCh38
rs995805832 Chr.3: 155806375-155806375 on Build GRCh38
rs189688427 Chr.3: 155806502-155806502 on Build GRCh38
rs900166983 Chr.3: 155806358-155806358 on Build GRCh38
rs761243525 Chr.3: 155806125-155806125 on Build GRCh38
rs768014362 Chr.3: 155806268-155806268 on Build GRCh38
rs1020119477 Chr.3: 155806049-155806049 on Build GRCh38
rs893206638 Chr.3: 155806516-155806516 on Build GRCh38
rs936164001 Chr.3: 155806059-155806059 on Build GRCh38
rs929637318 Chr.3: 155806204-155806204 on Build GRCh38
rs370899297 Chr.3: 155806386-155806386 on Build GRCh38
rs755993870 Chr.3: 155806328-155806328 on Build GRCh38
rs1004643177 Chr.3: 155806444-155806444 on Build GRCh38
rs980318152 Chr.3: 155806285-155806285 on Build GRCh38
rs1026698779 Chr.3: 155806343-155806343 on Build GRCh38
rs917255566 Chr.3: 155806272-155806272 on Build GRCh38
rs1027489450 Chr.3: 155806380-155806380 on Build GRCh38
rs992918786 Chr.3: 155806477-155806477 on Build GRCh38
rs769441633 Chr.3: 155806149-155806149 on Build GRCh38
rs958539687 Chr.3: 155806299-155806300 on Build GRCh38
rs961496531 Chr.3: 155806251-155806251 on Build GRCh38
rs922180725 Chr.3: 155806445-155806445 on Build GRCh38
rs764062155 Chr.3: 155806355-155806355 on Build GRCh38
rs543936719 Chr.3: 155806344-155806344 on Build GRCh38
rs760030246 Chr.3: 155806240-155806240 on Build GRCh38
rs993346836 Chr.3: 155806267-155806267 on Build GRCh38
rs981234239 Chr.3: 155806140-155806140 on Build GRCh38
rs887713249 Chr.3: 155806385-155806385 on Build GRCh38
rs528468416 Chr.3: 155806246-155806246 on Build GRCh38
rs996044580 Chr.3: 155806220-155806220 on Build GRCh38

View Full Product Details