Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 157569753-157570257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PQLC2L
Entrez Gene ID
152078
Gene Name
PQ loop repeat containing 2 like
Gene Aliases
C3orf55
UniGene
Hs.259046
Gene Chromosome Location
Chr.3: 157543344-157601233 on Build GRCh38
Transcripts
NM_001099777.3 NM_001130001.2 NM_001130002.2 NM_001243729.1 NR_024016.2 XM_017005754.1

SNP Detail

SNP ID Chromosome Location
rs1042111142 Chr.3: 157570087-157570087 on Build GRCh38
rs898164042 Chr.3: 157569942-157569942 on Build GRCh38
rs975382623 Chr.3: 157569779-157569779 on Build GRCh38
rs930955777 Chr.3: 157570154-157570154 on Build GRCh38
rs1045502617 Chr.3: 157569869-157569869 on Build GRCh38
rs1026303778 Chr.3: 157569983-157569983 on Build GRCh38
rs927571239 Chr.3: 157569781-157569781 on Build GRCh38
rs900799316 Chr.3: 157569879-157569879 on Build GRCh38
rs889722689 Chr.3: 157569905-157569905 on Build GRCh38
rs981646937 Chr.3: 157570088-157570088 on Build GRCh38
rs928389304 Chr.3: 157570111-157570111 on Build GRCh38
rs1055462488 Chr.3: 157570134-157570134 on Build GRCh38
rs1055347383 Chr.3: 157569921-157569921 on Build GRCh38
rs980538811 Chr.3: 157570010-157570010 on Build GRCh38
rs1005488303 Chr.3: 157569936-157569936 on Build GRCh38
rs1005437359 Chr.3: 157570160-157570160 on Build GRCh38
rs922371832 Chr.3: 157570094-157570094 on Build GRCh38
rs1054576522 Chr.3: 157569814-157569814 on Build GRCh38
rs542814499 Chr.3: 157570124-157570125 on Build GRCh38
rs937130539 Chr.3: 157570125-157570125 on Build GRCh38
rs571363864 Chr.3: 157570029-157570029 on Build GRCh38
rs539967336 Chr.3: 157570059-157570059 on Build GRCh38
rs561859078 Chr.3: 157569999-157569999 on Build GRCh38
rs183499421 Chr.3: 157570028-157570028 on Build GRCh38
rs559091033 Chr.3: 157569807-157569807 on Build GRCh38
rs913371052 Chr.3: 157570041-157570041 on Build GRCh38
rs998291774 Chr.3: 157569883-157569883 on Build GRCh38
rs937014748 Chr.3: 157569897-157569897 on Build GRCh38
rs896988030 Chr.3: 157570187-157570187 on Build GRCh38
rs964092500 Chr.3: 157570011-157570011 on Build GRCh38
rs1024849545 Chr.3: 157569975-157569975 on Build GRCh38
rs548606428 Chr.3: 157569951-157569951 on Build GRCh38
rs140562468 Chr.3: 157569793-157569793 on Build GRCh38
rs545116095 Chr.3: 157569886-157569891 on Build GRCh38
rs180920088 Chr.3: 157570017-157570017 on Build GRCh38
rs994213548 Chr.3: 157569966-157569966 on Build GRCh38
rs761602504 Chr.3: 157569783-157569783 on Build GRCh38
rs1020162802 Chr.3: 157570221-157570221 on Build GRCh38
rs531014784 Chr.3: 157570057-157570057 on Build GRCh38
rs117098017 Chr.3: 157569792-157569792 on Build GRCh38
rs913423718 Chr.3: 157569841-157569841 on Build GRCh38
rs1016748489 Chr.3: 157569941-157569941 on Build GRCh38
rs184105380 Chr.3: 157569843-157569843 on Build GRCh38
rs542921131 Chr.3: 157570182-157570182 on Build GRCh38
rs188448335 Chr.3: 157569894-157569894 on Build GRCh38
rs527970878 Chr.3: 157569913-157569913 on Build GRCh38
rs77822679 Chr.3: 157569846-157569846 on Build GRCh38
rs150762909 Chr.3: 157570147-157570147 on Build GRCh38
rs1049836271 Chr.3: 157570131-157570131 on Build GRCh38
rs538872833 Chr.3: 157570052-157570052 on Build GRCh38
rs563415819 Chr.3: 157570014-157570014 on Build GRCh38
rs142915324 Chr.3: 157569985-157569985 on Build GRCh38
rs933652393 Chr.3: 157570126-157570126 on Build GRCh38

View Full Product Details