Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 157578201-157578711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PQLC2L
Entrez Gene ID
152078
Gene Name
PQ loop repeat containing 2 like
Gene Aliases
C3orf55
UniGene
Hs.259046
Gene Chromosome Location
Chr.3: 157543344-157601233 on Build GRCh38
Transcripts
NM_001099777.3 NM_001130001.2 NM_001130002.2 NM_001243729.1 XM_017005754.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4542008 c.310G>A p.E104K Chr.3: 157578286-157578286 on Build GRCh38
5986218 c.328C>T p.L110F Chr.3: 157578304-157578304 on Build GRCh38
4920351 c.336C>T p.F112F Chr.3: 157578312-157578312 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs556193288 Chr.3: 157578346-157578346 on Build GRCh38
rs756656271 Chr.3: 157578671-157578671 on Build GRCh38
rs761136460 Chr.3: 157578381-157578381 on Build GRCh38
rs909731548 Chr.3: 157578325-157578325 on Build GRCh38
rs931022107 Chr.3: 157578629-157578629 on Build GRCh38
rs76526368 Chr.3: 157578432-157578432 on Build GRCh38
rs370364918 Chr.3: 157578271-157578271 on Build GRCh38
rs973954615 Chr.3: 157578605-157578605 on Build GRCh38
rs920774333 Chr.3: 157578616-157578616 on Build GRCh38
rs991192652 Chr.3: 157578256-157578256 on Build GRCh38
rs1035116147 Chr.3: 157578555-157578556 on Build GRCh38
rs1024087244 Chr.3: 157578625-157578625 on Build GRCh38
rs544937395 Chr.3: 157578374-157578374 on Build GRCh38
rs549798097 Chr.3: 157578586-157578586 on Build GRCh38
rs775672915 Chr.3: 157578377-157578377 on Build GRCh38
rs902708286 Chr.3: 157578503-157578503 on Build GRCh38
rs374639055 Chr.3: 157578239-157578239 on Build GRCh38
rs769577934 Chr.3: 157578333-157578333 on Build GRCh38
rs780890727 Chr.3: 157578328-157578328 on Build GRCh38
rs368403100 Chr.3: 157578330-157578330 on Build GRCh38
rs746153311 Chr.3: 157578317-157578317 on Build GRCh38
rs1035597251 Chr.3: 157578311-157578311 on Build GRCh38
rs773986007 Chr.3: 157578351-157578351 on Build GRCh38
rs759222951 Chr.3: 157578352-157578352 on Build GRCh38
rs563148436 Chr.3: 157578470-157578470 on Build GRCh38
rs773137494 Chr.3: 157578342-157578342 on Build GRCh38
rs898207506 Chr.3: 157578232-157578232 on Build GRCh38
rs753213543 Chr.3: 157578242-157578242 on Build GRCh38
rs71788167 Chr.3: 157578537-157578538 on Build GRCh38
rs754799255 Chr.3: 157578320-157578320 on Build GRCh38
rs60530908 Chr.3: 157578558-157578559 on Build GRCh38
rs910835628 Chr.3: 157578559-157578559 on Build GRCh38
rs577713212 Chr.3: 157578359-157578359 on Build GRCh38
rs368164404 Chr.3: 157578248-157578248 on Build GRCh38
rs560957261 Chr.3: 157578461-157578461 on Build GRCh38
rs774004092 Chr.3: 157578238-157578238 on Build GRCh38
rs201247963 Chr.3: 157578296-157578296 on Build GRCh38
rs762479715 Chr.3: 157578231-157578231 on Build GRCh38
rs769435188 Chr.3: 157578538-157578539 on Build GRCh38
rs372293767 Chr.3: 157578536-157578536 on Build GRCh38
rs368338960 Chr.3: 157578385-157578385 on Build GRCh38
rs202066379 Chr.3: 157578285-157578285 on Build GRCh38
rs531169352 Chr.3: 157578535-157578535 on Build GRCh38
rs961107252 Chr.3: 157578557-157578557 on Build GRCh38
rs572729354 Chr.3: 157578409-157578409 on Build GRCh38
rs147729645 Chr.3: 157578536-157578537 on Build GRCh38
rs781644773 Chr.3: 157578637-157578637 on Build GRCh38
rs746089504 Chr.3: 157578283-157578285 on Build GRCh38
rs987703920 Chr.3: 157578553-157578553 on Build GRCh38
rs573916280 Chr.3: 157578295-157578295 on Build GRCh38
rs372776979 Chr.3: 157578347-157578347 on Build GRCh38
rs555753331 Chr.3: 157578260-157578260 on Build GRCh38
rs765948195 Chr.3: 157578234-157578234 on Build GRCh38
rs1021477149 Chr.3: 157578375-157578375 on Build GRCh38
rs66748594 Chr.3: 157578639-157578639 on Build GRCh38
rs187905437 Chr.3: 157578372-157578372 on Build GRCh38
rs984058521 Chr.3: 157578675-157578675 on Build GRCh38
rs777372938 Chr.3: 157578241-157578241 on Build GRCh38
rs999609861 Chr.3: 157578510-157578510 on Build GRCh38
rs954732021 Chr.3: 157578466-157578466 on Build GRCh38

View Full Product Details