Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 158646078-158646582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFM1
Entrez Gene ID
85476
Gene Name
G elongation factor mitochondrial 1
Gene Aliases
COXPD1, EFG, EFG1, EFGM, EGF1, GFM, hEFG1
UniGene
Hs.518355
Gene Chromosome Location
Chr.3: 158644497-158692572 on Build GRCh38
Transcripts
NM_001308164.1 NM_001308166.1 NM_024996.5 XM_006713795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1420277 c.335A>C p.K112T Chr.3: 158646265-158646265 on Build GRCh38
4114864 c.266T>C p.M89T Chr.3: 158646196-158646196 on Build GRCh38
40822 c.277G>A p.E93K Chr.3: 158646207-158646207 on Build GRCh38
1040509 c.337G>T p.D113Y Chr.3: 158646267-158646267 on Build GRCh38
729464 c.273C>A p.S91S Chr.3: 158646203-158646203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762488275 Chr.3: 158646150-158646150 on Build GRCh38
rs781700550 Chr.3: 158646198-158646198 on Build GRCh38
rs776444859 Chr.3: 158646308-158646308 on Build GRCh38
rs764785102 Chr.3: 158646430-158646430 on Build GRCh38
rs1017672590 Chr.3: 158646530-158646530 on Build GRCh38
rs776494488 Chr.3: 158646117-158646117 on Build GRCh38
rs751161500 Chr.3: 158646329-158646329 on Build GRCh38
rs756718556 Chr.3: 158646237-158646237 on Build GRCh38
rs755786837 Chr.3: 158646233-158646239 on Build GRCh38
rs369021613 Chr.3: 158646509-158646509 on Build GRCh38
rs749429253 Chr.3: 158646104-158646104 on Build GRCh38
rs201727979 Chr.3: 158646184-158646184 on Build GRCh38
rs779253646 Chr.3: 158646274-158646274 on Build GRCh38
rs755447585 Chr.3: 158646191-158646191 on Build GRCh38
rs745537487 Chr.3: 158646311-158646311 on Build GRCh38
rs554128756 Chr.3: 158646419-158646419 on Build GRCh38
rs568400227 Chr.3: 158646319-158646319 on Build GRCh38
rs775118583 Chr.3: 158646114-158646114 on Build GRCh38
rs549720594 Chr.3: 158646285-158646285 on Build GRCh38
rs779634587 Chr.3: 158646387-158646387 on Build GRCh38
rs891431378 Chr.3: 158646358-158646358 on Build GRCh38
rs863224029 Chr.3: 158646168-158646168 on Build GRCh38
rs904809779 Chr.3: 158646424-158646424 on Build GRCh38
rs771204961 Chr.3: 158646108-158646108 on Build GRCh38
rs371977811 Chr.3: 158646224-158646224 on Build GRCh38
rs764690450 Chr.3: 158646133-158646133 on Build GRCh38
rs753187838 Chr.3: 158646204-158646204 on Build GRCh38
rs765930439 Chr.3: 158646324-158646324 on Build GRCh38
rs543336282 Chr.3: 158646536-158646536 on Build GRCh38
rs959536251 Chr.3: 158646503-158646503 on Build GRCh38
rs1033809327 Chr.3: 158646502-158646502 on Build GRCh38
rs113884757 Chr.3: 158646124-158646124 on Build GRCh38
rs752400894 Chr.3: 158646220-158646221 on Build GRCh38
rs1042725027 Chr.3: 158646403-158646403 on Build GRCh38
rs1046925437 Chr.3: 158646166-158646166 on Build GRCh38
rs911930051 Chr.3: 158646548-158646548 on Build GRCh38
rs771055207 Chr.3: 158646262-158646262 on Build GRCh38
rs529240264 Chr.3: 158646332-158646332 on Build GRCh38
rs568116224 Chr.3: 158646457-158646457 on Build GRCh38
rs757926529 Chr.3: 158646323-158646323 on Build GRCh38
rs759266378 Chr.3: 158646334-158646334 on Build GRCh38
rs768377828 Chr.3: 158646294-158646294 on Build GRCh38
rs767937072 Chr.3: 158646136-158646137 on Build GRCh38
rs536285807 Chr.3: 158646397-158646397 on Build GRCh38
rs749974602 Chr.3: 158646136-158646136 on Build GRCh38
rs201304690 Chr.3: 158646151-158646151 on Build GRCh38
rs369317856 Chr.3: 158646146-158646146 on Build GRCh38
rs373371132 Chr.3: 158646312-158646312 on Build GRCh38
rs879213259 Chr.3: 158646371-158646371 on Build GRCh38
rs572044445 Chr.3: 158646357-158646357 on Build GRCh38
rs531728845 Chr.3: 158646210-158646210 on Build GRCh38
rs955065891 Chr.3: 158646519-158646519 on Build GRCh38
rs761692289 Chr.3: 158646310-158646310 on Build GRCh38
rs56075263 Chr.3: 158646544-158646544 on Build GRCh38
rs751861999 Chr.3: 158646177-158646177 on Build GRCh38
rs375459830 Chr.3: 158646227-158646227 on Build GRCh38
rs2303910 Chr.3: 158646520-158646520 on Build GRCh38
rs951060350 Chr.3: 158646534-158646534 on Build GRCh38
rs908378995 Chr.3: 158646209-158646209 on Build GRCh38
rs553835635 Chr.3: 158646284-158646284 on Build GRCh38
rs773835614 Chr.3: 158646420-158646420 on Build GRCh38
rs767953284 Chr.3: 158646344-158646344 on Build GRCh38
rs999522136 Chr.3: 158646449-158646449 on Build GRCh38
rs369439447 Chr.3: 158646248-158646248 on Build GRCh38
rs777663037 Chr.3: 158646238-158646238 on Build GRCh38
rs1056918334 Chr.3: 158646372-158646372 on Build GRCh38
rs538813935 Chr.3: 158646116-158646116 on Build GRCh38
rs863224033 Chr.3: 158646203-158646203 on Build GRCh38
rs746460484 Chr.3: 158646390-158646390 on Build GRCh38

View Full Product Details