Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 158651968-158652478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFM1
Entrez Gene ID
85476
Gene Name
G elongation factor mitochondrial 1
Gene Aliases
COXPD1, EFG, EFG1, EFGM, EGF1, GFM, hEFG1
UniGene
Hs.518355
Gene Chromosome Location
Chr.3: 158644497-158692572 on Build GRCh38
Transcripts
NM_001308164.1 NM_001308166.1 NM_024996.5 XM_006713795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4584068 c.750G>T p.R250R Chr.3: 158652156-158652156 on Build GRCh38
3005724 c.798G>T p.E266D Chr.3: 158652204-158652204 on Build GRCh38
281357 c.797A>G p.E266G Chr.3: 158652203-158652203 on Build GRCh38
4114868 c.756G>A p.E252E Chr.3: 158652162-158652162 on Build GRCh38
4114867 c.731C>T p.A244V Chr.3: 158652137-158652137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778902849 Chr.3: 158652094-158652097 on Build GRCh38
rs35152071 Chr.3: 158652146-158652147 on Build GRCh38
rs538662087 Chr.3: 158652250-158652250 on Build GRCh38
rs1007055354 Chr.3: 158652018-158652018 on Build GRCh38
rs551478401 Chr.3: 158652077-158652077 on Build GRCh38
rs9810874 Chr.3: 158652009-158652009 on Build GRCh38
rs863224032 Chr.3: 158652106-158652106 on Build GRCh38
rs201621454 Chr.3: 158652143-158652143 on Build GRCh38
rs563041850 Chr.3: 158652429-158652429 on Build GRCh38
rs201360324 Chr.3: 158652111-158652111 on Build GRCh38
rs866197920 Chr.3: 158652351-158652351 on Build GRCh38
rs575835640 Chr.3: 158652394-158652394 on Build GRCh38
rs567363063 Chr.3: 158652003-158652003 on Build GRCh38
rs937833290 Chr.3: 158652435-158652435 on Build GRCh38
rs779127183 Chr.3: 158652282-158652282 on Build GRCh38
rs772686112 Chr.3: 158652190-158652190 on Build GRCh38
rs751966573 Chr.3: 158652292-158652292 on Build GRCh38
rs533254444 Chr.3: 158652439-158652439 on Build GRCh38
rs964267142 Chr.3: 158652313-158652313 on Build GRCh38
rs201685981 Chr.3: 158652091-158652091 on Build GRCh38
rs182221029 Chr.3: 158652401-158652401 on Build GRCh38
rs753449665 Chr.3: 158652262-158652262 on Build GRCh38
rs780670778 Chr.3: 158652299-158652299 on Build GRCh38
rs374427028 Chr.3: 158652217-158652217 on Build GRCh38
rs369346675 Chr.3: 158652272-158652272 on Build GRCh38
rs556268062 Chr.3: 158652172-158652172 on Build GRCh38
rs116385487 Chr.3: 158652338-158652338 on Build GRCh38
rs755679024 Chr.3: 158652252-158652252 on Build GRCh38
rs755783105 Chr.3: 158652071-158652071 on Build GRCh38
rs754969878 Chr.3: 158652153-158652153 on Build GRCh38
rs931007959 Chr.3: 158652377-158652377 on Build GRCh38
rs374061938 Chr.3: 158652025-158652026 on Build GRCh38
rs1039889051 Chr.3: 158652350-158652350 on Build GRCh38
rs776938494 Chr.3: 158652207-158652207 on Build GRCh38
rs942520711 Chr.3: 158652344-158652344 on Build GRCh38
rs958860760 Chr.3: 158652196-158652196 on Build GRCh38
rs905296441 Chr.3: 158652409-158652412 on Build GRCh38
rs200032705 Chr.3: 158652140-158652140 on Build GRCh38
rs751348727 Chr.3: 158652426-158652426 on Build GRCh38
rs777468400 Chr.3: 158652156-158652156 on Build GRCh38
rs759365696 Chr.3: 158652230-158652230 on Build GRCh38
rs745718158 Chr.3: 158652126-158652126 on Build GRCh38
rs377418512 Chr.3: 158652108-158652108 on Build GRCh38
rs56362855 Chr.3: 158652000-158652000 on Build GRCh38
rs567208295 Chr.3: 158652089-158652089 on Build GRCh38
rs55992564 Chr.3: 158652354-158652354 on Build GRCh38
rs375512235 Chr.3: 158652182-158652182 on Build GRCh38
rs763523648 Chr.3: 158652025-158652025 on Build GRCh38
rs377037517 Chr.3: 158652169-158652169 on Build GRCh38
rs758828876 Chr.3: 158652293-158652293 on Build GRCh38
rs767528432 Chr.3: 158652236-158652236 on Build GRCh38
rs768991090 Chr.3: 158652205-158652205 on Build GRCh38
rs752251570 Chr.3: 158652155-158652155 on Build GRCh38
rs1046501617 Chr.3: 158652409-158652409 on Build GRCh38
rs778088445 Chr.3: 158652072-158652072 on Build GRCh38
rs373030031 Chr.3: 158652079-158652079 on Build GRCh38
rs878859919 Chr.3: 158652314-158652314 on Build GRCh38
rs762127044 Chr.3: 158652211-158652211 on Build GRCh38
rs935241149 Chr.3: 158652323-158652323 on Build GRCh38
rs892517916 Chr.3: 158652074-158652074 on Build GRCh38
rs774215742 Chr.3: 158652226-158652226 on Build GRCh38
rs553465700 Chr.3: 158652330-158652330 on Build GRCh38
rs112368036 Chr.3: 158652000-158652000 on Build GRCh38
rs977373803 Chr.3: 158652302-158652302 on Build GRCh38
rs192374828 Chr.3: 158652335-158652335 on Build GRCh38
rs111853162 Chr.3: 158652010-158652011 on Build GRCh38
rs185807995 Chr.3: 158652258-158652258 on Build GRCh38
rs747710750 Chr.3: 158652070-158652070 on Build GRCh38
rs139430866 Chr.3: 158652154-158652154 on Build GRCh38
rs760959060 Chr.3: 158652109-158652109 on Build GRCh38
rs913941688 Chr.3: 158652400-158652400 on Build GRCh38
rs115984741 Chr.3: 158652194-158652194 on Build GRCh38
rs150024625 Chr.3: 158652160-158652160 on Build GRCh38
rs180934936 Chr.3: 158652056-158652056 on Build GRCh38
rs766836270 Chr.3: 158652283-158652283 on Build GRCh38
rs914631990 Chr.3: 158652268-158652268 on Build GRCh38
rs923897975 Chr.3: 158652310-158652310 on Build GRCh38
rs954460694 Chr.3: 158652290-158652290 on Build GRCh38
rs766213740 Chr.3: 158652141-158652141 on Build GRCh38
rs867158942 Chr.3: 158652049-158652049 on Build GRCh38
rs150903461 Chr.3: 158652137-158652137 on Build GRCh38
rs898169717 Chr.3: 158652078-158652078 on Build GRCh38
rs747155546 Chr.3: 158652203-158652203 on Build GRCh38
rs1020187204 Chr.3: 158652297-158652298 on Build GRCh38
rs750841861 Chr.3: 158652287-158652287 on Build GRCh38
rs771865940 Chr.3: 158652232-158652233 on Build GRCh38
rs757551544 Chr.3: 158652163-158652163 on Build GRCh38
rs879807884 Chr.3: 158652213-158652213 on Build GRCh38

View Full Product Details