Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 160225852-160226392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf80
Entrez Gene ID
401097
Gene Name
chromosome 3 open reading frame 80
Gene Aliases
-
UniGene
Hs.729755
Gene Chromosome Location
Chr.3: 160225636-160228213 on Build GRCh38
Transcripts
NM_001168214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5663440 c.113G>T p.R38L Chr.3: 160225955-160225955 on Build GRCh38
5675753 c.144A>T p.G48G Chr.3: 160225986-160225986 on Build GRCh38
4987831 c.191G>A p.G64E Chr.3: 160226033-160226033 on Build GRCh38
5032345 c.177G>A p.P59P Chr.3: 160226019-160226019 on Build GRCh38
87472 c.176C>T p.P59L Chr.3: 160226018-160226018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901072306 Chr.3: 160226194-160226194 on Build GRCh38
rs903009062 Chr.3: 160226004-160226004 on Build GRCh38
rs902599781 Chr.3: 160226208-160226208 on Build GRCh38
rs868184667 Chr.3: 160226276-160226276 on Build GRCh38
rs1022575775 Chr.3: 160226049-160226049 on Build GRCh38
rs771146945 Chr.3: 160226071-160226071 on Build GRCh38
rs776278119 Chr.3: 160226115-160226116 on Build GRCh38
rs961007320 Chr.3: 160226362-160226362 on Build GRCh38
rs754975775 Chr.3: 160225882-160225882 on Build GRCh38
rs139484601 Chr.3: 160226062-160226062 on Build GRCh38
rs531152164 Chr.3: 160226033-160226033 on Build GRCh38
rs1042330858 Chr.3: 160226180-160226180 on Build GRCh38
rs981155822 Chr.3: 160226061-160226061 on Build GRCh38
rs546861198 Chr.3: 160226115-160226115 on Build GRCh38
rs535572881 Chr.3: 160226172-160226172 on Build GRCh38
rs1046985865 Chr.3: 160225902-160225902 on Build GRCh38
rs920143000 Chr.3: 160226135-160226135 on Build GRCh38
rs1049769866 Chr.3: 160225982-160225982 on Build GRCh38
rs775962517 Chr.3: 160226004-160226004 on Build GRCh38
rs764161762 Chr.3: 160226145-160226145 on Build GRCh38
rs747328781 Chr.3: 160226040-160226042 on Build GRCh38
rs932617553 Chr.3: 160225968-160225968 on Build GRCh38
rs1042462955 Chr.3: 160226191-160226191 on Build GRCh38
rs945541748 Chr.3: 160226139-160226139 on Build GRCh38
rs747660323 Chr.3: 160226289-160226289 on Build GRCh38
rs780818702 Chr.3: 160225958-160225958 on Build GRCh38
rs998061943 Chr.3: 160226226-160226226 on Build GRCh38
rs746815437 Chr.3: 160226171-160226171 on Build GRCh38
rs769557972 Chr.3: 160226046-160226046 on Build GRCh38
rs777359507 Chr.3: 160226338-160226338 on Build GRCh38
rs569233933 Chr.3: 160226349-160226349 on Build GRCh38
rs867160733 Chr.3: 160225874-160225874 on Build GRCh38
rs998708045 Chr.3: 160226018-160226018 on Build GRCh38
rs974694838 Chr.3: 160225926-160225926 on Build GRCh38
rs754945034 Chr.3: 160226260-160226260 on Build GRCh38
rs770643668 Chr.3: 160226325-160226325 on Build GRCh38
rs965056928 Chr.3: 160226004-160226021 on Build GRCh38
rs551033898 Chr.3: 160226263-160226263 on Build GRCh38
rs1046602872 Chr.3: 160226333-160226333 on Build GRCh38
rs985732270 Chr.3: 160226161-160226161 on Build GRCh38
rs920190098 Chr.3: 160225962-160225962 on Build GRCh38
rs961802450 Chr.3: 160226083-160226083 on Build GRCh38
rs768413497 Chr.3: 160226039-160226040 on Build GRCh38
rs779121195 Chr.3: 160226363-160226363 on Build GRCh38
rs953797795 Chr.3: 160226306-160226306 on Build GRCh38
rs912563305 Chr.3: 160226166-160226166 on Build GRCh38
rs757520939 Chr.3: 160226167-160226167 on Build GRCh38
rs768059103 Chr.3: 160226129-160226129 on Build GRCh38
rs938073598 Chr.3: 160226116-160226116 on Build GRCh38
rs529008906 Chr.3: 160226334-160226334 on Build GRCh38
rs753949156 Chr.3: 160226154-160226154 on Build GRCh38
rs568566401 Chr.3: 160226143-160226143 on Build GRCh38
rs891160368 Chr.3: 160225995-160225995 on Build GRCh38
rs773068598 Chr.3: 160226053-160226053 on Build GRCh38
rs892990349 Chr.3: 160226330-160226330 on Build GRCh38
rs944085168 Chr.3: 160225996-160225996 on Build GRCh38
rs185340430 Chr.3: 160226355-160226355 on Build GRCh38
rs748847483 Chr.3: 160226343-160226343 on Build GRCh38
rs759925512 Chr.3: 160226118-160226118 on Build GRCh38
rs934465973 Chr.3: 160226319-160226319 on Build GRCh38
rs751410853 Chr.3: 160226232-160226232 on Build GRCh38
rs376126153 Chr.3: 160226035-160226035 on Build GRCh38
rs775531076 Chr.3: 160226132-160226132 on Build GRCh38

View Full Product Details