Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 160227088-160227599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf80
Entrez Gene ID
401097
Gene Name
chromosome 3 open reading frame 80
Gene Aliases
-
UniGene
Hs.729755
Gene Chromosome Location
Chr.3: 160225636-160228213 on Build GRCh38
Transcripts
NM_001168214.1

SNP Detail

SNP ID Chromosome Location
rs955478561 Chr.3: 160227274-160227274 on Build GRCh38
rs553483968 Chr.3: 160227557-160227557 on Build GRCh38
rs4680571 Chr.3: 160227422-160227422 on Build GRCh38
rs1029830062 Chr.3: 160227517-160227517 on Build GRCh38
rs752098599 Chr.3: 160227119-160227119 on Build GRCh38
rs1003501483 Chr.3: 160227553-160227553 on Build GRCh38
rs1006897324 Chr.3: 160227544-160227544 on Build GRCh38
rs1035073296 Chr.3: 160227307-160227308 on Build GRCh38
rs1053018750 Chr.3: 160227507-160227507 on Build GRCh38
rs866861201 Chr.3: 160227128-160227128 on Build GRCh38
rs911183536 Chr.3: 160227317-160227317 on Build GRCh38
rs931211451 Chr.3: 160227383-160227385 on Build GRCh38
rs1030191038 Chr.3: 160227270-160227270 on Build GRCh38
rs1011127047 Chr.3: 160227182-160227182 on Build GRCh38
rs754129599 Chr.3: 160227427-160227428 on Build GRCh38
rs116763145 Chr.3: 160227172-160227172 on Build GRCh38
rs967254154 Chr.3: 160227237-160227237 on Build GRCh38
rs1044391205 Chr.3: 160227467-160227467 on Build GRCh38
rs537554547 Chr.3: 160227347-160227347 on Build GRCh38
rs565298736 Chr.3: 160227432-160227434 on Build GRCh38
rs1023571849 Chr.3: 160227259-160227259 on Build GRCh38
rs551784002 Chr.3: 160227158-160227158 on Build GRCh38
rs779712226 Chr.3: 160227201-160227201 on Build GRCh38
rs1040783636 Chr.3: 160227459-160227459 on Build GRCh38
rs555579240 Chr.3: 160227328-160227328 on Build GRCh38
rs1035045577 Chr.3: 160227570-160227570 on Build GRCh38
rs955461138 Chr.3: 160227526-160227526 on Build GRCh38
rs1023876152 Chr.3: 160227539-160227539 on Build GRCh38
rs1022473000 Chr.3: 160227184-160227184 on Build GRCh38
rs900927928 Chr.3: 160227462-160227462 on Build GRCh38
rs1011179378 Chr.3: 160227455-160227455 on Build GRCh38
rs918445692 Chr.3: 160227349-160227349 on Build GRCh38
rs902754498 Chr.3: 160227470-160227470 on Build GRCh38
rs753338484 Chr.3: 160227329-160227329 on Build GRCh38
rs892776610 Chr.3: 160227432-160227432 on Build GRCh38
rs138522487 Chr.3: 160227153-160227154 on Build GRCh38
rs942410889 Chr.3: 160227124-160227124 on Build GRCh38
rs928412527 Chr.3: 160227379-160227379 on Build GRCh38
rs999679317 Chr.3: 160227263-160227263 on Build GRCh38
rs533842307 Chr.3: 160227298-160227298 on Build GRCh38
rs191975786 Chr.3: 160227391-160227391 on Build GRCh38
rs935482820 Chr.3: 160227472-160227472 on Build GRCh38
rs184227092 Chr.3: 160227546-160227546 on Build GRCh38
rs142639983 Chr.3: 160227340-160227340 on Build GRCh38
rs942379838 Chr.3: 160227444-160227444 on Build GRCh38
rs1000138680 Chr.3: 160227489-160227489 on Build GRCh38
rs1045561424 Chr.3: 160227357-160227357 on Build GRCh38
rs901335167 Chr.3: 160227140-160227140 on Build GRCh38
rs532718589 Chr.3: 160227356-160227356 on Build GRCh38

View Full Product Details