Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 160678198-160678729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL14
Entrez Gene ID
80117
Gene Name
ADP ribosylation factor like GTPase 14
Gene Aliases
ARF7
UniGene
Hs.287702
Gene Chromosome Location
Chr.3: 160677160-160678448 on Build GRCh38
Transcripts
NM_025047.2

SNP Detail

SNP ID Chromosome Location
rs565870078 Chr.3: 160678384-160678384 on Build GRCh38
rs75884210 Chr.3: 160678388-160678388 on Build GRCh38
rs963363874 Chr.3: 160678601-160678601 on Build GRCh38
rs1042896152 Chr.3: 160678567-160678567 on Build GRCh38
rs996606668 Chr.3: 160678334-160678334 on Build GRCh38
rs569318803 Chr.3: 160678387-160678387 on Build GRCh38
rs761815313 Chr.3: 160678287-160678287 on Build GRCh38
rs907679419 Chr.3: 160678269-160678269 on Build GRCh38
rs533133113 Chr.3: 160678440-160678440 on Build GRCh38
rs1052085914 Chr.3: 160678643-160678643 on Build GRCh38
rs973192101 Chr.3: 160678605-160678605 on Build GRCh38
rs557528609 Chr.3: 160678619-160678619 on Build GRCh38
rs4133252 Chr.3: 160678652-160678652 on Build GRCh38
rs757665619 Chr.3: 160678309-160678309 on Build GRCh38
rs567925919 Chr.3: 160678345-160678345 on Build GRCh38
rs890468402 Chr.3: 160678646-160678646 on Build GRCh38
rs555756004 Chr.3: 160678308-160678308 on Build GRCh38
rs913080199 Chr.3: 160678298-160678298 on Build GRCh38
rs938464570 Chr.3: 160678276-160678276 on Build GRCh38
rs74781289 Chr.3: 160678412-160678412 on Build GRCh38
rs368986985 Chr.3: 160678637-160678637 on Build GRCh38
rs988564020 Chr.3: 160678525-160678525 on Build GRCh38
rs779537169 Chr.3: 160678321-160678321 on Build GRCh38
rs568904894 Chr.3: 160678606-160678606 on Build GRCh38
rs537850282 Chr.3: 160678670-160678670 on Build GRCh38
rs759742202 Chr.3: 160678623-160678623 on Build GRCh38
rs890426332 Chr.3: 160678349-160678349 on Build GRCh38
rs969649482 Chr.3: 160678454-160678454 on Build GRCh38
rs928397729 Chr.3: 160678259-160678259 on Build GRCh38
rs1042343794 Chr.3: 160678399-160678399 on Build GRCh38
rs6806755 Chr.3: 160678553-160678553 on Build GRCh38
rs371655763 Chr.3: 160678247-160678247 on Build GRCh38
rs982828618 Chr.3: 160678594-160678594 on Build GRCh38
rs760346510 Chr.3: 160678510-160678510 on Build GRCh38
rs191982249 Chr.3: 160678333-160678333 on Build GRCh38
rs143305825 Chr.3: 160678491-160678491 on Build GRCh38
rs748919791 Chr.3: 160678325-160678325 on Build GRCh38
rs931934119 Chr.3: 160678617-160678617 on Build GRCh38
rs1015245472 Chr.3: 160678595-160678595 on Build GRCh38
rs770728459 Chr.3: 160678524-160678524 on Build GRCh38
rs555683813 Chr.3: 160678593-160678593 on Build GRCh38
rs551676857 Chr.3: 160678411-160678411 on Build GRCh38
rs921865312 Chr.3: 160678609-160678609 on Build GRCh38
rs1036444753 Chr.3: 160678690-160678690 on Build GRCh38
rs752707263 Chr.3: 160678694-160678694 on Build GRCh38
rs4133253 Chr.3: 160678671-160678671 on Build GRCh38
rs973264444 Chr.3: 160678324-160678324 on Build GRCh38
rs535367340 Chr.3: 160678554-160678554 on Build GRCh38

View Full Product Details