Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170892119-170892668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF5A2
Entrez Gene ID
56648
Gene Name
eukaryotic translation initiation factor 5A2
Gene Aliases
EIF-5A2, eIF5AII
UniGene
Hs.164144
Gene Chromosome Location
Chr.3: 170888415-170908637 on Build GRCh38
Transcripts
NM_020390.5

SNP Detail

SNP ID Chromosome Location
rs34403814 Chr.3: 170892448-170892448 on Build GRCh38
rs893996166 Chr.3: 170892505-170892505 on Build GRCh38
rs34539321 Chr.3: 170892464-170892464 on Build GRCh38
rs777701457 Chr.3: 170892556-170892556 on Build GRCh38
rs141092398 Chr.3: 170892304-170892304 on Build GRCh38
rs879168640 Chr.3: 170892622-170892622 on Build GRCh38
rs921093440 Chr.3: 170892148-170892148 on Build GRCh38
rs921052998 Chr.3: 170892246-170892246 on Build GRCh38
rs932520415 Chr.3: 170892153-170892153 on Build GRCh38
rs895488317 Chr.3: 170892255-170892255 on Build GRCh38
rs543865338 Chr.3: 170892395-170892395 on Build GRCh38
rs1384976 Chr.3: 170892444-170892444 on Build GRCh38
rs909601996 Chr.3: 170892583-170892583 on Build GRCh38
rs190193397 Chr.3: 170892554-170892554 on Build GRCh38
rs1384977 Chr.3: 170892406-170892406 on Build GRCh38
rs925884863 Chr.3: 170892188-170892188 on Build GRCh38
rs906025092 Chr.3: 170892290-170892290 on Build GRCh38
rs949736124 Chr.3: 170892524-170892524 on Build GRCh38
rs77597980 Chr.3: 170892453-170892453 on Build GRCh38
rs145468371 Chr.3: 170892254-170892254 on Build GRCh38
rs1019227263 Chr.3: 170892593-170892593 on Build GRCh38
rs1044209019 Chr.3: 170892282-170892282 on Build GRCh38
rs1037371711 Chr.3: 170892320-170892320 on Build GRCh38
rs116305447 Chr.3: 170892579-170892579 on Build GRCh38
rs1019329540 Chr.3: 170892387-170892387 on Build GRCh38
rs1002252958 Chr.3: 170892474-170892474 on Build GRCh38
rs996156267 Chr.3: 170892640-170892640 on Build GRCh38
rs530411894 Chr.3: 170892591-170892591 on Build GRCh38
rs905635807 Chr.3: 170892537-170892537 on Build GRCh38
rs934611430 Chr.3: 170892214-170892214 on Build GRCh38
rs61792989 Chr.3: 170892248-170892248 on Build GRCh38
rs769739793 Chr.3: 170892558-170892558 on Build GRCh38
rs190861407 Chr.3: 170892162-170892162 on Build GRCh38
rs1013972642 Chr.3: 170892258-170892258 on Build GRCh38
rs895722103 Chr.3: 170892418-170892419 on Build GRCh38
rs6792592 Chr.3: 170892470-170892470 on Build GRCh38
rs965653977 Chr.3: 170892544-170892544 on Build GRCh38
rs922047386 Chr.3: 170892416-170892416 on Build GRCh38
rs966004098 Chr.3: 170892602-170892602 on Build GRCh38
rs10581157 Chr.3: 170892638-170892639 on Build GRCh38
rs976425480 Chr.3: 170892194-170892194 on Build GRCh38
rs538565044 Chr.3: 170892319-170892319 on Build GRCh38
rs974790318 Chr.3: 170892411-170892411 on Build GRCh38
rs188044242 Chr.3: 170892354-170892354 on Build GRCh38
rs6779919 Chr.3: 170892484-170892484 on Build GRCh38
rs555889684 Chr.3: 170892462-170892462 on Build GRCh38
rs994608195 Chr.3: 170892553-170892553 on Build GRCh38
rs983836407 Chr.3: 170892249-170892249 on Build GRCh38
rs966020442 Chr.3: 170892402-170892402 on Build GRCh38
rs182579351 Chr.3: 170892310-170892310 on Build GRCh38
rs6792607 Chr.3: 170892527-170892527 on Build GRCh38
rs771306682 Chr.3: 170892635-170892635 on Build GRCh38
rs576526348 Chr.3: 170892377-170892377 on Build GRCh38
rs965445622 Chr.3: 170892164-170892164 on Build GRCh38
rs534804933 Chr.3: 170892286-170892286 on Build GRCh38
rs939879333 Chr.3: 170892584-170892584 on Build GRCh38
rs1005148003 Chr.3: 170892356-170892356 on Build GRCh38
rs916901410 Chr.3: 170892511-170892511 on Build GRCh38
rs953861340 Chr.3: 170892419-170892419 on Build GRCh38

View Full Product Details