Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 172133361-172133760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC3B
Entrez Gene ID
64778
Gene Name
fibronectin type III domain containing 3B
Gene Aliases
FAD104, PRO4979, YVTM2421
UniGene
Hs.744888
Gene Chromosome Location
Chr.3: 172039628-172400703 on Build GRCh38
Transcripts
NM_001135095.1 NM_022763.3 XM_017007062.1 XM_017007064.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5006960 c.133C>G p.P45A Chr.3: 172133492-172133492 on Build GRCh38
4054980 c.187+5G>A p.? Chr.3: 172133551-172133551 on Build GRCh38
729083 c.160G>T p.E54* Chr.3: 172133519-172133519 on Build GRCh38
4954768 c.150A>G p.T50T Chr.3: 172133509-172133509 on Build GRCh38
1158420 c.121G>A p.V41I Chr.3: 172133480-172133480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367594838 Chr.3: 172133689-172133689 on Build GRCh38
rs530804931 Chr.3: 172133672-172133672 on Build GRCh38
rs569350066 Chr.3: 172133610-172133610 on Build GRCh38
rs528420349 Chr.3: 172133408-172133408 on Build GRCh38
rs747419375 Chr.3: 172133398-172133398 on Build GRCh38
rs935917967 Chr.3: 172133581-172133582 on Build GRCh38
rs563751151 Chr.3: 172133600-172133600 on Build GRCh38
rs756157879 Chr.3: 172133540-172133540 on Build GRCh38
rs759960000 Chr.3: 172133673-172133673 on Build GRCh38
rs769023379 Chr.3: 172133459-172133459 on Build GRCh38
rs761382945 Chr.3: 172133456-172133456 on Build GRCh38
rs987777089 Chr.3: 172133691-172133691 on Build GRCh38
rs150488615 Chr.3: 172133480-172133480 on Build GRCh38
rs762560445 Chr.3: 172133629-172133629 on Build GRCh38
rs779013733 Chr.3: 172133453-172133453 on Build GRCh38
rs370524615 Chr.3: 172133632-172133632 on Build GRCh38
rs766795993 Chr.3: 172133630-172133630 on Build GRCh38
rs201913233 Chr.3: 172133597-172133597 on Build GRCh38
rs980091105 Chr.3: 172133449-172133449 on Build GRCh38
rs763501921 Chr.3: 172133479-172133479 on Build GRCh38
rs758274956 Chr.3: 172133577-172133577 on Build GRCh38
rs566465641 Chr.3: 172133701-172133701 on Build GRCh38
rs756280555 Chr.3: 172133652-172133652 on Build GRCh38
rs926506908 Chr.3: 172133732-172133732 on Build GRCh38
rs767966179 Chr.3: 172133639-172133639 on Build GRCh38
rs532649939 Chr.3: 172133590-172133590 on Build GRCh38
rs763580982 Chr.3: 172133661-172133662 on Build GRCh38
rs374846620 Chr.3: 172133650-172133650 on Build GRCh38
rs772787716 Chr.3: 172133624-172133624 on Build GRCh38
rs768189211 Chr.3: 172133439-172133439 on Build GRCh38
rs917318184 Chr.3: 172133409-172133409 on Build GRCh38
rs111245223 Chr.3: 172133528-172133528 on Build GRCh38
rs192019662 Chr.3: 172133708-172133708 on Build GRCh38
rs754900301 Chr.3: 172133692-172133692 on Build GRCh38
rs553099022 Chr.3: 172133581-172133581 on Build GRCh38
rs780989362 Chr.3: 172133697-172133697 on Build GRCh38
rs777791357 Chr.3: 172133570-172133570 on Build GRCh38
rs749393147 Chr.3: 172133576-172133576 on Build GRCh38
rs752013295 Chr.3: 172133672-172133673 on Build GRCh38
rs888847520 Chr.3: 172133607-172133607 on Build GRCh38
rs764315016 Chr.3: 172133658-172133658 on Build GRCh38
rs747760320 Chr.3: 172133616-172133616 on Build GRCh38
rs769588621 Chr.3: 172133619-172133619 on Build GRCh38
rs751808398 Chr.3: 172133499-172133499 on Build GRCh38
rs116529569 Chr.3: 172133454-172133454 on Build GRCh38
rs764877604 Chr.3: 172133458-172133458 on Build GRCh38
rs188271722 Chr.3: 172133687-172133687 on Build GRCh38
rs961864646 Chr.3: 172133731-172133731 on Build GRCh38
rs780104558 Chr.3: 172133586-172133586 on Build GRCh38
rs972828818 Chr.3: 172133404-172133404 on Build GRCh38
rs138140214 Chr.3: 172133681-172133681 on Build GRCh38
rs866383009 Chr.3: 172133706-172133706 on Build GRCh38
rs901593242 Chr.3: 172133674-172133674 on Build GRCh38
rs766957338 Chr.3: 172133665-172133666 on Build GRCh38
rs767624523 Chr.3: 172133536-172133536 on Build GRCh38
rs755441800 Chr.3: 172133501-172133501 on Build GRCh38

View Full Product Details