Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 172754578-172755090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECT2
Entrez Gene ID
1894
Gene Name
epithelial cell transforming 2
Gene Aliases
ARHGEF31
UniGene
Hs.518299
Gene Chromosome Location
Chr.3: 172750682-172829273 on Build GRCh38
Transcripts
NM_001258315.1 NM_001258316.1 NM_018098.5 XM_005247176.1 XM_006713523.3 XM_006713524.3 XM_011512514.2 XM_011512515.2 XM_011512516.1 XM_017005830.1 XM_017005831.1 XM_017005832.1 XM_017005833.1 XM_017005834.1 XM_017005835.1 XM_017005836.1 XM_017005837.1 XM_017005838.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5398673 c.89C>T p.S30F Chr.3: 172754619-172754619 on Build GRCh38
3735642 c.71C>T p.S24F Chr.3: 172754601-172754601 on Build GRCh38
4841215 c.84G>T p.E28D Chr.3: 172754614-172754614 on Build GRCh38
3660349 c.106A>T p.I36F Chr.3: 172754636-172754636 on Build GRCh38
3590297 c.123T>A p.Y41* Chr.3: 172754653-172754653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906484567 Chr.3: 172754843-172754843 on Build GRCh38
rs183566950 Chr.3: 172754737-172754737 on Build GRCh38
rs748486976 Chr.3: 172754637-172754637 on Build GRCh38
rs766590049 Chr.3: 172754685-172754688 on Build GRCh38
rs981941182 Chr.3: 172754945-172754945 on Build GRCh38
rs545408401 Chr.3: 172754972-172754972 on Build GRCh38
rs776838286 Chr.3: 172754919-172754919 on Build GRCh38
rs897970356 Chr.3: 172754728-172754728 on Build GRCh38
rs1032311889 Chr.3: 172754849-172754849 on Build GRCh38
rs199950814 Chr.3: 172754714-172754714 on Build GRCh38
rs773482993 Chr.3: 172754657-172754657 on Build GRCh38
rs555919529 Chr.3: 172754717-172754717 on Build GRCh38
rs140381162 Chr.3: 172754656-172754656 on Build GRCh38
rs150569451 Chr.3: 172754634-172754634 on Build GRCh38
rs1020812998 Chr.3: 172754939-172754939 on Build GRCh38
rs575276736 Chr.3: 172754706-172754706 on Build GRCh38
rs941808437 Chr.3: 172754766-172754766 on Build GRCh38
rs1031013098 Chr.3: 172754807-172754807 on Build GRCh38
rs935221725 Chr.3: 172754863-172754863 on Build GRCh38
rs757872372 Chr.3: 172754611-172754611 on Build GRCh38
rs373132353 Chr.3: 172754633-172754633 on Build GRCh38
rs774530249 Chr.3: 172754669-172754669 on Build GRCh38
rs759691315 Chr.3: 172754672-172754672 on Build GRCh38
rs889350488 Chr.3: 172754834-172754834 on Build GRCh38
rs962822916 Chr.3: 172755030-172755030 on Build GRCh38
rs750066658 Chr.3: 172754691-172754691 on Build GRCh38
rs764030791 Chr.3: 172754673-172754673 on Build GRCh38
rs757026653 Chr.3: 172755044-172755044 on Build GRCh38
rs1007569594 Chr.3: 172754841-172754841 on Build GRCh38
rs982979432 Chr.3: 172755027-172755027 on Build GRCh38
rs766547704 Chr.3: 172754664-172754664 on Build GRCh38
rs560062224 Chr.3: 172754768-172754768 on Build GRCh38
rs1052289633 Chr.3: 172754872-172754872 on Build GRCh38
rs546574061 Chr.3: 172755028-172755028 on Build GRCh38
rs549165637 Chr.3: 172755053-172755053 on Build GRCh38
rs1011006053 Chr.3: 172754889-172754889 on Build GRCh38
rs958144280 Chr.3: 172754875-172754875 on Build GRCh38
rs763309386 Chr.3: 172754661-172754661 on Build GRCh38
rs973104527 Chr.3: 172754798-172754798 on Build GRCh38
rs746543430 Chr.3: 172754628-172754628 on Build GRCh38
rs770114928 Chr.3: 172754652-172754652 on Build GRCh38
rs142626979 Chr.3: 172754610-172754610 on Build GRCh38
rs1046484493 Chr.3: 172754821-172754821 on Build GRCh38
rs74697599 Chr.3: 172754941-172754941 on Build GRCh38
rs988611866 Chr.3: 172754907-172754907 on Build GRCh38
rs1030071256 Chr.3: 172754966-172754966 on Build GRCh38
rs757070002 Chr.3: 172754675-172754675 on Build GRCh38
rs764980892 Chr.3: 172754690-172754690 on Build GRCh38
rs780165590 Chr.3: 172754951-172754953 on Build GRCh38
rs186775483 Chr.3: 172754749-172754749 on Build GRCh38
rs937896163 Chr.3: 172754973-172754973 on Build GRCh38
rs753774488 Chr.3: 172754674-172754674 on Build GRCh38
rs768907305 Chr.3: 172754833-172754833 on Build GRCh38
rs566838050 Chr.3: 172755048-172755048 on Build GRCh38
rs551013726 Chr.3: 172754920-172754920 on Build GRCh38
rs947707691 Chr.3: 172754819-172754819 on Build GRCh38
rs536677886 Chr.3: 172754983-172754983 on Build GRCh38
rs753603847 Chr.3: 172755036-172755036 on Build GRCh38
rs760949053 Chr.3: 172754810-172754810 on Build GRCh38
rs764318469 Chr.3: 172754731-172754731 on Build GRCh38
rs1018327558 Chr.3: 172754908-172754908 on Build GRCh38
rs779586664 Chr.3: 172754617-172754617 on Build GRCh38

View Full Product Details