Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 172762524-172763023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECT2
Entrez Gene ID
1894
Gene Name
epithelial cell transforming 2
Gene Aliases
ARHGEF31
UniGene
Hs.518299
Gene Chromosome Location
Chr.3: 172750682-172829273 on Build GRCh38
Transcripts
NM_001258315.1 NM_001258316.1 NM_018098.5 XM_005247176.1 XM_006713523.3 XM_006713524.3 XM_011512514.2 XM_011512515.2 XM_011512516.1 XM_017005830.1 XM_017005831.1 XM_017005832.1 XM_017005833.1 XM_017005834.1 XM_017005835.1 XM_017005836.1 XM_017005837.1 XM_017005838.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680115 c.806A>G p.Y269C Chr.3: 172762700-172762700 on Build GRCh38
3205123 c.803A>C p.K268T Chr.3: 172762697-172762697 on Build GRCh38
1637973 c.943C>T p.R315* Chr.3: 172762940-172762940 on Build GRCh38
6453840 c.905C>T p.P302L Chr.3: 172762706-172762706 on Build GRCh38
4944755 c.823G>A p.E275K Chr.3: 172762717-172762717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900671938 Chr.3: 172762634-172762634 on Build GRCh38
rs775334397 Chr.3: 172762995-172762995 on Build GRCh38
rs150790038 Chr.3: 172762776-172762776 on Build GRCh38
rs758384949 Chr.3: 172762761-172762761 on Build GRCh38
rs747123435 Chr.3: 172762773-172762773 on Build GRCh38
rs1039879182 Chr.3: 172762847-172762847 on Build GRCh38
rs760287034 Chr.3: 172762875-172762875 on Build GRCh38
rs757484043 Chr.3: 172762591-172762591 on Build GRCh38
rs754531638 Chr.3: 172762555-172762555 on Build GRCh38
rs750969640 Chr.3: 172762549-172762549 on Build GRCh38
rs368163367 Chr.3: 172762746-172762746 on Build GRCh38
rs374691305 Chr.3: 172762955-172762955 on Build GRCh38
rs918533119 Chr.3: 172762780-172762780 on Build GRCh38
rs370177109 Chr.3: 172762570-172762570 on Build GRCh38
rs749311125 Chr.3: 172762965-172762965 on Build GRCh38
rs773544097 Chr.3: 172762587-172762587 on Build GRCh38
rs932569715 Chr.3: 172762724-172762724 on Build GRCh38
rs767688416 Chr.3: 172762757-172762758 on Build GRCh38
rs185535917 Chr.3: 172762732-172762732 on Build GRCh38
rs375708654 Chr.3: 172762714-172762714 on Build GRCh38
rs1011058816 Chr.3: 172762924-172762924 on Build GRCh38
rs1015823294 Chr.3: 172762774-172762774 on Build GRCh38
rs960855534 Chr.3: 172762725-172762725 on Build GRCh38
rs141100382 Chr.3: 172762846-172762846 on Build GRCh38
rs769746235 Chr.3: 172762668-172762668 on Build GRCh38
rs772259525 Chr.3: 172762597-172762597 on Build GRCh38
rs775104619 Chr.3: 172762857-172762857 on Build GRCh38
rs980097774 Chr.3: 172762905-172762905 on Build GRCh38
rs765891768 Chr.3: 172762705-172762705 on Build GRCh38
rs991318628 Chr.3: 172762731-172762731 on Build GRCh38
rs759926948 Chr.3: 172762838-172762838 on Build GRCh38
rs978086847 Chr.3: 172762674-172762674 on Build GRCh38
rs890796274 Chr.3: 172762836-172762836 on Build GRCh38
rs774113446 Chr.3: 172762706-172762706 on Build GRCh38
rs763067010 Chr.3: 172762647-172762647 on Build GRCh38
rs1035981302 Chr.3: 172762934-172762934 on Build GRCh38
rs371741696 Chr.3: 172762883-172762883 on Build GRCh38
rs200063253 Chr.3: 172762975-172762975 on Build GRCh38
rs377204259 Chr.3: 172762977-172762977 on Build GRCh38
rs750766918 Chr.3: 172762938-172762938 on Build GRCh38
rs980316456 Chr.3: 172762626-172762626 on Build GRCh38
rs745685915 Chr.3: 172762986-172762986 on Build GRCh38
rs181315491 Chr.3: 172762589-172762589 on Build GRCh38
rs200765639 Chr.3: 172762828-172762828 on Build GRCh38
rs369898272 Chr.3: 172762994-172762994 on Build GRCh38
rs745920922 Chr.3: 172762594-172762594 on Build GRCh38
rs989378584 Chr.3: 172762639-172762639 on Build GRCh38
rs770797910 Chr.3: 172762827-172762827 on Build GRCh38
rs780284987 Chr.3: 172762941-172762941 on Build GRCh38
rs759287302 Chr.3: 172762837-172762837 on Build GRCh38
rs778996603 Chr.3: 172762752-172762752 on Build GRCh38
rs762742246 Chr.3: 172762697-172762697 on Build GRCh38
rs780132969 Chr.3: 172762768-172762768 on Build GRCh38
rs138074177 Chr.3: 172762790-172762790 on Build GRCh38
rs777991375 Chr.3: 172762940-172762940 on Build GRCh38
rs371845005 Chr.3: 172762751-172762751 on Build GRCh38
rs754193804 Chr.3: 172762744-172762744 on Build GRCh38
rs995967083 Chr.3: 172762815-172762815 on Build GRCh38
rs970008086 Chr.3: 172762881-172762881 on Build GRCh38
rs373313106 Chr.3: 172762996-172762996 on Build GRCh38
rs1017991574 Chr.3: 172762633-172762633 on Build GRCh38
rs755097248 Chr.3: 172762961-172762961 on Build GRCh38
rs767899343 Chr.3: 172762560-172762560 on Build GRCh38
rs772951610 Chr.3: 172762673-172762673 on Build GRCh38
rs970312294 Chr.3: 172762615-172762615 on Build GRCh38
rs141974807 Chr.3: 172762756-172762756 on Build GRCh38
rs1050733701 Chr.3: 172762585-172762585 on Build GRCh38
rs1050784769 Chr.3: 172762835-172762835 on Build GRCh38
rs756644935 Chr.3: 172762719-172762719 on Build GRCh38
rs80293636 Chr.3: 172762663-172762663 on Build GRCh38
rs756520153 Chr.3: 172762579-172762579 on Build GRCh38
rs762081406 Chr.3: 172762921-172762921 on Build GRCh38
rs1008089831 Chr.3: 172762750-172762750 on Build GRCh38
rs749057791 Chr.3: 172762824-172762824 on Build GRCh38
rs145065176 Chr.3: 172762915-172762915 on Build GRCh38
rs547764309 Chr.3: 172762930-172762930 on Build GRCh38
rs34623300 Chr.3: 172762707-172762707 on Build GRCh38
rs374516578 Chr.3: 172762593-172762593 on Build GRCh38
rs778707047 Chr.3: 172762976-172762976 on Build GRCh38
rs759733611 Chr.3: 172762848-172762849 on Build GRCh38
rs753237241 Chr.3: 172762715-172762715 on Build GRCh38
rs777654187 Chr.3: 172762963-172762963 on Build GRCh38
rs1007045477 Chr.3: 172762603-172762603 on Build GRCh38
rs35786384 Chr.3: 172762688-172762688 on Build GRCh38

View Full Product Details