Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 180951228-180951727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXR1
Entrez Gene ID
8087
Gene Name
FMR1 autosomal homolog 1
Gene Aliases
FXR1P
UniGene
Hs.478407
Gene Chromosome Location
Chr.3: 180912446-180982753 on Build GRCh38
Transcripts
NM_001013438.2 NM_001013439.2 NM_005087.3 XM_005247813.3 XM_005247814.3 XM_005247815.3 XM_005247816.3 XM_006713775.3 XM_017007298.1 XM_017007299.1 XM_017007300.1 XM_017007301.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
446043 c.786C>A p.F262L Chr.3: 180951453-180951453 on Build GRCh38
6667965 c.682G>T p.D228Y Chr.3: 180951349-180951349 on Build GRCh38
1726507 c.692G>A p.G231D Chr.3: 180951359-180951359 on Build GRCh38
5863985 c.760G>T p.E254* Chr.3: 180951427-180951427 on Build GRCh38
4685912 c.686T>G p.L229* Chr.3: 180951353-180951353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551449074 Chr.3: 180951528-180951528 on Build GRCh38
rs767215804 Chr.3: 180951516-180951516 on Build GRCh38
rs953480919 Chr.3: 180951594-180951594 on Build GRCh38
rs374396414 Chr.3: 180951351-180951351 on Build GRCh38
rs766297434 Chr.3: 180951354-180951354 on Build GRCh38
rs759645322 Chr.3: 180951276-180951276 on Build GRCh38
rs373538881 Chr.3: 180951515-180951515 on Build GRCh38
rs940649917 Chr.3: 180951603-180951603 on Build GRCh38
rs764264412 Chr.3: 180951325-180951325 on Build GRCh38
rs184718040 Chr.3: 180951600-180951600 on Build GRCh38
rs962098022 Chr.3: 180951549-180951549 on Build GRCh38
rs933159953 Chr.3: 180951638-180951638 on Build GRCh38
rs755691527 Chr.3: 180951453-180951453 on Build GRCh38
rs73176330 Chr.3: 180951420-180951420 on Build GRCh38
rs376533605 Chr.3: 180951518-180951518 on Build GRCh38
rs984903481 Chr.3: 180951596-180951596 on Build GRCh38
rs760912085 Chr.3: 180951492-180951492 on Build GRCh38
rs759700778 Chr.3: 180951263-180951263 on Build GRCh38
rs754313359 Chr.3: 180951644-180951644 on Build GRCh38
rs371035174 Chr.3: 180951484-180951484 on Build GRCh38
rs200507471 Chr.3: 180951448-180951448 on Build GRCh38
rs767740881 Chr.3: 180951269-180951269 on Build GRCh38
rs918479725 Chr.3: 180951697-180951697 on Build GRCh38
rs376260999 Chr.3: 180951508-180951508 on Build GRCh38
rs1003487718 Chr.3: 180951343-180951343 on Build GRCh38
rs751427528 Chr.3: 180951509-180951509 on Build GRCh38
rs972124243 Chr.3: 180951563-180951563 on Build GRCh38
rs760887774 Chr.3: 180951282-180951282 on Build GRCh38
rs751326790 Chr.3: 180951378-180951378 on Build GRCh38
rs150549760 Chr.3: 180951584-180951584 on Build GRCh38
rs1003351615 Chr.3: 180951267-180951267 on Build GRCh38
rs200414784 Chr.3: 180951279-180951279 on Build GRCh38
rs559357948 Chr.3: 180951690-180951690 on Build GRCh38
rs763011616 Chr.3: 180951500-180951500 on Build GRCh38
rs779178820 Chr.3: 180951474-180951474 on Build GRCh38
rs536907868 Chr.3: 180951579-180951579 on Build GRCh38
rs541044881 Chr.3: 180951632-180951632 on Build GRCh38
rs750391093 Chr.3: 180951330-180951330 on Build GRCh38
rs747745510 Chr.3: 180951449-180951449 on Build GRCh38
rs142641159 Chr.3: 180951454-180951454 on Build GRCh38
rs199498739 Chr.3: 180951476-180951476 on Build GRCh38
rs749812307 Chr.3: 180951461-180951461 on Build GRCh38
rs1021907404 Chr.3: 180951441-180951441 on Build GRCh38
rs776802797 Chr.3: 180951498-180951498 on Build GRCh38
rs774723399 Chr.3: 180951258-180951258 on Build GRCh38
rs892189752 Chr.3: 180951260-180951260 on Build GRCh38
rs771379697 Chr.3: 180951462-180951462 on Build GRCh38
rs780942609 Chr.3: 180951432-180951432 on Build GRCh38
rs768870495 Chr.3: 180951496-180951496 on Build GRCh38
rs199617499 Chr.3: 180951486-180951486 on Build GRCh38

View Full Product Details