Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 180957695-180958219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXR1
Entrez Gene ID
8087
Gene Name
FMR1 autosomal homolog 1
Gene Aliases
FXR1P
UniGene
Hs.478407
Gene Chromosome Location
Chr.3: 180912446-180982753 on Build GRCh38
Transcripts
NM_001013438.2 NM_001013439.2 NM_005087.3 XM_005247813.3 XM_005247814.3 XM_005247815.3 XM_005247816.3 XM_006713775.3 XM_017007298.1 XM_017007299.1 XM_017007300.1 XM_017007301.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1207673 c.943C>T p.R315* Chr.3: 180957881-180957881 on Build GRCh38
1566545 c.885A>G p.K295K Chr.3: 180957823-180957823 on Build GRCh38
1041697 c.990+1G>A p.? Chr.3: 180957929-180957929 on Build GRCh38
6163828 c.948_949GA>TT p.R317* Chr.3: 180957886-180957887 on Build GRCh38
4513282 c.932C>A p.S311Y Chr.3: 180957870-180957870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941156596 Chr.3: 180957734-180957734 on Build GRCh38
rs374344744 Chr.3: 180957942-180957942 on Build GRCh38
rs538850059 Chr.3: 180957718-180957718 on Build GRCh38
rs776340627 Chr.3: 180958072-180958072 on Build GRCh38
rs939137135 Chr.3: 180957757-180957757 on Build GRCh38
rs532505719 Chr.3: 180958060-180958060 on Build GRCh38
rs984816521 Chr.3: 180958046-180958046 on Build GRCh38
rs779695858 Chr.3: 180957775-180957797 on Build GRCh38
rs576038253 Chr.3: 180957980-180957980 on Build GRCh38
rs772508533 Chr.3: 180957743-180957743 on Build GRCh38
rs771204399 Chr.3: 180957939-180957940 on Build GRCh38
rs775514702 Chr.3: 180957959-180957960 on Build GRCh38
rs1056089590 Chr.3: 180958181-180958181 on Build GRCh38
rs907460146 Chr.3: 180957767-180957767 on Build GRCh38
rs745844006 Chr.3: 180957898-180957898 on Build GRCh38
rs199859306 Chr.3: 180957793-180957793 on Build GRCh38
rs768063286 Chr.3: 180957954-180957955 on Build GRCh38
rs776449660 Chr.3: 180957932-180957933 on Build GRCh38
rs909751241 Chr.3: 180958079-180958079 on Build GRCh38
rs1042323195 Chr.3: 180957754-180957754 on Build GRCh38
rs976194083 Chr.3: 180958004-180958004 on Build GRCh38
rs946172986 Chr.3: 180958056-180958056 on Build GRCh38
rs769925166 Chr.3: 180957841-180957841 on Build GRCh38
rs760774217 Chr.3: 180957961-180957962 on Build GRCh38
rs553965452 Chr.3: 180957759-180957759 on Build GRCh38
rs966110541 Chr.3: 180958002-180958002 on Build GRCh38
rs773110019 Chr.3: 180957870-180957870 on Build GRCh38
rs1011934094 Chr.3: 180957950-180957950 on Build GRCh38
rs757894126 Chr.3: 180957954-180957954 on Build GRCh38
rs1031208908 Chr.3: 180957997-180957997 on Build GRCh38
rs904114772 Chr.3: 180957969-180957969 on Build GRCh38
rs774846301 Chr.3: 180957952-180957953 on Build GRCh38
rs761312916 Chr.3: 180957949-180957949 on Build GRCh38
rs76978300 Chr.3: 180958017-180958017 on Build GRCh38
rs985735238 Chr.3: 180957995-180957995 on Build GRCh38
rs750952239 Chr.3: 180957784-180957784 on Build GRCh38
rs748373761 Chr.3: 180957826-180957826 on Build GRCh38
rs755368035 Chr.3: 180957798-180957798 on Build GRCh38
rs775375701 Chr.3: 180957917-180957917 on Build GRCh38
rs767039013 Chr.3: 180957956-180957956 on Build GRCh38
rs372060185 Chr.3: 180957976-180957976 on Build GRCh38
rs755457990 Chr.3: 180957962-180957962 on Build GRCh38
rs181799042 Chr.3: 180958031-180958031 on Build GRCh38
rs569904802 Chr.3: 180957943-180957943 on Build GRCh38
rs768386596 Chr.3: 180957931-180957932 on Build GRCh38
rs939083461 Chr.3: 180958169-180958169 on Build GRCh38
rs549292600 Chr.3: 180958165-180958165 on Build GRCh38
rs763388651 Chr.3: 180958084-180958084 on Build GRCh38
rs759948184 Chr.3: 180957953-180957954 on Build GRCh38
rs749861166 Chr.3: 180957953-180957953 on Build GRCh38
rs746560734 Chr.3: 180957928-180957929 on Build GRCh38
rs972967976 Chr.3: 180958108-180958108 on Build GRCh38
rs918446788 Chr.3: 180958114-180958114 on Build GRCh38
rs530320996 Chr.3: 180958180-180958180 on Build GRCh38
rs376336776 Chr.3: 180957808-180957808 on Build GRCh38
rs760512627 Chr.3: 180957928-180957928 on Build GRCh38
rs765698766 Chr.3: 180958000-180958000 on Build GRCh38
rs368200929 Chr.3: 180957951-180957951 on Build GRCh38
rs958720175 Chr.3: 180957892-180957892 on Build GRCh38
rs752094655 Chr.3: 180957960-180957960 on Build GRCh38
rs747415158 Chr.3: 180957794-180957794 on Build GRCh38
rs894473677 Chr.3: 180958185-180958185 on Build GRCh38
rs907433118 Chr.3: 180958119-180958119 on Build GRCh38
rs771868461 Chr.3: 180957913-180957913 on Build GRCh38
rs184217446 Chr.3: 180958092-180958092 on Build GRCh38
rs1056042499 Chr.3: 180957772-180957772 on Build GRCh38
rs923145232 Chr.3: 180957968-180957968 on Build GRCh38
rs76531300 Chr.3: 180957830-180957830 on Build GRCh38
rs896904456 Chr.3: 180957744-180957744 on Build GRCh38
rs559943175 Chr.3: 180958097-180958097 on Build GRCh38
rs190435322 Chr.3: 180957964-180957964 on Build GRCh38
rs3026197 Chr.3: 180957731-180957731 on Build GRCh38
rs754019564 Chr.3: 180957967-180957968 on Build GRCh38
rs779671749 Chr.3: 180958087-180958088 on Build GRCh38
rs200843705 Chr.3: 180957792-180957792 on Build GRCh38
rs749612035 Chr.3: 180957938-180957939 on Build GRCh38
rs750039510 Chr.3: 180958029-180958031 on Build GRCh38

View Full Product Details