Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 182845347-182845853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11B
Entrez Gene ID
23200
Gene Name
ATPase phospholipid transporting 11B (putative)
Gene Aliases
ATPIF, ATPIR
UniGene
Hs.478429
Gene Chromosome Location
Chr.3: 182793503-182923010 on Build GRCh38
Transcripts
NM_014616.2 XM_005247241.2 XM_005247243.2 XM_011512593.2 XM_011512594.2 XM_011512597.2 XM_017006008.1 XM_017006009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1041713 c.760G>T p.E254* Chr.3: 182845513-182845513 on Build GRCh38
4973224 c.734G>A p.R245H Chr.3: 182845487-182845487 on Build GRCh38
3915429 c.707C>T p.P236L Chr.3: 182845460-182845460 on Build GRCh38
6351559 c.716C>T p.P239L Chr.3: 182845469-182845469 on Build GRCh38
3365086 c.748A>G p.K250E Chr.3: 182845501-182845501 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375235909 Chr.3: 182845528-182845528 on Build GRCh38
rs767143988 Chr.3: 182845574-182845574 on Build GRCh38
rs749328307 Chr.3: 182845585-182845585 on Build GRCh38
rs766248151 Chr.3: 182845534-182845534 on Build GRCh38
rs112781753 Chr.3: 182845618-182845618 on Build GRCh38
rs964571109 Chr.3: 182845654-182845654 on Build GRCh38
rs995672011 Chr.3: 182845828-182845828 on Build GRCh38
rs142938523 Chr.3: 182845470-182845470 on Build GRCh38
rs751362703 Chr.3: 182845552-182845552 on Build GRCh38
rs55668228 Chr.3: 182845759-182845759 on Build GRCh38
rs760746973 Chr.3: 182845511-182845511 on Build GRCh38
rs760788784 Chr.3: 182845758-182845758 on Build GRCh38
rs761051610 Chr.3: 182845446-182845447 on Build GRCh38
rs1010987677 Chr.3: 182845550-182845550 on Build GRCh38
rs975517784 Chr.3: 182845700-182845700 on Build GRCh38
rs1004495345 Chr.3: 182845409-182845409 on Build GRCh38
rs914645929 Chr.3: 182845717-182845717 on Build GRCh38
rs759288667 Chr.3: 182845567-182845567 on Build GRCh38
rs368559733 Chr.3: 182845576-182845576 on Build GRCh38
rs60524783 Chr.3: 182845736-182845736 on Build GRCh38
rs999237620 Chr.3: 182845390-182845390 on Build GRCh38
rs75637615 Chr.3: 182845516-182845516 on Build GRCh38
rs907554360 Chr.3: 182845742-182845742 on Build GRCh38
rs182771870 Chr.3: 182845797-182845797 on Build GRCh38
rs879376795 Chr.3: 182845807-182845807 on Build GRCh38
rs906757720 Chr.3: 182845455-182845455 on Build GRCh38
rs572600718 Chr.3: 182845662-182845662 on Build GRCh38
rs902418981 Chr.3: 182845384-182845384 on Build GRCh38
rs762897338 Chr.3: 182845530-182845530 on Build GRCh38
rs756592871 Chr.3: 182845437-182845437 on Build GRCh38
rs766332860 Chr.3: 182845541-182845541 on Build GRCh38
rs771024083 Chr.3: 182845589-182845589 on Build GRCh38
rs555324755 Chr.3: 182845722-182845722 on Build GRCh38
rs1025930037 Chr.3: 182845611-182845614 on Build GRCh38
rs756267967 Chr.3: 182845466-182845466 on Build GRCh38
rs866508705 Chr.3: 182845791-182845791 on Build GRCh38
rs920404125 Chr.3: 182845720-182845720 on Build GRCh38
rs746058315 Chr.3: 182845471-182845471 on Build GRCh38
rs545085939 Chr.3: 182845826-182845826 on Build GRCh38
rs771273511 Chr.3: 182845469-182845469 on Build GRCh38
rs550157097 Chr.3: 182845454-182845454 on Build GRCh38
rs754577880 Chr.3: 182845408-182845408 on Build GRCh38
rs775454339 Chr.3: 182845494-182845494 on Build GRCh38
rs749720577 Chr.3: 182845468-182845468 on Build GRCh38
rs747650546 Chr.3: 182845436-182845436 on Build GRCh38
rs974200035 Chr.3: 182845596-182845596 on Build GRCh38
rs1023779610 Chr.3: 182845556-182845556 on Build GRCh38
rs774218790 Chr.3: 182845676-182845676 on Build GRCh38
rs906790402 Chr.3: 182845787-182845787 on Build GRCh38
rs576838519 Chr.3: 182845723-182845723 on Build GRCh38
rs36041795 Chr.3: 182845757-182845757 on Build GRCh38
rs35039982 Chr.3: 182845725-182845725 on Build GRCh38
rs570990408 Chr.3: 182845395-182845395 on Build GRCh38
rs780602611 Chr.3: 182845428-182845428 on Build GRCh38
rs192662576 Chr.3: 182845680-182845680 on Build GRCh38
rs772241980 Chr.3: 182845487-182845487 on Build GRCh38
rs141764997 Chr.3: 182845811-182845811 on Build GRCh38
rs775130833 Chr.3: 182845786-182845786 on Build GRCh38

View Full Product Details