Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 182944950-182945463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCUN1D1
Entrez Gene ID
54165
Gene Name
defective in cullin neddylation 1 domain containing 1
Gene Aliases
DCNL1, DCUN1L1, RP42, SCCRO, SCRO, Tes3
UniGene
Hs.592789
Gene Chromosome Location
Chr.3: 182938158-182986364 on Build GRCh38
Transcripts
NM_001308101.1 NM_020640.3 XM_011512912.1 XM_011512914.1 XM_011512915.2 XM_011512916.2 XM_017006633.1 XM_017006634.1 XM_017006635.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5498871 c.742C>T p.R248C Chr.3: 182945132-182945132 on Build GRCh38
4395407 c.744C>T p.R248R Chr.3: 182945130-182945130 on Build GRCh38
1633006 c.733G>T p.E245* Chr.3: 182945141-182945141 on Build GRCh38
4115557 c.743G>A p.R248H Chr.3: 182945131-182945131 on Build GRCh38
6968649 c.708G>T p.W236C Chr.3: 182945166-182945166 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755135817 Chr.3: 182945107-182945107 on Build GRCh38
rs533434313 Chr.3: 182944996-182944996 on Build GRCh38
rs1054615430 Chr.3: 182945211-182945211 on Build GRCh38
rs765815861 Chr.3: 182945212-182945212 on Build GRCh38
rs539767818 Chr.3: 182945331-182945331 on Build GRCh38
rs950041409 Chr.3: 182945379-182945379 on Build GRCh38
rs34134647 Chr.3: 182945366-182945367 on Build GRCh38
rs770949224 Chr.3: 182945169-182945169 on Build GRCh38
rs747254493 Chr.3: 182945105-182945105 on Build GRCh38
rs753217232 Chr.3: 182945051-182945051 on Build GRCh38
rs745778347 Chr.3: 182945184-182945184 on Build GRCh38
rs779690107 Chr.3: 182945194-182945197 on Build GRCh38
rs757017923 Chr.3: 182945315-182945315 on Build GRCh38
rs758736228 Chr.3: 182945082-182945082 on Build GRCh38
rs757847059 Chr.3: 182945055-182945055 on Build GRCh38
rs1010469483 Chr.3: 182945227-182945227 on Build GRCh38
rs557458040 Chr.3: 182945388-182945388 on Build GRCh38
rs760403543 Chr.3: 182945193-182945193 on Build GRCh38
rs111464424 Chr.3: 182945057-182945057 on Build GRCh38
rs780360395 Chr.3: 182945087-182945087 on Build GRCh38
rs972256012 Chr.3: 182945410-182945410 on Build GRCh38
rs185328412 Chr.3: 182945178-182945178 on Build GRCh38
rs776259271 Chr.3: 182945203-182945203 on Build GRCh38
rs775363027 Chr.3: 182945187-182945187 on Build GRCh38
rs967318450 Chr.3: 182945297-182945297 on Build GRCh38
rs761210517 Chr.3: 182945210-182945210 on Build GRCh38
rs56199887 Chr.3: 182945035-182945035 on Build GRCh38
rs763707789 Chr.3: 182945045-182945045 on Build GRCh38
rs908954176 Chr.3: 182945395-182945395 on Build GRCh38
rs759110660 Chr.3: 182945305-182945308 on Build GRCh38
rs1017312585 Chr.3: 182945294-182945294 on Build GRCh38
rs1036602048 Chr.3: 182944985-182944985 on Build GRCh38
rs34640949 Chr.3: 182945355-182945356 on Build GRCh38
rs765652635 Chr.3: 182945064-182945064 on Build GRCh38
rs917160607 Chr.3: 182945362-182945362 on Build GRCh38
rs750928578 Chr.3: 182945217-182945217 on Build GRCh38
rs963548321 Chr.3: 182945408-182945408 on Build GRCh38
rs556671532 Chr.3: 182945249-182945250 on Build GRCh38
rs73883963 Chr.3: 182945239-182945239 on Build GRCh38
rs1008985308 Chr.3: 182945291-182945291 on Build GRCh38
rs964419296 Chr.3: 182945310-182945310 on Build GRCh38
rs781338469 Chr.3: 182945113-182945113 on Build GRCh38
rs190569356 Chr.3: 182945325-182945325 on Build GRCh38
rs1054563608 Chr.3: 182945440-182945440 on Build GRCh38
rs753829144 Chr.3: 182945274-182945274 on Build GRCh38
rs973153811 Chr.3: 182945311-182945311 on Build GRCh38
rs927307468 Chr.3: 182945431-182945431 on Build GRCh38
rs748173202 Chr.3: 182945145-182945145 on Build GRCh38
rs758476133 Chr.3: 182945178-182945178 on Build GRCh38
rs750851979 Chr.3: 182945071-182945071 on Build GRCh38
rs778821168 Chr.3: 182945335-182945335 on Build GRCh38
rs772014555 Chr.3: 182945186-182945186 on Build GRCh38
rs750389110 Chr.3: 182945088-182945091 on Build GRCh38
rs1051776339 Chr.3: 182945067-182945067 on Build GRCh38
rs891992431 Chr.3: 182945213-182945213 on Build GRCh38
rs936052656 Chr.3: 182945437-182945437 on Build GRCh38
rs919509485 Chr.3: 182945027-182945027 on Build GRCh38
rs768417171 Chr.3: 182945196-182945196 on Build GRCh38

View Full Product Details