Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 182961111-182961659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCUN1D1
Entrez Gene ID
54165
Gene Name
defective in cullin neddylation 1 domain containing 1
Gene Aliases
DCNL1, DCUN1L1, RP42, SCCRO, SCRO, Tes3
UniGene
Hs.592789
Gene Chromosome Location
Chr.3: 182938158-182986364 on Build GRCh38
Transcripts
NM_001308101.1 NM_020640.3 XM_011512912.1 XM_011512914.1 XM_011512915.2 XM_011512916.2 XM_017006633.1 XM_017006634.1 XM_017006635.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5003702 c.457C>T p.R153* Chr.3: 182961289-182961289 on Build GRCh38
1203176 c.498G>T p.K166N Chr.3: 182961248-182961248 on Build GRCh38
3682980 c.423A>C p.I141I Chr.3: 182961323-182961323 on Build GRCh38
280324 c.390-8C>T p.? Chr.3: 182961364-182961364 on Build GRCh38
6535224 c.413A>C p.K138T Chr.3: 182961333-182961333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773792110 Chr.3: 182961313-182961313 on Build GRCh38
rs368498955 Chr.3: 182961567-182961570 on Build GRCh38
rs200237071 Chr.3: 182961303-182961303 on Build GRCh38
rs755782222 Chr.3: 182961245-182961245 on Build GRCh38
rs144588807 Chr.3: 182961338-182961338 on Build GRCh38
rs1049665835 Chr.3: 182961340-182961340 on Build GRCh38
rs61735023 Chr.3: 182961311-182961311 on Build GRCh38
rs936464672 Chr.3: 182961629-182961629 on Build GRCh38
rs754590083 Chr.3: 182961531-182961531 on Build GRCh38
rs966639 Chr.3: 182961541-182961541 on Build GRCh38
rs144878955 Chr.3: 182961181-182961181 on Build GRCh38
rs989028585 Chr.3: 182961327-182961327 on Build GRCh38
rs922979204 Chr.3: 182961346-182961346 on Build GRCh38
rs780576525 Chr.3: 182961386-182961386 on Build GRCh38
rs374108147 Chr.3: 182961377-182961377 on Build GRCh38
rs985313395 Chr.3: 182961595-182961595 on Build GRCh38
rs777384692 Chr.3: 182961288-182961288 on Build GRCh38
rs769168770 Chr.3: 182961405-182961405 on Build GRCh38
rs571613788 Chr.3: 182961165-182961165 on Build GRCh38
rs746363788 Chr.3: 182961386-182961386 on Build GRCh38
rs377642178 Chr.3: 182961300-182961300 on Build GRCh38
rs955189143 Chr.3: 182961570-182961570 on Build GRCh38
rs924922865 Chr.3: 182961578-182961578 on Build GRCh38
rs922433283 Chr.3: 182961335-182961335 on Build GRCh38
rs892616351 Chr.3: 182961404-182961405 on Build GRCh38
rs780854096 Chr.3: 182961231-182961231 on Build GRCh38
rs370774289 Chr.3: 182961354-182961354 on Build GRCh38
rs777028043 Chr.3: 182961341-182961341 on Build GRCh38
rs969372083 Chr.3: 182961171-182961171 on Build GRCh38
rs527257512 Chr.3: 182961564-182961564 on Build GRCh38
rs772501205 Chr.3: 182961382-182961382 on Build GRCh38
rs753239851 Chr.3: 182961484-182961484 on Build GRCh38
rs765370463 Chr.3: 182961214-182961214 on Build GRCh38
rs755909007 Chr.3: 182961363-182961363 on Build GRCh38
rs1013827767 Chr.3: 182961156-182961156 on Build GRCh38
rs747971698 Chr.3: 182961244-182961244 on Build GRCh38
rs747431223 Chr.3: 182961395-182961395 on Build GRCh38
rs968301128 Chr.3: 182961493-182961493 on Build GRCh38
rs765925164 Chr.3: 182961461-182961461 on Build GRCh38
rs373395712 Chr.3: 182961190-182961190 on Build GRCh38
rs756967292 Chr.3: 182961574-182961574 on Build GRCh38
rs1023825801 Chr.3: 182961478-182961478 on Build GRCh38
rs922380977 Chr.3: 182961535-182961535 on Build GRCh38
rs1056993243 Chr.3: 182961451-182961451 on Build GRCh38
rs753785444 Chr.3: 182961200-182961200 on Build GRCh38
rs905408191 Chr.3: 182961469-182961469 on Build GRCh38
rs1013881474 Chr.3: 182961437-182961437 on Build GRCh38
rs778334909 Chr.3: 182961610-182961610 on Build GRCh38
rs758158829 Chr.3: 182961453-182961453 on Build GRCh38
rs988587771 Chr.3: 182961533-182961533 on Build GRCh38
rs771333618 Chr.3: 182961214-182961214 on Build GRCh38
rs1032697786 Chr.3: 182961517-182961517 on Build GRCh38
rs879177878 Chr.3: 182961509-182961509 on Build GRCh38
rs761075296 Chr.3: 182961337-182961337 on Build GRCh38
rs146767924 Chr.3: 182961607-182961607 on Build GRCh38
rs1030213108 Chr.3: 182961554-182961554 on Build GRCh38
rs576612301 Chr.3: 182961316-182961316 on Build GRCh38
rs760539146 Chr.3: 182961144-182961144 on Build GRCh38
rs775772964 Chr.3: 182961329-182961329 on Build GRCh38
rs961849411 Chr.3: 182961533-182961535 on Build GRCh38
rs750468790 Chr.3: 182961359-182961359 on Build GRCh38
rs202126248 Chr.3: 182961348-182961348 on Build GRCh38
rs1012401461 Chr.3: 182961477-182961477 on Build GRCh38
rs554123358 Chr.3: 182961164-182961164 on Build GRCh38
rs772613014 Chr.3: 182961388-182961427 on Build GRCh38
rs367888827 Chr.3: 182961361-182961361 on Build GRCh38
rs764991235 Chr.3: 182961389-182961389 on Build GRCh38
rs911128904 Chr.3: 182961596-182961596 on Build GRCh38
rs779321439 Chr.3: 182961366-182961366 on Build GRCh38
rs34676061 Chr.3: 182961562-182961562 on Build GRCh38
rs184671977 Chr.3: 182961364-182961364 on Build GRCh38
rs751573642 Chr.3: 182961230-182961230 on Build GRCh38
rs984656284 Chr.3: 182961268-182961268 on Build GRCh38
rs192147858 Chr.3: 182961360-182961360 on Build GRCh38
rs375877720 Chr.3: 182961202-182961202 on Build GRCh38
rs751131725 Chr.3: 182961513-182961513 on Build GRCh38
rs750046380 Chr.3: 182961421-182961421 on Build GRCh38
rs745366157 Chr.3: 182961381-182961381 on Build GRCh38

View Full Product Details