Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 183253118-183253648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT5
Entrez Gene ID
84002
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5
Gene Aliases
B3GN-T5, beta3Gn-T5
UniGene
Hs.718506
Gene Chromosome Location
Chr.3: 183253207-183273391 on Build GRCh38
Transcripts
NM_032047.4 XM_011513227.2 XM_011513228.2 NM_015078.3 XM_011512585.2 XM_011512586.2 XM_017005943.1 XM_017005945.1 XM_017005946.1

Gene Symbol
MCF2L2
Entrez Gene ID
23101
Gene Name
MCF.2 cell line derived transforming sequence-like 2
Gene Aliases
ARHGEF22
UniGene
Hs.208267
Gene Chromosome Location
Chr.3: 183176116-183428990 on Build GRCh38
Transcripts
NM_032047.4 XM_011513227.2 XM_011513228.2 NM_015078.3 XM_011512585.2 XM_011512586.2 XM_017005943.1 XM_017005945.1 XM_017005946.1

SNP Detail

SNP ID Chromosome Location
rs996713214 Chr.3: 183253563-183253563 on Build GRCh38
rs554646222 Chr.3: 183253220-183253220 on Build GRCh38
rs758536035 Chr.3: 183253491-183253491 on Build GRCh38
rs569787589 Chr.3: 183253366-183253366 on Build GRCh38
rs960573352 Chr.3: 183253614-183253614 on Build GRCh38
rs1019868692 Chr.3: 183253470-183253470 on Build GRCh38
rs997818612 Chr.3: 183253210-183253210 on Build GRCh38
rs553773069 Chr.3: 183253520-183253520 on Build GRCh38
rs888026868 Chr.3: 183253580-183253580 on Build GRCh38
rs145838225 Chr.3: 183253468-183253468 on Build GRCh38
rs80163073 Chr.3: 183253193-183253194 on Build GRCh38
rs923870181 Chr.3: 183253250-183253250 on Build GRCh38
rs1043702537 Chr.3: 183253509-183253509 on Build GRCh38
rs1016732243 Chr.3: 183253426-183253426 on Build GRCh38
rs186051455 Chr.3: 183253409-183253409 on Build GRCh38
rs916447143 Chr.3: 183253490-183253490 on Build GRCh38
rs376442899 Chr.3: 183253180-183253180 on Build GRCh38
rs796648149 Chr.3: 183253221-183253236 on Build GRCh38
rs947855809 Chr.3: 183253355-183253355 on Build GRCh38
rs902116010 Chr.3: 183253143-183253143 on Build GRCh38
rs917790248 Chr.3: 183253313-183253313 on Build GRCh38
rs1029627823 Chr.3: 183253287-183253287 on Build GRCh38
rs1028371748 Chr.3: 183253222-183253222 on Build GRCh38
rs898219117 Chr.3: 183253392-183253392 on Build GRCh38
rs572014065 Chr.3: 183253561-183253561 on Build GRCh38
rs924058308 Chr.3: 183253512-183253512 on Build GRCh38
rs1040834729 Chr.3: 183253526-183253526 on Build GRCh38
rs1050702374 Chr.3: 183253577-183253577 on Build GRCh38
rs536205886 Chr.3: 183253182-183253182 on Build GRCh38
rs977898216 Chr.3: 183253584-183253584 on Build GRCh38
rs571890929 Chr.3: 183253607-183253607 on Build GRCh38
rs977425485 Chr.3: 183253213-183253213 on Build GRCh38
rs377422556 Chr.3: 183253261-183253261 on Build GRCh38
rs398092038 Chr.3: 183253194-183253195 on Build GRCh38
rs996458995 Chr.3: 183253281-183253281 on Build GRCh38
rs13313978 Chr.3: 183253604-183253604 on Build GRCh38
rs112987327 Chr.3: 183253155-183253156 on Build GRCh38
rs542594311 Chr.3: 183253590-183253590 on Build GRCh38
rs993945648 Chr.3: 183253612-183253612 on Build GRCh38
rs964215673 Chr.3: 183253508-183253509 on Build GRCh38
rs889711914 Chr.3: 183253506-183253506 on Build GRCh38
rs927838624 Chr.3: 183253437-183253437 on Build GRCh38
rs577740403 Chr.3: 183253436-183253436 on Build GRCh38
rs1035148487 Chr.3: 183253613-183253613 on Build GRCh38
rs149822915 Chr.3: 183253427-183253427 on Build GRCh38
rs371835467 Chr.3: 183253140-183253140 on Build GRCh38
rs953699140 Chr.3: 183253320-183253320 on Build GRCh38
rs990760113 Chr.3: 183253474-183253474 on Build GRCh38
rs867747894 Chr.3: 183253552-183253552 on Build GRCh38

View Full Product Details