Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 183270194-183270703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT5
Entrez Gene ID
84002
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5
Gene Aliases
B3GN-T5, beta3Gn-T5
UniGene
Hs.718506
Gene Chromosome Location
Chr.3: 183253207-183273391 on Build GRCh38
Transcripts
NM_032047.4 XM_005247823.4 XM_005247824.3 XM_005247825.3 XM_011513225.2 XM_011513226.2 XM_011513227.2 XM_011513228.2 XM_011513229.2 XM_017007309.1 NM_015078.3 XM_011512585.2 XM_011512586.2 XM_017005943.1 XM_017005945.1 XM_017005946.1

Gene Symbol
MCF2L2
Entrez Gene ID
23101
Gene Name
MCF.2 cell line derived transforming sequence-like 2
Gene Aliases
ARHGEF22
UniGene
Hs.208267
Gene Chromosome Location
Chr.3: 183176116-183428990 on Build GRCh38
Transcripts
NM_032047.4 XM_005247823.4 XM_005247824.3 XM_005247825.3 XM_011513225.2 XM_011513226.2 XM_011513227.2 XM_011513228.2 XM_011513229.2 XM_017007309.1 NM_015078.3 XM_011512585.2 XM_011512586.2 XM_017005943.1 XM_017005945.1 XM_017005946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3373098 c.533G>A p.W178* Chr.3: 183270331-183270331 on Build GRCh38
78877 c.714C>A p.Y238* Chr.3: 183270512-183270512 on Build GRCh38
5026084 c.490T>A p.S164T Chr.3: 183270288-183270288 on Build GRCh38
3206587 c.553C>T p.H185Y Chr.3: 183270351-183270351 on Build GRCh38
4831553 c.451C>G p.Q151E Chr.3: 183270249-183270249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148521313 Chr.3: 183270581-183270581 on Build GRCh38
rs142129979 Chr.3: 183270344-183270344 on Build GRCh38
rs769134426 Chr.3: 183270374-183270376 on Build GRCh38
rs757410219 Chr.3: 183270432-183270432 on Build GRCh38
rs144646232 Chr.3: 183270681-183270681 on Build GRCh38
rs770531165 Chr.3: 183270627-183270627 on Build GRCh38
rs944436859 Chr.3: 183270495-183270495 on Build GRCh38
rs180964101 Chr.3: 183270334-183270334 on Build GRCh38
rs369205848 Chr.3: 183270358-183270358 on Build GRCh38
rs777449742 Chr.3: 183270613-183270613 on Build GRCh38
rs766934598 Chr.3: 183270383-183270383 on Build GRCh38
rs151026623 Chr.3: 183270644-183270644 on Build GRCh38
rs772844252 Chr.3: 183270513-183270513 on Build GRCh38
rs372670786 Chr.3: 183270380-183270380 on Build GRCh38
rs543575415 Chr.3: 183270512-183270512 on Build GRCh38
rs747306979 Chr.3: 183270319-183270319 on Build GRCh38
rs752499455 Chr.3: 183270604-183270604 on Build GRCh38
rs760733397 Chr.3: 183270462-183270462 on Build GRCh38
rs957615603 Chr.3: 183270292-183270292 on Build GRCh38
rs751976718 Chr.3: 183270228-183270228 on Build GRCh38
rs200721442 Chr.3: 183270607-183270607 on Build GRCh38
rs759992079 Chr.3: 183270391-183270391 on Build GRCh38
rs759746825 Chr.3: 183270222-183270222 on Build GRCh38
rs866974372 Chr.3: 183270589-183270589 on Build GRCh38
rs781516085 Chr.3: 183270236-183270236 on Build GRCh38
rs930566130 Chr.3: 183270527-183270527 on Build GRCh38
rs760987845 Chr.3: 183270680-183270680 on Build GRCh38
rs753064388 Chr.3: 183270402-183270402 on Build GRCh38
rs765544080 Chr.3: 183270608-183270608 on Build GRCh38
rs907298861 Chr.3: 183270597-183270597 on Build GRCh38
rs892992489 Chr.3: 183270475-183270475 on Build GRCh38
rs776397039 Chr.3: 183270338-183270338 on Build GRCh38
rs910501603 Chr.3: 183270449-183270449 on Build GRCh38
rs767794526 Chr.3: 183270395-183270395 on Build GRCh38
rs1038857854 Chr.3: 183270508-183270508 on Build GRCh38
rs369390470 Chr.3: 183270411-183270411 on Build GRCh38
rs745325444 Chr.3: 183270650-183270650 on Build GRCh38
rs558711608 Chr.3: 183270520-183270520 on Build GRCh38
rs756296159 Chr.3: 183270409-183270409 on Build GRCh38
rs1035756136 Chr.3: 183270281-183270281 on Build GRCh38
rs765432993 Chr.3: 183270582-183270582 on Build GRCh38
rs768307976 Chr.3: 183270336-183270336 on Build GRCh38
rs747782074 Chr.3: 183270343-183270343 on Build GRCh38
rs1045707401 Chr.3: 183270591-183270591 on Build GRCh38
rs767733409 Chr.3: 183270229-183270229 on Build GRCh38
rs139703393 Chr.3: 183270651-183270651 on Build GRCh38
rs768000813 Chr.3: 183270560-183270560 on Build GRCh38
rs747915894 Chr.3: 183270471-183270471 on Build GRCh38
rs146378010 Chr.3: 183270572-183270572 on Build GRCh38
rs752838845 Chr.3: 183270249-183270249 on Build GRCh38
rs769405398 Chr.3: 183270232-183270232 on Build GRCh38
rs113447284 Chr.3: 183270588-183270588 on Build GRCh38
rs142874374 Chr.3: 183270618-183270618 on Build GRCh38
rs544060812 Chr.3: 183270440-183270440 on Build GRCh38
rs759992762 Chr.3: 183270547-183270547 on Build GRCh38
rs947225960 Chr.3: 183270493-183270493 on Build GRCh38
rs756310252 Chr.3: 183270265-183270265 on Build GRCh38
rs775732268 Chr.3: 183270561-183270561 on Build GRCh38
rs746922841 Chr.3: 183270436-183270436 on Build GRCh38
rs1035370710 Chr.3: 183270625-183270625 on Build GRCh38
rs1002571616 Chr.3: 183270257-183270257 on Build GRCh38
rs369857642 Chr.3: 183270298-183270298 on Build GRCh38
rs762202095 Chr.3: 183270468-183270484 on Build GRCh38
rs754899411 Chr.3: 183270448-183270448 on Build GRCh38
rs764354287 Chr.3: 183270410-183270410 on Build GRCh38
rs774807011 Chr.3: 183270384-183270384 on Build GRCh38
rs780884644 Chr.3: 183270463-183270463 on Build GRCh38
rs140197372 Chr.3: 183270329-183270329 on Build GRCh38
rs777108603 Chr.3: 183270406-183270409 on Build GRCh38
rs750480690 Chr.3: 183270585-183270585 on Build GRCh38
rs755289892 Chr.3: 183270230-183270230 on Build GRCh38
rs770404604 Chr.3: 183270522-183270522 on Build GRCh38
rs957522638 Chr.3: 183270640-183270640 on Build GRCh38
rs902915427 Chr.3: 183270237-183270237 on Build GRCh38
rs773800140 Chr.3: 183270525-183270525 on Build GRCh38
rs780183264 Chr.3: 183270433-183270433 on Build GRCh38
rs761087698 Chr.3: 183270566-183270566 on Build GRCh38
rs373017855 Chr.3: 183270319-183270319 on Build GRCh38
rs754881748 Chr.3: 183270587-183270587 on Build GRCh38
rs777744768 Chr.3: 183270283-183270283 on Build GRCh38
rs577147009 Chr.3: 183270573-183270573 on Build GRCh38
rs776062689 Chr.3: 183270671-183270671 on Build GRCh38

View Full Product Details