Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 183272488-183272995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT5
Entrez Gene ID
84002
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5
Gene Aliases
B3GN-T5, beta3Gn-T5
UniGene
Hs.718506
Gene Chromosome Location
Chr.3: 183253207-183273391 on Build GRCh38
Transcripts
NM_032047.4 XM_005247823.4 XM_005247824.3 XM_005247825.3 XM_011513225.2 XM_011513226.2 XM_011513227.2 XM_011513228.2 XM_011513229.2 XM_017007309.1 NM_015078.3 XM_011512585.2 XM_011512586.2 XM_017005943.1 XM_017005944.1 XM_017005945.1 XM_017005946.1

Gene Symbol
MCF2L2
Entrez Gene ID
23101
Gene Name
MCF.2 cell line derived transforming sequence-like 2
Gene Aliases
ARHGEF22
UniGene
Hs.208267
Gene Chromosome Location
Chr.3: 183176116-183428990 on Build GRCh38
Transcripts
NM_032047.4 XM_005247823.4 XM_005247824.3 XM_005247825.3 XM_011513225.2 XM_011513226.2 XM_011513227.2 XM_011513228.2 XM_011513229.2 XM_017007309.1 NM_015078.3 XM_011512585.2 XM_011512586.2 XM_017005943.1 XM_017005944.1 XM_017005945.1 XM_017005946.1

SNP Detail

SNP ID Chromosome Location
rs1012280577 Chr.3: 183272924-183272924 on Build GRCh38
rs974801058 Chr.3: 183272759-183272759 on Build GRCh38
rs950472789 Chr.3: 183272708-183272708 on Build GRCh38
rs547197310 Chr.3: 183272811-183272811 on Build GRCh38
rs976801173 Chr.3: 183272621-183272621 on Build GRCh38
rs1056895440 Chr.3: 183272871-183272871 on Build GRCh38
rs777149179 Chr.3: 183272541-183272541 on Build GRCh38
rs1043918904 Chr.3: 183272810-183272810 on Build GRCh38
rs924070130 Chr.3: 183272816-183272816 on Build GRCh38
rs541750476 Chr.3: 183272650-183272650 on Build GRCh38
rs374900978 Chr.3: 183272742-183272742 on Build GRCh38
rs73884619 Chr.3: 183272575-183272575 on Build GRCh38
rs16857228 Chr.3: 183272959-183272959 on Build GRCh38
rs922212471 Chr.3: 183272763-183272763 on Build GRCh38
rs946947901 Chr.3: 183272789-183272789 on Build GRCh38
rs921465281 Chr.3: 183272623-183272623 on Build GRCh38
rs16857226 Chr.3: 183272556-183272556 on Build GRCh38
rs897456841 Chr.3: 183272777-183272777 on Build GRCh38
rs16857227 Chr.3: 183272754-183272754 on Build GRCh38
rs1027441884 Chr.3: 183272705-183272705 on Build GRCh38
rs940237909 Chr.3: 183272730-183272730 on Build GRCh38
rs568556719 Chr.3: 183272843-183272843 on Build GRCh38
rs777582524 Chr.3: 183272718-183272718 on Build GRCh38
rs536184575 Chr.3: 183272917-183272917 on Build GRCh38
rs753071286 Chr.3: 183272747-183272750 on Build GRCh38
rs1032053428 Chr.3: 183272571-183272571 on Build GRCh38
rs896459409 Chr.3: 183272615-183272615 on Build GRCh38
rs1035441678 Chr.3: 183272598-183272599 on Build GRCh38
rs765569853 Chr.3: 183272826-183272826 on Build GRCh38
rs186812949 Chr.3: 183272728-183272728 on Build GRCh38
rs530381643 Chr.3: 183272703-183272703 on Build GRCh38
rs73884620 Chr.3: 183272691-183272691 on Build GRCh38
rs969149588 Chr.3: 183272548-183272548 on Build GRCh38
rs894344221 Chr.3: 183272892-183272892 on Build GRCh38
rs935424718 Chr.3: 183272824-183272824 on Build GRCh38
rs973243061 Chr.3: 183272700-183272700 on Build GRCh38
rs1014603119 Chr.3: 183272668-183272668 on Build GRCh38
rs932807585 Chr.3: 183272681-183272681 on Build GRCh38
rs73184919 Chr.3: 183272942-183272942 on Build GRCh38
rs528156182 Chr.3: 183272775-183272775 on Build GRCh38
rs969933436 Chr.3: 183272694-183272694 on Build GRCh38
rs1056531279 Chr.3: 183272827-183272827 on Build GRCh38
rs191025704 Chr.3: 183272836-183272836 on Build GRCh38

View Full Product Details