Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184176859-184177354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2M1
Entrez Gene ID
1173
Gene Name
adaptor related protein complex 2 mu 1 subunit
Gene Aliases
AP50, CLAPM1, mu2
UniGene
Hs.518460
Gene Chromosome Location
Chr.3: 184174846-184184091 on Build GRCh38
Transcripts
NM_001025205.1 NM_001311198.1 NM_004068.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6568970 c.64G>A p.D22N Chr.3: 184177057-184177057 on Build GRCh38
6097176 c.42G>C p.V14V Chr.3: 184177035-184177035 on Build GRCh38
3945214 c.66T>A p.D22E Chr.3: 184177059-184177059 on Build GRCh38
3702375 c.73G>A p.G25R Chr.3: 184177066-184177066 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs997346356 Chr.3: 184177136-184177136 on Build GRCh38
rs919386343 Chr.3: 184176950-184176950 on Build GRCh38
rs1017122917 Chr.3: 184177070-184177070 on Build GRCh38
rs1054284197 Chr.3: 184177242-184177242 on Build GRCh38
rs564705439 Chr.3: 184176931-184176931 on Build GRCh38
rs780587205 Chr.3: 184177090-184177090 on Build GRCh38
rs546499164 Chr.3: 184176888-184176888 on Build GRCh38
rs1003475359 Chr.3: 184177313-184177313 on Build GRCh38
rs776152866 Chr.3: 184177116-184177117 on Build GRCh38
rs752850608 Chr.3: 184176976-184176976 on Build GRCh38
rs992944541 Chr.3: 184177314-184177314 on Build GRCh38
rs772599064 Chr.3: 184177081-184177081 on Build GRCh38
rs945479577 Chr.3: 184177260-184177260 on Build GRCh38
rs757550426 Chr.3: 184177048-184177048 on Build GRCh38
rs911120756 Chr.3: 184177190-184177190 on Build GRCh38
rs760460731 Chr.3: 184177241-184177241 on Build GRCh38
rs142388295 Chr.3: 184177181-184177181 on Build GRCh38
rs376237979 Chr.3: 184176991-184176991 on Build GRCh38
rs746308252 Chr.3: 184177074-184177074 on Build GRCh38
rs948860846 Chr.3: 184177032-184177032 on Build GRCh38
rs550921790 Chr.3: 184177143-184177143 on Build GRCh38
rs796938439 Chr.3: 184176910-184176910 on Build GRCh38
rs985910237 Chr.3: 184177174-184177174 on Build GRCh38
rs151304119 Chr.3: 184177285-184177285 on Build GRCh38
rs536982073 Chr.3: 184177253-184177253 on Build GRCh38
rs878937674 Chr.3: 184177038-184177038 on Build GRCh38
rs908904889 Chr.3: 184176896-184176896 on Build GRCh38
rs776385102 Chr.3: 184176958-184176958 on Build GRCh38
rs1057271546 Chr.3: 184176966-184176966 on Build GRCh38
rs1016319370 Chr.3: 184177232-184177232 on Build GRCh38
rs372679705 Chr.3: 184177099-184177099 on Build GRCh38
rs756315784 Chr.3: 184176990-184176990 on Build GRCh38
rs146101904 Chr.3: 184176953-184176953 on Build GRCh38
rs1020947170 Chr.3: 184176923-184176923 on Build GRCh38
rs985385426 Chr.3: 184177177-184177177 on Build GRCh38
rs201516833 Chr.3: 184176970-184176970 on Build GRCh38
rs200441721 Chr.3: 184176945-184176945 on Build GRCh38
rs776133057 Chr.3: 184177114-184177114 on Build GRCh38
rs1042826637 Chr.3: 184177265-184177265 on Build GRCh38
rs768276985 Chr.3: 184177113-184177113 on Build GRCh38
rs796997967 Chr.3: 184176898-184176898 on Build GRCh38
rs919507396 Chr.3: 184177278-184177278 on Build GRCh38
rs779351528 Chr.3: 184177062-184177062 on Build GRCh38
rs200980522 Chr.3: 184176975-184176975 on Build GRCh38
rs760265104 Chr.3: 184176964-184176964 on Build GRCh38
rs753312758 Chr.3: 184176986-184176986 on Build GRCh38
rs763658618 Chr.3: 184176974-184176974 on Build GRCh38
rs113379783 Chr.3: 184176948-184176948 on Build GRCh38
rs1003753822 Chr.3: 184176977-184176977 on Build GRCh38
rs889629165 Chr.3: 184176880-184176882 on Build GRCh38
rs953234157 Chr.3: 184177170-184177170 on Build GRCh38
rs556151421 Chr.3: 184177109-184177109 on Build GRCh38
rs1054301035 Chr.3: 184176895-184176895 on Build GRCh38
rs2231214 Chr.3: 184177035-184177035 on Build GRCh38
rs113086272 Chr.3: 184177069-184177069 on Build GRCh38

View Full Product Details