Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184181590-184182100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2M1
Entrez Gene ID
1173
Gene Name
adaptor related protein complex 2 mu 1 subunit
Gene Aliases
AP50, CLAPM1, mu2
UniGene
Hs.518460
Gene Chromosome Location
Chr.3: 184174846-184184091 on Build GRCh38
Transcripts
NM_001025205.1 NM_001311198.1 NM_004068.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
136393 c.799C>T p.P267S Chr.3: 184181787-184181787 on Build GRCh38
6601628 c.813G>T p.E271D Chr.3: 184181801-184181801 on Build GRCh38
3945215 c.872C>T p.P291L Chr.3: 184181956-184181956 on Build GRCh38
6601633 c.734A>G p.D245G Chr.3: 184181722-184181722 on Build GRCh38
5864014 c.961G>A p.E321K Chr.3: 184182045-184182045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779389033 Chr.3: 184181786-184181786 on Build GRCh38
rs752798083 Chr.3: 184181708-184181708 on Build GRCh38
rs754078750 Chr.3: 184181939-184181939 on Build GRCh38
rs921161297 Chr.3: 184181645-184181645 on Build GRCh38
rs144760576 Chr.3: 184181771-184181771 on Build GRCh38
rs756552319 Chr.3: 184181845-184181858 on Build GRCh38
rs746493726 Chr.3: 184181987-184181987 on Build GRCh38
rs8478 Chr.3: 184182044-184182044 on Build GRCh38
rs750886669 Chr.3: 184181963-184181963 on Build GRCh38
rs974810130 Chr.3: 184181623-184181623 on Build GRCh38
rs932142551 Chr.3: 184181854-184181854 on Build GRCh38
rs768679941 Chr.3: 184182072-184182072 on Build GRCh38
rs962288740 Chr.3: 184181662-184181662 on Build GRCh38
rs369968833 Chr.3: 184182005-184182005 on Build GRCh38
rs1019440607 Chr.3: 184182060-184182060 on Build GRCh38
rs542405163 Chr.3: 184181693-184181693 on Build GRCh38
rs557482117 Chr.3: 184181789-184181789 on Build GRCh38
rs762986870 Chr.3: 184181874-184181874 on Build GRCh38
rs761989264 Chr.3: 184181632-184181632 on Build GRCh38
rs776182971 Chr.3: 184181858-184181858 on Build GRCh38
rs746590871 Chr.3: 184181851-184181851 on Build GRCh38
rs374946932 Chr.3: 184181974-184181974 on Build GRCh38
rs754512906 Chr.3: 184182002-184182002 on Build GRCh38
rs373467264 Chr.3: 184182029-184182029 on Build GRCh38
rs765231211 Chr.3: 184181668-184181668 on Build GRCh38
rs757847867 Chr.3: 184181824-184181824 on Build GRCh38
rs767666661 Chr.3: 184181884-184181884 on Build GRCh38
rs953991301 Chr.3: 184181656-184181656 on Build GRCh38
rs377598426 Chr.3: 184182064-184182064 on Build GRCh38
rs1011177817 Chr.3: 184181918-184181918 on Build GRCh38
rs760808659 Chr.3: 184182065-184182065 on Build GRCh38
rs768328122 Chr.3: 184181852-184181852 on Build GRCh38
rs767715842 Chr.3: 184181696-184181696 on Build GRCh38
rs764508843 Chr.3: 184182056-184182057 on Build GRCh38
rs200439653 Chr.3: 184182057-184182057 on Build GRCh38
rs368390936 Chr.3: 184181714-184181714 on Build GRCh38
rs976295117 Chr.3: 184181670-184181670 on Build GRCh38
rs908158889 Chr.3: 184181646-184181646 on Build GRCh38
rs747952613 Chr.3: 184181860-184181860 on Build GRCh38
rs11539600 Chr.3: 184181932-184181932 on Build GRCh38
rs769769242 Chr.3: 184181864-184181864 on Build GRCh38
rs770879947 Chr.3: 184182061-184182061 on Build GRCh38
rs375009921 Chr.3: 184181822-184181822 on Build GRCh38
rs757608518 Chr.3: 184181950-184181950 on Build GRCh38
rs765602148 Chr.3: 184181957-184181957 on Build GRCh38
rs371916595 Chr.3: 184181893-184181893 on Build GRCh38
rs776779281 Chr.3: 184182073-184182073 on Build GRCh38
rs773174636 Chr.3: 184182056-184182056 on Build GRCh38
rs773223946 Chr.3: 184181873-184181873 on Build GRCh38
rs369667214 Chr.3: 184182053-184182053 on Build GRCh38
rs779644185 Chr.3: 184181829-184181829 on Build GRCh38
rs867287194 Chr.3: 184181942-184181942 on Build GRCh38
rs780471771 Chr.3: 184181980-184181980 on Build GRCh38
rs368900561 Chr.3: 184181865-184181865 on Build GRCh38
rs984777849 Chr.3: 184181628-184181628 on Build GRCh38
rs375041873 Chr.3: 184181654-184181654 on Build GRCh38
rs754435714 Chr.3: 184182056-184182056 on Build GRCh38
rs754212923 Chr.3: 184181729-184181729 on Build GRCh38
rs773346740 Chr.3: 184181673-184181673 on Build GRCh38
rs775662402 Chr.3: 184181885-184181885 on Build GRCh38
rs756393805 Chr.3: 184181711-184181711 on Build GRCh38
rs764346596 Chr.3: 184181919-184181919 on Build GRCh38

View Full Product Details