Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184337401-184337897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM131A
Entrez Gene ID
131408
Gene Name
family with sequence similarity 131 member A
Gene Aliases
C3orf40, FLAT715, PRO1378
UniGene
Hs.591307
Gene Chromosome Location
Chr.3: 184335924-184346275 on Build GRCh38
Transcripts
NM_001171093.1 NM_144635.4 XM_005247113.2 XM_005247114.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6351246 c.7A>G p.M3V Chr.3: 184337637-184337637 on Build GRCh38
3767336 c.66G>A p.W22* Chr.3: 184337696-184337696 on Build GRCh38
446100 c.26A>G p.K9R Chr.3: 184337656-184337656 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1020310001 Chr.3: 184337694-184337694 on Build GRCh38
rs752515459 Chr.3: 184337808-184337808 on Build GRCh38
rs942924358 Chr.3: 184337612-184337612 on Build GRCh38
rs887504286 Chr.3: 184337695-184337695 on Build GRCh38
rs536297259 Chr.3: 184337850-184337850 on Build GRCh38
rs776058971 Chr.3: 184337645-184337645 on Build GRCh38
rs541800165 Chr.3: 184337470-184337470 on Build GRCh38
rs547207412 Chr.3: 184337609-184337609 on Build GRCh38
rs935253898 Chr.3: 184337535-184337535 on Build GRCh38
rs772195107 Chr.3: 184337586-184337587 on Build GRCh38
rs746205957 Chr.3: 184337622-184337622 on Build GRCh38
rs532297333 Chr.3: 184337587-184337587 on Build GRCh38
rs565836225 Chr.3: 184337873-184337873 on Build GRCh38
rs775965342 Chr.3: 184337734-184337734 on Build GRCh38
rs759503274 Chr.3: 184337431-184337431 on Build GRCh38
rs779126703 Chr.3: 184337592-184337592 on Build GRCh38
rs775665731 Chr.3: 184337594-184337594 on Build GRCh38
rs891330206 Chr.3: 184337480-184337480 on Build GRCh38
rs977374017 Chr.3: 184337517-184337517 on Build GRCh38
rs1021771294 Chr.3: 184337863-184337863 on Build GRCh38
rs767456348 Chr.3: 184337551-184337551 on Build GRCh38
rs924634771 Chr.3: 184337532-184337532 on Build GRCh38
rs923216890 Chr.3: 184337590-184337590 on Build GRCh38
rs1016094184 Chr.3: 184337744-184337744 on Build GRCh38
rs1006001622 Chr.3: 184337709-184337709 on Build GRCh38
rs1050264961 Chr.3: 184337626-184337626 on Build GRCh38
rs931769895 Chr.3: 184337615-184337615 on Build GRCh38
rs747525323 Chr.3: 184337674-184337674 on Build GRCh38
rs918944773 Chr.3: 184337844-184337844 on Build GRCh38
rs897519885 Chr.3: 184337739-184337739 on Build GRCh38
rs560898403 Chr.3: 184337500-184337500 on Build GRCh38
rs145928718 Chr.3: 184337430-184337430 on Build GRCh38
rs1003393161 Chr.3: 184337723-184337723 on Build GRCh38
rs571360080 Chr.3: 184337571-184337571 on Build GRCh38
rs763661353 Chr.3: 184337867-184337867 on Build GRCh38
rs968791590 Chr.3: 184337493-184337493 on Build GRCh38
rs760431357 Chr.3: 184337857-184337857 on Build GRCh38
rs549626607 Chr.3: 184337550-184337550 on Build GRCh38
rs1035525789 Chr.3: 184337735-184337735 on Build GRCh38
rs991623378 Chr.3: 184337482-184337482 on Build GRCh38
rs1036017356 Chr.3: 184337713-184337713 on Build GRCh38
rs548638516 Chr.3: 184337667-184337667 on Build GRCh38
rs963281717 Chr.3: 184337762-184337762 on Build GRCh38
rs1011561782 Chr.3: 184337692-184337692 on Build GRCh38
rs772466270 Chr.3: 184337639-184337639 on Build GRCh38
rs958819728 Chr.3: 184337736-184337736 on Build GRCh38
rs757428154 Chr.3: 184337591-184337591 on Build GRCh38
rs531495416 Chr.3: 184337533-184337533 on Build GRCh38
rs752489739 Chr.3: 184337465-184337466 on Build GRCh38
rs914628233 Chr.3: 184337491-184337491 on Build GRCh38
rs565744092 Chr.3: 184337804-184337804 on Build GRCh38
rs1036316598 Chr.3: 184337807-184337807 on Build GRCh38
rs769212848 Chr.3: 184337688-184337688 on Build GRCh38
rs1002543681 Chr.3: 184337768-184337768 on Build GRCh38
rs765245475 Chr.3: 184337800-184337800 on Build GRCh38

View Full Product Details