Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184342660-184343166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM131A
Entrez Gene ID
131408
Gene Name
family with sequence similarity 131 member A
Gene Aliases
C3orf40, FLAT715, PRO1378
UniGene
Hs.591307
Gene Chromosome Location
Chr.3: 184335924-184346275 on Build GRCh38
Transcripts
NM_001171093.1 NM_144635.4 XM_005247113.2 XM_005247114.3 XM_011512413.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1042041 c.573G>T p.E191D Chr.3: 184342808-184342808 on Build GRCh38
3590758 c.546C>T p.L182L Chr.3: 184342781-184342781 on Build GRCh38
4115765 c.512T>C p.F171S Chr.3: 184342840-184342840 on Build GRCh38
6850144 c.562G>T p.V188L Chr.3: 184342797-184342797 on Build GRCh38
1241142 c.570C>T p.G190G Chr.3: 184342805-184342805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554199917 Chr.3: 184342919-184342919 on Build GRCh38
rs753635359 Chr.3: 184342808-184342808 on Build GRCh38
rs369256278 Chr.3: 184342701-184342701 on Build GRCh38
rs995848800 Chr.3: 184343143-184343143 on Build GRCh38
rs770060271 Chr.3: 184342754-184342754 on Build GRCh38
rs895403567 Chr.3: 184342911-184342911 on Build GRCh38
rs755081249 Chr.3: 184342741-184342741 on Build GRCh38
rs771348837 Chr.3: 184342893-184342893 on Build GRCh38
rs972845096 Chr.3: 184343000-184343000 on Build GRCh38
rs753689374 Chr.3: 184342736-184342736 on Build GRCh38
rs543300173 Chr.3: 184342968-184342968 on Build GRCh38
rs111824196 Chr.3: 184342867-184342867 on Build GRCh38
rs763735013 Chr.3: 184342735-184342735 on Build GRCh38
rs774882447 Chr.3: 184342783-184342783 on Build GRCh38
rs748260920 Chr.3: 184342753-184342753 on Build GRCh38
rs200132624 Chr.3: 184342738-184342738 on Build GRCh38
rs200914816 Chr.3: 184342894-184342894 on Build GRCh38
rs766907588 Chr.3: 184342702-184342702 on Build GRCh38
rs748294579 Chr.3: 184342849-184342849 on Build GRCh38
rs779910983 Chr.3: 184342898-184342898 on Build GRCh38
rs567075819 Chr.3: 184342739-184342739 on Build GRCh38
rs3064291 Chr.3: 184343122-184343123 on Build GRCh38
rs759098355 Chr.3: 184342712-184342712 on Build GRCh38
rs374971509 Chr.3: 184343098-184343098 on Build GRCh38
rs543576227 Chr.3: 184343113-184343113 on Build GRCh38
rs5855023 Chr.3: 184343143-184343144 on Build GRCh38
rs751024706 Chr.3: 184342710-184342710 on Build GRCh38
rs766989260 Chr.3: 184342789-184342789 on Build GRCh38
rs775077047 Chr.3: 184342796-184342796 on Build GRCh38
rs746314462 Chr.3: 184342896-184342896 on Build GRCh38
rs201459098 Chr.3: 184342914-184342914 on Build GRCh38
rs770114794 Chr.3: 184342697-184342697 on Build GRCh38
rs781068515 Chr.3: 184342848-184342848 on Build GRCh38
rs962077748 Chr.3: 184342826-184342826 on Build GRCh38
rs781465189 Chr.3: 184342748-184342748 on Build GRCh38
rs933887009 Chr.3: 184342696-184342696 on Build GRCh38
rs183160243 Chr.3: 184343060-184343060 on Build GRCh38
rs773412803 Chr.3: 184342768-184342768 on Build GRCh38
rs572764470 Chr.3: 184342965-184342965 on Build GRCh38
rs760175064 Chr.3: 184342797-184342797 on Build GRCh38
rs749626106 Chr.3: 184342891-184342891 on Build GRCh38
rs757165844 Chr.3: 184342814-184342814 on Build GRCh38
rs142482105 Chr.3: 184342829-184342829 on Build GRCh38
rs537556690 Chr.3: 184342769-184342769 on Build GRCh38
rs762235607 Chr.3: 184342695-184342695 on Build GRCh38
rs746513812 Chr.3: 184343119-184343119 on Build GRCh38
rs758370307 Chr.3: 184342846-184342846 on Build GRCh38
rs564918794 Chr.3: 184342733-184342733 on Build GRCh38
rs543561165 Chr.3: 184342722-184342722 on Build GRCh38
rs746248453 Chr.3: 184342687-184342687 on Build GRCh38
rs774821983 Chr.3: 184342908-184342908 on Build GRCh38
rs771408226 Chr.3: 184342778-184342778 on Build GRCh38
rs1029757921 Chr.3: 184343027-184343027 on Build GRCh38
rs571174963 Chr.3: 184342805-184342805 on Build GRCh38
rs946568026 Chr.3: 184342951-184342951 on Build GRCh38
rs145559471 Chr.3: 184342787-184342787 on Build GRCh38
rs9682844 Chr.3: 184342973-184342973 on Build GRCh38
rs777602901 Chr.3: 184342723-184342723 on Build GRCh38
rs565360413 Chr.3: 184343146-184343146 on Build GRCh38
rs750444368 Chr.3: 184342839-184342839 on Build GRCh38
rs116476249 Chr.3: 184342699-184342699 on Build GRCh38
rs754713023 Chr.3: 184342726-184342726 on Build GRCh38

View Full Product Details