Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184344779-184345285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM131A
Entrez Gene ID
131408
Gene Name
family with sequence similarity 131 member A
Gene Aliases
C3orf40, FLAT715, PRO1378
UniGene
Hs.591307
Gene Chromosome Location
Chr.3: 184335924-184346275 on Build GRCh38
Transcripts
NM_001171093.1 NM_144635.4 XM_005247113.2 XM_005247114.3 XM_011512413.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4625025 c.1014C>A p.S338R Chr.3: 184344883-184344883 on Build GRCh38
6097151 c.979G>C p.E327Q Chr.3: 184344941-184344941 on Build GRCh38
6408208 c.935A>G p.E312G Chr.3: 184344804-184344804 on Build GRCh38
4682540 c.1007C>T p.T336M Chr.3: 184344876-184344876 on Build GRCh38
6313242 c.916G>T p.G306C Chr.3: 184344878-184344878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749385469 Chr.3: 184344953-184344953 on Build GRCh38
rs774589741 Chr.3: 184344963-184344963 on Build GRCh38
rs759969947 Chr.3: 184344877-184344877 on Build GRCh38
rs767940819 Chr.3: 184344883-184344883 on Build GRCh38
rs1037583549 Chr.3: 184345076-184345076 on Build GRCh38
rs62287503 Chr.3: 184344925-184344925 on Build GRCh38
rs778477208 Chr.3: 184344829-184344829 on Build GRCh38
rs757704272 Chr.3: 184344836-184344837 on Build GRCh38
rs746890853 Chr.3: 184344839-184344839 on Build GRCh38
rs1056889602 Chr.3: 184345063-184345063 on Build GRCh38
rs890208085 Chr.3: 184345106-184345107 on Build GRCh38
rs768618051 Chr.3: 184344940-184344940 on Build GRCh38
rs1021847342 Chr.3: 184345200-184345200 on Build GRCh38
rs911254096 Chr.3: 184345179-184345179 on Build GRCh38
rs948567685 Chr.3: 184345071-184345071 on Build GRCh38
rs948957320 Chr.3: 184344802-184344802 on Build GRCh38
rs370485685 Chr.3: 184344987-184344987 on Build GRCh38
rs529758058 Chr.3: 184344922-184344922 on Build GRCh38
rs565108026 Chr.3: 184344852-184344852 on Build GRCh38
rs768137101 Chr.3: 184344810-184344810 on Build GRCh38
rs142507952 Chr.3: 184344835-184344835 on Build GRCh38
rs753318197 Chr.3: 184344817-184344817 on Build GRCh38
rs762993503 Chr.3: 184344858-184344858 on Build GRCh38
rs759770014 Chr.3: 184344971-184344971 on Build GRCh38
rs1013935841 Chr.3: 184345022-184345022 on Build GRCh38
rs868610503 Chr.3: 184344899-184344899 on Build GRCh38
rs746829651 Chr.3: 184344927-184344927 on Build GRCh38
rs547386620 Chr.3: 184345005-184345005 on Build GRCh38
rs777265957 Chr.3: 184344821-184344821 on Build GRCh38
rs534866278 Chr.3: 184345091-184345091 on Build GRCh38
rs979462973 Chr.3: 184345087-184345087 on Build GRCh38
rs745630112 Chr.3: 184344830-184344830 on Build GRCh38
rs754154368 Chr.3: 184344823-184344825 on Build GRCh38
rs375453499 Chr.3: 184344820-184344820 on Build GRCh38
rs922699349 Chr.3: 184345105-184345105 on Build GRCh38
rs926183999 Chr.3: 184345096-184345096 on Build GRCh38
rs34826748 Chr.3: 184344848-184344849 on Build GRCh38
rs559476814 Chr.3: 184344897-184344897 on Build GRCh38
rs768507394 Chr.3: 184344842-184344842 on Build GRCh38
rs144826588 Chr.3: 184344876-184344876 on Build GRCh38
rs368029423 Chr.3: 184345004-184345004 on Build GRCh38
rs956219492 Chr.3: 184345103-184345103 on Build GRCh38
rs569505156 Chr.3: 184345025-184345025 on Build GRCh38
rs771155670 Chr.3: 184344868-184344868 on Build GRCh38
rs144919867 Chr.3: 184345237-184345237 on Build GRCh38
rs148217150 Chr.3: 184344890-184344890 on Build GRCh38
rs779272839 Chr.3: 184345015-184345016 on Build GRCh38
rs9839000 Chr.3: 184344988-184344988 on Build GRCh38
rs868402817 Chr.3: 184344944-184344944 on Build GRCh38
rs745621367 Chr.3: 184344884-184344885 on Build GRCh38
rs764528978 Chr.3: 184344887-184344887 on Build GRCh38
rs926071707 Chr.3: 184344816-184344816 on Build GRCh38
rs757638263 Chr.3: 184345056-184345056 on Build GRCh38
rs988957272 Chr.3: 184345104-184345104 on Build GRCh38
rs879139083 Chr.3: 184345036-184345036 on Build GRCh38
rs748776023 Chr.3: 184344822-184344822 on Build GRCh38
rs374349895 Chr.3: 184344886-184344886 on Build GRCh38
rs141220191 Chr.3: 184344915-184344915 on Build GRCh38
rs779785469 Chr.3: 184344923-184344923 on Build GRCh38
rs975659748 Chr.3: 184345000-184345000 on Build GRCh38
rs924684733 Chr.3: 184345047-184345047 on Build GRCh38
rs781033931 Chr.3: 184344949-184344949 on Build GRCh38
rs1049755568 Chr.3: 184345158-184345158 on Build GRCh38
rs770977453 Chr.3: 184344954-184344954 on Build GRCh38
rs969404057 Chr.3: 184345206-184345207 on Build GRCh38
rs779777777 Chr.3: 184344819-184344819 on Build GRCh38
rs771364273 Chr.3: 184345189-184345189 on Build GRCh38
rs775392036 Chr.3: 184344838-184344838 on Build GRCh38
rs1001939576 Chr.3: 184345185-184345194 on Build GRCh38
rs934037989 Chr.3: 184345143-184345143 on Build GRCh38
rs889048318 Chr.3: 184344854-184344854 on Build GRCh38
rs377287428 Chr.3: 184344891-184344891 on Build GRCh38
rs993309960 Chr.3: 184345252-184345252 on Build GRCh38
rs149106753 Chr.3: 184345064-184345064 on Build GRCh38
rs749715474 Chr.3: 184345180-184345180 on Build GRCh38
rs552560133 Chr.3: 184345046-184345046 on Build GRCh38
rs755437344 Chr.3: 184344807-184344807 on Build GRCh38
rs899070214 Chr.3: 184345238-184345238 on Build GRCh38
rs1017781951 Chr.3: 184345169-184345169 on Build GRCh38
rs1016182850 Chr.3: 184344882-184344882 on Build GRCh38
rs934813185 Chr.3: 184344818-184344818 on Build GRCh38
rs1008688102 Chr.3: 184345120-184345120 on Build GRCh38
rs993151373 Chr.3: 184345095-184345095 on Build GRCh38
rs764628737 Chr.3: 184345003-184345003 on Build GRCh38
rs760991776 Chr.3: 184344995-184344995 on Build GRCh38
rs977403458 Chr.3: 184345035-184345035 on Build GRCh38
rs1019833919 Chr.3: 184344907-184344907 on Build GRCh38
rs756881641 Chr.3: 184344823-184344823 on Build GRCh38
rs537757987 Chr.3: 184345030-184345030 on Build GRCh38

View Full Product Details