Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184380415-184380922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRD
Entrez Gene ID
8646
Gene Name
chordin
Gene Aliases
-
UniGene
Hs.166186
Gene Chromosome Location
Chr.3: 184380073-184389835 on Build GRCh38
Transcripts
NM_001304472.1 NM_001304473.1 NM_001304474.1 NM_003741.3 NR_130747.1 XM_011513254.1 XM_017007388.1 XM_017007389.1 XM_017007390.1 XM_017007391.1 XM_017007392.1 XM_017007393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6036000 c.208G>C p.E70Q Chr.3: 184380751-184380751 on Build GRCh38
6500581 c.136C>T p.R46W Chr.3: 184380454-184380454 on Build GRCh38
6391187 c.160G>C p.G54R Chr.3: 184380703-184380703 on Build GRCh38
1042064 c.157T>G p.F53V Chr.3: 184380700-184380700 on Build GRCh38
6391188 c.160G>C p.G54R Chr.3: 184380703-184380703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960579507 Chr.3: 184380468-184380468 on Build GRCh38
rs750135804 Chr.3: 184380681-184380681 on Build GRCh38
rs1014404275 Chr.3: 184380497-184380497 on Build GRCh38
rs143811087 Chr.3: 184380641-184380641 on Build GRCh38
rs950986539 Chr.3: 184380878-184380878 on Build GRCh38
rs902440604 Chr.3: 184380687-184380687 on Build GRCh38
rs765337014 Chr.3: 184380808-184380808 on Build GRCh38
rs772927377 Chr.3: 184380824-184380824 on Build GRCh38
rs754935921 Chr.3: 184380688-184380688 on Build GRCh38
rs372672105 Chr.3: 184380822-184380822 on Build GRCh38
rs767309911 Chr.3: 184380784-184380784 on Build GRCh38
rs563846109 Chr.3: 184380478-184380478 on Build GRCh38
rs767385245 Chr.3: 184380830-184380830 on Build GRCh38
rs757304300 Chr.3: 184380799-184380799 on Build GRCh38
rs761831874 Chr.3: 184380616-184380616 on Build GRCh38
rs751498580 Chr.3: 184380685-184380685 on Build GRCh38
rs974101920 Chr.3: 184380540-184380540 on Build GRCh38
rs779022396 Chr.3: 184380801-184380801 on Build GRCh38
rs1023447001 Chr.3: 184380486-184380486 on Build GRCh38
rs994197153 Chr.3: 184380818-184380818 on Build GRCh38
rs1023131086 Chr.3: 184380835-184380835 on Build GRCh38
rs1004821616 Chr.3: 184380880-184380880 on Build GRCh38
rs190522280 Chr.3: 184380774-184380774 on Build GRCh38
rs757349750 Chr.3: 184380698-184380698 on Build GRCh38
rs545545389 Chr.3: 184380457-184380457 on Build GRCh38
rs983431085 Chr.3: 184380582-184380582 on Build GRCh38
rs900399850 Chr.3: 184380450-184380450 on Build GRCh38
rs763657800 Chr.3: 184380662-184380662 on Build GRCh38
rs368754452 Chr.3: 184380796-184380796 on Build GRCh38
rs766189404 Chr.3: 184380748-184380748 on Build GRCh38
rs569519997 Chr.3: 184380825-184380825 on Build GRCh38
rs769465205 Chr.3: 184380731-184380731 on Build GRCh38
rs759391287 Chr.3: 184380782-184380782 on Build GRCh38
rs1023580769 Chr.3: 184380515-184380515 on Build GRCh38
rs896253227 Chr.3: 184380492-184380492 on Build GRCh38
rs9880198 Chr.3: 184380771-184380771 on Build GRCh38
rs781515509 Chr.3: 184380819-184380819 on Build GRCh38
rs974589552 Chr.3: 184380650-184380650 on Build GRCh38
rs1056163964 Chr.3: 184380719-184380719 on Build GRCh38
rs778785026 Chr.3: 184380642-184380643 on Build GRCh38
rs902221751 Chr.3: 184380477-184380477 on Build GRCh38
rs761145242 Chr.3: 184380649-184380649 on Build GRCh38
rs885650 Chr.3: 184380859-184380859 on Build GRCh38
rs761370761 Chr.3: 184380722-184380722 on Build GRCh38
rs779067519 Chr.3: 184380700-184380700 on Build GRCh38
rs762871495 Chr.3: 184380868-184380868 on Build GRCh38
rs373677799 Chr.3: 184380674-184380674 on Build GRCh38
rs769092393 Chr.3: 184380815-184380815 on Build GRCh38
rs750637775 Chr.3: 184380802-184380802 on Build GRCh38
rs748717476 Chr.3: 184380820-184380820 on Build GRCh38
rs772413432 Chr.3: 184380703-184380703 on Build GRCh38
rs974174807 Chr.3: 184380581-184380581 on Build GRCh38
rs760671906 Chr.3: 184380844-184380844 on Build GRCh38
rs746138623 Chr.3: 184380702-184380702 on Build GRCh38
rs771072986 Chr.3: 184380455-184380455 on Build GRCh38
rs772466378 Chr.3: 184380480-184380480 on Build GRCh38
rs747284058 Chr.3: 184380809-184380809 on Build GRCh38
rs777220814 Chr.3: 184380717-184380717 on Build GRCh38
rs963147124 Chr.3: 184380500-184380500 on Build GRCh38
rs752617161 Chr.3: 184380841-184380841 on Build GRCh38
rs561705843 Chr.3: 184380682-184380682 on Build GRCh38
rs764938722 Chr.3: 184380679-184380679 on Build GRCh38
rs375577594 Chr.3: 184380829-184380829 on Build GRCh38
rs768990411 Chr.3: 184380715-184380715 on Build GRCh38
rs1027834704 Chr.3: 184380863-184380863 on Build GRCh38
rs781192276 Chr.3: 184380692-184380692 on Build GRCh38
rs970328243 Chr.3: 184380530-184380530 on Build GRCh38
rs140559964 Chr.3: 184380738-184380738 on Build GRCh38
rs766133113 Chr.3: 184380684-184380684 on Build GRCh38
rs868352240 Chr.3: 184380546-184380546 on Build GRCh38
rs1043394021 Chr.3: 184380471-184380471 on Build GRCh38
rs775787241 Chr.3: 184380704-184380704 on Build GRCh38
rs747406248 Chr.3: 184380705-184380705 on Build GRCh38
rs867146249 Chr.3: 184380451-184380451 on Build GRCh38
rs762749709 Chr.3: 184380741-184380741 on Build GRCh38
rs911954704 Chr.3: 184380635-184380635 on Build GRCh38
rs763528309 Chr.3: 184380851-184380851 on Build GRCh38
rs774076650 Chr.3: 184380828-184380828 on Build GRCh38
rs747385357 Chr.3: 184380547-184380547 on Build GRCh38

View Full Product Details