Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184381154-184381655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRD
Entrez Gene ID
8646
Gene Name
chordin
Gene Aliases
-
UniGene
Hs.166186
Gene Chromosome Location
Chr.3: 184380073-184389835 on Build GRCh38
Transcripts
NM_001304472.1 NM_001304473.1 NM_001304474.1 NM_003741.3 NR_130747.1 XM_011513254.1 XM_011513256.1 XM_017007388.1 XM_017007389.1 XM_017007390.1 XM_017007391.1 XM_017007392.1 XM_017007393.1 XM_017007394.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3945233 c.472C>A p.P158T Chr.3: 184381585-184381585 on Build GRCh38
6628588 c.408G>A p.P136P Chr.3: 184381521-184381521 on Build GRCh38
1421376 c.284G>A p.G95D Chr.3: 184381266-184381266 on Build GRCh38
7314099 c.348G>T p.Q116H Chr.3: 184381330-184381330 on Build GRCh38
127096 c.511+1G>A p.? Chr.3: 184381625-184381625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776748999 Chr.3: 184381514-184381514 on Build GRCh38
rs767859402 Chr.3: 184381242-184381242 on Build GRCh38
rs533637821 Chr.3: 184381383-184381383 on Build GRCh38
rs572984824 Chr.3: 184381499-184381499 on Build GRCh38
rs1005012756 Chr.3: 184381441-184381441 on Build GRCh38
rs3749228 Chr.3: 184381554-184381554 on Build GRCh38
rs113468239 Chr.3: 184381193-184381193 on Build GRCh38
rs775144152 Chr.3: 184381308-184381308 on Build GRCh38
rs35929225 Chr.3: 184381590-184381590 on Build GRCh38
rs1010020967 Chr.3: 184381579-184381579 on Build GRCh38
rs748373553 Chr.3: 184381589-184381589 on Build GRCh38
rs777487132 Chr.3: 184381574-184381574 on Build GRCh38
rs781112267 Chr.3: 184381448-184381448 on Build GRCh38
rs746958838 Chr.3: 184381240-184381240 on Build GRCh38
rs776406416 Chr.3: 184381339-184381339 on Build GRCh38
rs567035947 Chr.3: 184381415-184381415 on Build GRCh38
rs771653468 Chr.3: 184381305-184381305 on Build GRCh38
rs1009906844 Chr.3: 184381375-184381375 on Build GRCh38
rs974259217 Chr.3: 184381436-184381436 on Build GRCh38
rs923801669 Chr.3: 184381226-184381226 on Build GRCh38
rs753000064 Chr.3: 184381380-184381380 on Build GRCh38
rs766987083 Chr.3: 184381624-184381624 on Build GRCh38
rs772319456 Chr.3: 184381470-184381470 on Build GRCh38
rs145485926 Chr.3: 184381405-184381405 on Build GRCh38
rs773495582 Chr.3: 184381596-184381596 on Build GRCh38
rs373792812 Chr.3: 184381523-184381523 on Build GRCh38
rs760850880 Chr.3: 184381512-184381512 on Build GRCh38
rs752381713 Chr.3: 184381628-184381628 on Build GRCh38
rs373375780 Chr.3: 184381409-184381409 on Build GRCh38
rs138217536 Chr.3: 184381293-184381293 on Build GRCh38
rs763515621 Chr.3: 184381536-184381536 on Build GRCh38
rs376941518 Chr.3: 184381377-184381377 on Build GRCh38
rs766879394 Chr.3: 184381544-184381544 on Build GRCh38
rs754487046 Chr.3: 184381257-184381264 on Build GRCh38
rs759057662 Chr.3: 184381622-184381622 on Build GRCh38
rs79103946 Chr.3: 184381206-184381206 on Build GRCh38
rs201383201 Chr.3: 184381569-184381569 on Build GRCh38
rs778386345 Chr.3: 184381248-184381248 on Build GRCh38
rs368192345 Chr.3: 184381205-184381205 on Build GRCh38
rs538246209 Chr.3: 184381456-184381456 on Build GRCh38
rs149400260 Chr.3: 184381483-184381484 on Build GRCh38
rs755790440 Chr.3: 184381629-184381629 on Build GRCh38
rs774872526 Chr.3: 184381621-184381621 on Build GRCh38
rs777956202 Chr.3: 184381570-184381570 on Build GRCh38
rs1050852388 Chr.3: 184381631-184381631 on Build GRCh38
rs762907259 Chr.3: 184381211-184381211 on Build GRCh38
rs766825882 Chr.3: 184381616-184381616 on Build GRCh38
rs760475645 Chr.3: 184381204-184381204 on Build GRCh38
rs192063183 Chr.3: 184381576-184381576 on Build GRCh38
rs199868165 Chr.3: 184381218-184381218 on Build GRCh38
rs754172359 Chr.3: 184381484-184381484 on Build GRCh38
rs373789484 Chr.3: 184381585-184381585 on Build GRCh38
rs779015930 Chr.3: 184381582-184381582 on Build GRCh38
rs1032783409 Chr.3: 184381323-184381323 on Build GRCh38
rs766368325 Chr.3: 184381355-184381355 on Build GRCh38
rs759551290 Chr.3: 184381368-184381368 on Build GRCh38
rs992004711 Chr.3: 184381500-184381500 on Build GRCh38
rs771758431 Chr.3: 184381188-184381188 on Build GRCh38
rs373534825 Chr.3: 184381487-184381488 on Build GRCh38
rs749287992 Chr.3: 184381504-184381504 on Build GRCh38
rs201984321 Chr.3: 184381335-184381335 on Build GRCh38
rs749842503 Chr.3: 184381250-184381250 on Build GRCh38
rs918721378 Chr.3: 184381446-184381446 on Build GRCh38
rs746709131 Chr.3: 184381411-184381411 on Build GRCh38
rs34095724 Chr.3: 184381262-184381262 on Build GRCh38
rs377128785 Chr.3: 184381388-184381388 on Build GRCh38
rs780725085 Chr.3: 184381487-184381487 on Build GRCh38
rs765566662 Chr.3: 184381524-184381524 on Build GRCh38
rs751722209 Chr.3: 184381224-184381224 on Build GRCh38
rs779299629 Chr.3: 184381389-184381389 on Build GRCh38
rs544184044 Chr.3: 184381360-184381360 on Build GRCh38
rs754508907 Chr.3: 184381552-184381552 on Build GRCh38
rs73053787 Chr.3: 184381481-184381481 on Build GRCh38
rs968181969 Chr.3: 184381178-184381178 on Build GRCh38
rs185841327 Chr.3: 184381419-184381419 on Build GRCh38
rs540307891 Chr.3: 184381309-184381309 on Build GRCh38
rs201020400 Chr.3: 184381340-184381340 on Build GRCh38
rs376181413 Chr.3: 184381176-184381176 on Build GRCh38
rs754295476 Chr.3: 184381386-184381386 on Build GRCh38
rs149561137 Chr.3: 184381610-184381610 on Build GRCh38
rs745592395 Chr.3: 184381186-184381186 on Build GRCh38
rs776887601 Chr.3: 184381586-184381586 on Build GRCh38
rs567606357 Chr.3: 184381420-184381420 on Build GRCh38
rs947681671 Chr.3: 184381348-184381348 on Build GRCh38
rs753056932 Chr.3: 184381244-184381244 on Build GRCh38
rs372908853 Chr.3: 184381331-184381331 on Build GRCh38
rs769509825 Chr.3: 184381488-184381488 on Build GRCh38
rs775833174 Chr.3: 184381473-184381473 on Build GRCh38
rs756571427 Chr.3: 184381247-184381247 on Build GRCh38
rs773048521 Chr.3: 184381457-184381457 on Build GRCh38
rs539327743 Chr.3: 184381485-184381485 on Build GRCh38
rs746244283 Chr.3: 184381533-184381535 on Build GRCh38
rs533718758 Chr.3: 184381480-184381481 on Build GRCh38
rs775197496 Chr.3: 184381201-184381201 on Build GRCh38
rs747755283 Chr.3: 184381481-184381481 on Build GRCh38
rs202122376 Chr.3: 184381486-184381486 on Build GRCh38
rs745620244 Chr.3: 184381561-184381562 on Build GRCh38
rs376748609 Chr.3: 184381546-184381546 on Build GRCh38
rs755298701 Chr.3: 184381234-184381234 on Build GRCh38
rs772266252 Chr.3: 184381509-184381509 on Build GRCh38
rs750912862 Chr.3: 184381531-184381531 on Build GRCh38
rs3749229 Chr.3: 184381418-184381418 on Build GRCh38
rs768871806 Chr.3: 184381513-184381513 on Build GRCh38
rs980833271 Chr.3: 184381520-184381520 on Build GRCh38
rs780735490 Chr.3: 184381301-184381301 on Build GRCh38
rs1017918330 Chr.3: 184381630-184381630 on Build GRCh38
rs757202403 Chr.3: 184381581-184381581 on Build GRCh38
rs200272021 Chr.3: 184381458-184381458 on Build GRCh38
rs747043337 Chr.3: 184381584-184381584 on Build GRCh38
rs35087408 Chr.3: 184381510-184381510 on Build GRCh38
rs745331545 Chr.3: 184381401-184381401 on Build GRCh38
rs760225685 Chr.3: 184381296-184381296 on Build GRCh38
rs764601926 Chr.3: 184381385-184381385 on Build GRCh38
rs745373807 Chr.3: 184381297-184381297 on Build GRCh38

View Full Product Details