Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184576400-184576913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB3
Entrez Gene ID
2049
Gene Name
EPH receptor B3
Gene Aliases
EK2, ETK2, HEK2, TYRO6
UniGene
Hs.2913
Gene Chromosome Location
Chr.3: 184561799-184582408 on Build GRCh38
Transcripts
NM_004443.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3767341 c.1049T>C p.V350A Chr.3: 184576878-184576878 on Build GRCh38
1421389 c.1084C>T p.P362S Chr.3: 184576913-184576913 on Build GRCh38
5995246 c.1030C>T p.R344* Chr.3: 184576859-184576859 on Build GRCh38
730191 c.1072G>A p.E358K Chr.3: 184576901-184576901 on Build GRCh38
4115806 c.1073A>C p.E358A Chr.3: 184576902-184576902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776902371 Chr.3: 184576821-184576821 on Build GRCh38
rs1043684395 Chr.3: 184576726-184576726 on Build GRCh38
rs902535627 Chr.3: 184576761-184576761 on Build GRCh38
rs900408452 Chr.3: 184576539-184576539 on Build GRCh38
rs868057603 Chr.3: 184576769-184576769 on Build GRCh38
rs372575808 Chr.3: 184576859-184576859 on Build GRCh38
rs764300784 Chr.3: 184576818-184576818 on Build GRCh38
rs1034769281 Chr.3: 184576663-184576663 on Build GRCh38
rs549588186 Chr.3: 184576687-184576687 on Build GRCh38
rs1023326163 Chr.3: 184576759-184576759 on Build GRCh38
rs960474934 Chr.3: 184576665-184576665 on Build GRCh38
rs961765476 Chr.3: 184576568-184576568 on Build GRCh38
rs190816816 Chr.3: 184576467-184576467 on Build GRCh38
rs1040417624 Chr.3: 184576786-184576786 on Build GRCh38
rs973956398 Chr.3: 184576433-184576433 on Build GRCh38
rs981198925 Chr.3: 184576584-184576584 on Build GRCh38
rs758560752 Chr.3: 184576506-184576506 on Build GRCh38
rs947309868 Chr.3: 184576712-184576712 on Build GRCh38
rs780837073 Chr.3: 184576873-184576873 on Build GRCh38
rs560735975 Chr.3: 184576560-184576560 on Build GRCh38
rs1047811625 Chr.3: 184576458-184576458 on Build GRCh38
rs999807147 Chr.3: 184576616-184576616 on Build GRCh38
rs771463060 Chr.3: 184576601-184576601 on Build GRCh38
rs74553333 Chr.3: 184576711-184576711 on Build GRCh38
rs182950285 Chr.3: 184576766-184576766 on Build GRCh38
rs112508507 Chr.3: 184576817-184576817 on Build GRCh38
rs537970978 Chr.3: 184576443-184576443 on Build GRCh38
rs572777020 Chr.3: 184576548-184576548 on Build GRCh38
rs531405354 Chr.3: 184576561-184576561 on Build GRCh38
rs766853413 Chr.3: 184576852-184576852 on Build GRCh38
rs574038132 Chr.3: 184576440-184576440 on Build GRCh38
rs991373593 Chr.3: 184576707-184576707 on Build GRCh38
rs80234080 Chr.3: 184576459-184576459 on Build GRCh38
rs763398988 Chr.3: 184576850-184576850 on Build GRCh38
rs775452153 Chr.3: 184576807-184576807 on Build GRCh38
rs778200335 Chr.3: 184576530-184576530 on Build GRCh38
rs911963053 Chr.3: 184576799-184576799 on Build GRCh38
rs928429838 Chr.3: 184576621-184576621 on Build GRCh38
rs767285345 Chr.3: 184576469-184576469 on Build GRCh38
rs111611525 Chr.3: 184576552-184576552 on Build GRCh38
rs762008751 Chr.3: 184576830-184576830 on Build GRCh38
rs901995191 Chr.3: 184576447-184576447 on Build GRCh38
rs148453943 Chr.3: 184576534-184576534 on Build GRCh38
rs545654539 Chr.3: 184576486-184576486 on Build GRCh38
rs937171123 Chr.3: 184576634-184576634 on Build GRCh38
rs201146779 Chr.3: 184576843-184576843 on Build GRCh38
rs149247223 Chr.3: 184576844-184576844 on Build GRCh38
rs563635301 Chr.3: 184576422-184576422 on Build GRCh38
rs902349669 Chr.3: 184576609-184576609 on Build GRCh38
rs920721459 Chr.3: 184576780-184576780 on Build GRCh38
rs562993499 Chr.3: 184576860-184576860 on Build GRCh38
rs950911007 Chr.3: 184576781-184576781 on Build GRCh38

View Full Product Details