Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 185218128-185218676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHHADH
Entrez Gene ID
1962
Gene Name
enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase
Gene Aliases
ECHD, FRTS3, L-PBE, LBFP, LBP, PBFE
UniGene
Hs.429879
Gene Chromosome Location
Chr.3: 185190624-185282886 on Build GRCh38
Transcripts
NM_001166415.1 NM_001966.3 XM_006713525.1 XM_011512517.1 XM_017005839.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4834701 c.556C>T p.Q186* Chr.3: 185218148-185218148 on Build GRCh38
4115858 c.502A>G p.I168V Chr.3: 185218202-185218202 on Build GRCh38
1042174 c.496C>T p.L166L Chr.3: 185218208-185218208 on Build GRCh38
6652459 c.532G>T p.V178F Chr.3: 185218172-185218172 on Build GRCh38
3590861 c.464G>A p.G155E Chr.3: 185218240-185218240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770272693 Chr.3: 185218188-185218188 on Build GRCh38
rs1018639616 Chr.3: 185218565-185218565 on Build GRCh38
rs148411037 Chr.3: 185218173-185218173 on Build GRCh38
rs748785259 Chr.3: 185218202-185218202 on Build GRCh38
rs1016018121 Chr.3: 185218597-185218597 on Build GRCh38
rs1011451986 Chr.3: 185218458-185218458 on Build GRCh38
rs551812065 Chr.3: 185218630-185218630 on Build GRCh38
rs985576203 Chr.3: 185218349-185218349 on Build GRCh38
rs1022052890 Chr.3: 185218469-185218469 on Build GRCh38
rs895354671 Chr.3: 185218429-185218429 on Build GRCh38
rs764049343 Chr.3: 185218244-185218244 on Build GRCh38
rs1054935039 Chr.3: 185218551-185218551 on Build GRCh38
rs199809736 Chr.3: 185218528-185218528 on Build GRCh38
rs760532670 Chr.3: 185218234-185218234 on Build GRCh38
rs995465914 Chr.3: 185218632-185218632 on Build GRCh38
rs750488107 Chr.3: 185218247-185218247 on Build GRCh38
rs9852812 Chr.3: 185218631-185218631 on Build GRCh38
rs781694865 Chr.3: 185218285-185218285 on Build GRCh38
rs562578928 Chr.3: 185218470-185218470 on Build GRCh38
rs976173975 Chr.3: 185218422-185218422 on Build GRCh38
rs755198066 Chr.3: 185218282-185218282 on Build GRCh38
rs937854280 Chr.3: 185218548-185218548 on Build GRCh38
rs529394652 Chr.3: 185218416-185218416 on Build GRCh38
rs149820014 Chr.3: 185218208-185218208 on Build GRCh38
rs944276533 Chr.3: 185218395-185218395 on Build GRCh38
rs775296210 Chr.3: 185218227-185218227 on Build GRCh38
rs912835676 Chr.3: 185218385-185218385 on Build GRCh38
rs561850049 Chr.3: 185218318-185218318 on Build GRCh38
rs999215218 Chr.3: 185218543-185218543 on Build GRCh38
rs781519143 Chr.3: 185218261-185218263 on Build GRCh38
rs774034501 Chr.3: 185218209-185218209 on Build GRCh38
rs974694206 Chr.3: 185218619-185218619 on Build GRCh38
rs533657502 Chr.3: 185218480-185218480 on Build GRCh38
rs771912925 Chr.3: 185218219-185218219 on Build GRCh38
rs544192756 Chr.3: 185218425-185218425 on Build GRCh38
rs369360134 Chr.3: 185218268-185218268 on Build GRCh38
rs1027052037 Chr.3: 185218646-185218646 on Build GRCh38
rs903585434 Chr.3: 185218501-185218501 on Build GRCh38
rs781683788 Chr.3: 185218174-185218174 on Build GRCh38
rs758834472 Chr.3: 185218249-185218251 on Build GRCh38
rs572168718 Chr.3: 185218218-185218218 on Build GRCh38
rs753740694 Chr.3: 185218596-185218596 on Build GRCh38
rs965672761 Chr.3: 185218605-185218605 on Build GRCh38
rs763137355 Chr.3: 185218252-185218252 on Build GRCh38
rs777180874 Chr.3: 185218196-185218196 on Build GRCh38
rs376317448 Chr.3: 185218263-185218263 on Build GRCh38
rs868468862 Chr.3: 185218172-185218172 on Build GRCh38
rs55956553 Chr.3: 185218249-185218249 on Build GRCh38
rs751806587 Chr.3: 185218151-185218151 on Build GRCh38
rs1013464281 Chr.3: 185218578-185218578 on Build GRCh38

View Full Product Details