Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186188119-186188618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKG
Entrez Gene ID
1608
Gene Name
diacylglycerol kinase gamma
Gene Aliases
DAGK3, DGK-GAMMA
UniGene
Hs.683449
Gene Chromosome Location
Chr.3: 186147201-186362234 on Build GRCh38
Transcripts
NM_001080744.1 NM_001080745.1 NM_001346.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3945257 c.1961G>A p.G654D Chr.3: 186188336-186188336 on Build GRCh38
4848023 c.2025G>C p.K675N Chr.3: 186188272-186188272 on Build GRCh38
5864091 c.2048G>A p.S683N Chr.3: 186188249-186188249 on Build GRCh38
3332211 c.2006T>A p.L669H Chr.3: 186188291-186188291 on Build GRCh38
7133475 c.2088C>T p.C696C Chr.3: 186188209-186188209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960287811 Chr.3: 186188491-186188491 on Build GRCh38
rs761818632 Chr.3: 186188401-186188401 on Build GRCh38
rs141461004 Chr.3: 186188389-186188389 on Build GRCh38
rs1038908811 Chr.3: 186188309-186188309 on Build GRCh38
rs562868830 Chr.3: 186188454-186188454 on Build GRCh38
rs766731251 Chr.3: 186188421-186188421 on Build GRCh38
rs771858343 Chr.3: 186188392-186188392 on Build GRCh38
rs187896616 Chr.3: 186188143-186188143 on Build GRCh38
rs979191438 Chr.3: 186188548-186188548 on Build GRCh38
rs766370553 Chr.3: 186188171-186188171 on Build GRCh38
rs763352937 Chr.3: 186188419-186188419 on Build GRCh38
rs531462977 Chr.3: 186188160-186188160 on Build GRCh38
rs113020411 Chr.3: 186188388-186188388 on Build GRCh38
rs747611243 Chr.3: 186188343-186188343 on Build GRCh38
rs377679950 Chr.3: 186188172-186188172 on Build GRCh38
rs3217178 Chr.3: 186188458-186188461 on Build GRCh38
rs750627171 Chr.3: 186188415-186188415 on Build GRCh38
rs762238205 Chr.3: 186188460-186188461 on Build GRCh38
rs992606030 Chr.3: 186188435-186188436 on Build GRCh38
rs142953458 Chr.3: 186188267-186188267 on Build GRCh38
rs903675867 Chr.3: 186188292-186188292 on Build GRCh38
rs879059977 Chr.3: 186188461-186188462 on Build GRCh38
rs763013531 Chr.3: 186188170-186188170 on Build GRCh38
rs765514375 Chr.3: 186188408-186188408 on Build GRCh38
rs879035686 Chr.3: 186188507-186188507 on Build GRCh38
rs34284245 Chr.3: 186188278-186188278 on Build GRCh38
rs759551064 Chr.3: 186188391-186188391 on Build GRCh38
rs773320582 Chr.3: 186188158-186188158 on Build GRCh38
rs375480424 Chr.3: 186188246-186188246 on Build GRCh38
rs746564333 Chr.3: 186188230-186188230 on Build GRCh38
rs757659817 Chr.3: 186188192-186188192 on Build GRCh38
rs139876309 Chr.3: 186188310-186188310 on Build GRCh38
rs750998877 Chr.3: 186188380-186188380 on Build GRCh38
rs749153792 Chr.3: 186188385-186188385 on Build GRCh38
rs747526080 Chr.3: 186188333-186188333 on Build GRCh38
rs372919637 Chr.3: 186188409-186188409 on Build GRCh38
rs760717232 Chr.3: 186188294-186188294 on Build GRCh38
rs201791341 Chr.3: 186188305-186188305 on Build GRCh38
rs768195151 Chr.3: 186188238-186188238 on Build GRCh38
rs1050640548 Chr.3: 186188542-186188542 on Build GRCh38
rs1016792662 Chr.3: 186188457-186188457 on Build GRCh38
rs917051919 Chr.3: 186188445-186188445 on Build GRCh38
rs1040580197 Chr.3: 186188586-186188586 on Build GRCh38
rs780783240 Chr.3: 186188336-186188336 on Build GRCh38
rs769584231 Chr.3: 186188353-186188353 on Build GRCh38
rs201532965 Chr.3: 186188461-186188461 on Build GRCh38
rs566926961 Chr.3: 186188544-186188544 on Build GRCh38
rs758754601 Chr.3: 186188321-186188321 on Build GRCh38
rs1035915913 Chr.3: 186188462-186188462 on Build GRCh38
rs199878379 Chr.3: 186188208-186188208 on Build GRCh38
rs371007519 Chr.3: 186188174-186188174 on Build GRCh38
rs73178197 Chr.3: 186188328-186188328 on Build GRCh38
rs776192898 Chr.3: 186188242-186188242 on Build GRCh38
rs544713321 Chr.3: 186188249-186188249 on Build GRCh38
rs527785021 Chr.3: 186188545-186188545 on Build GRCh38
rs751828559 Chr.3: 186188431-186188431 on Build GRCh38
rs778488993 Chr.3: 186188425-186188425 on Build GRCh38
rs59681256 Chr.3: 186188471-186188472 on Build GRCh38
rs913567273 Chr.3: 186188532-186188532 on Build GRCh38
rs200697757 Chr.3: 186188453-186188453 on Build GRCh38
rs764281620 Chr.3: 186188178-186188178 on Build GRCh38
rs779153569 Chr.3: 186188209-186188209 on Build GRCh38
rs200480631 Chr.3: 186188304-186188304 on Build GRCh38
rs754483425 Chr.3: 186188335-186188335 on Build GRCh38
rs56134684 Chr.3: 186188468-186188471 on Build GRCh38
rs776788995 Chr.3: 186188397-186188397 on Build GRCh38
rs985328849 Chr.3: 186188140-186188140 on Build GRCh38
rs754070147 Chr.3: 186188184-186188184 on Build GRCh38
rs56840967 Chr.3: 186188458-186188458 on Build GRCh38
rs765388941 Chr.3: 186188311-186188311 on Build GRCh38
rs888851224 Chr.3: 186188567-186188567 on Build GRCh38
rs561957681 Chr.3: 186188371-186188371 on Build GRCh38
rs750796251 Chr.3: 186188316-186188316 on Build GRCh38
rs779284601 Chr.3: 186188202-186188202 on Build GRCh38
rs551498564 Chr.3: 186188515-186188515 on Build GRCh38
rs16860539 Chr.3: 186188570-186188570 on Build GRCh38
rs200595122 Chr.3: 186188325-186188325 on Build GRCh38
rs545594405 Chr.3: 186188590-186188590 on Build GRCh38
rs139820498 Chr.3: 186188237-186188237 on Build GRCh38
rs769530605 Chr.3: 186188255-186188255 on Build GRCh38

View Full Product Details