Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186211711-186212212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKG
Entrez Gene ID
1608
Gene Name
diacylglycerol kinase gamma
Gene Aliases
DAGK3, DGK-GAMMA
UniGene
Hs.683449
Gene Chromosome Location
Chr.3: 186147201-186362234 on Build GRCh38
Transcripts
NM_001080744.1 NM_001080745.1 NM_001346.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3849631 c.1900G>A p.D634N Chr.3: 186211812-186211812 on Build GRCh38
6990536 c.1914G>C p.L638F Chr.3: 186211798-186211798 on Build GRCh38
1421483 c.1833G>T p.K611N Chr.3: 186211879-186211879 on Build GRCh38
5398806 c.1848C>T p.Y616Y Chr.3: 186211864-186211864 on Build GRCh38
5809291 c.1854A>G p.E618E Chr.3: 186211858-186211858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549881049 Chr.3: 186212100-186212100 on Build GRCh38
rs75943036 Chr.3: 186212071-186212071 on Build GRCh38
rs61752077 Chr.3: 186211795-186211795 on Build GRCh38
rs200349299 Chr.3: 186211813-186211813 on Build GRCh38
rs766460411 Chr.3: 186211800-186211800 on Build GRCh38
rs574782806 Chr.3: 186211884-186211884 on Build GRCh38
rs780012300 Chr.3: 186211872-186211872 on Build GRCh38
rs1038636707 Chr.3: 186212060-186212060 on Build GRCh38
rs745552430 Chr.3: 186211776-186211777 on Build GRCh38
rs574596649 Chr.3: 186212100-186212100 on Build GRCh38
rs759751438 Chr.3: 186211806-186211806 on Build GRCh38
rs542163508 Chr.3: 186211918-186211918 on Build GRCh38
rs751965324 Chr.3: 186211805-186211805 on Build GRCh38
rs761376942 Chr.3: 186212088-186212088 on Build GRCh38
rs543861928 Chr.3: 186211986-186211986 on Build GRCh38
rs201258836 Chr.3: 186211757-186211757 on Build GRCh38
rs770451943 Chr.3: 186211756-186211756 on Build GRCh38
rs1002188331 Chr.3: 186211752-186211752 on Build GRCh38
rs775406036 Chr.3: 186211765-186211765 on Build GRCh38
rs903693527 Chr.3: 186211973-186211973 on Build GRCh38
rs781553784 Chr.3: 186211900-186211901 on Build GRCh38
rs530849235 Chr.3: 186211946-186211946 on Build GRCh38
rs763641014 Chr.3: 186212063-186212063 on Build GRCh38
rs567256712 Chr.3: 186212075-186212075 on Build GRCh38
rs1042468450 Chr.3: 186212152-186212152 on Build GRCh38
rs768537842 Chr.3: 186211889-186211889 on Build GRCh38
rs939860735 Chr.3: 186212013-186212013 on Build GRCh38
rs563321602 Chr.3: 186211921-186211921 on Build GRCh38
rs368496064 Chr.3: 186211902-186211902 on Build GRCh38
rs773460746 Chr.3: 186211783-186211783 on Build GRCh38
rs891908823 Chr.3: 186211955-186211955 on Build GRCh38
rs745594053 Chr.3: 186211759-186211759 on Build GRCh38
rs920942740 Chr.3: 186211958-186211958 on Build GRCh38
rs1053094437 Chr.3: 186211882-186211882 on Build GRCh38
rs908335930 Chr.3: 186211993-186211993 on Build GRCh38
rs777457614 Chr.3: 186211831-186211831 on Build GRCh38
rs528230251 Chr.3: 186212133-186212133 on Build GRCh38
rs973854190 Chr.3: 186212175-186212175 on Build GRCh38
rs547522584 Chr.3: 186211846-186211846 on Build GRCh38
rs772054884 Chr.3: 186211763-186211763 on Build GRCh38
rs749266684 Chr.3: 186211891-186211891 on Build GRCh38
rs768946569 Chr.3: 186211841-186211841 on Build GRCh38
rs145474986 Chr.3: 186211870-186211870 on Build GRCh38
rs769838562 Chr.3: 186211782-186211782 on Build GRCh38
rs368357864 Chr.3: 186211772-186211772 on Build GRCh38
rs760002403 Chr.3: 186211908-186211908 on Build GRCh38
rs745746020 Chr.3: 186211852-186211852 on Build GRCh38
rs767188118 Chr.3: 186212097-186212097 on Build GRCh38
rs896491505 Chr.3: 186212093-186212093 on Build GRCh38
rs755725819 Chr.3: 186211829-186211829 on Build GRCh38
rs552357282 Chr.3: 186211961-186211961 on Build GRCh38
rs748919383 Chr.3: 186211745-186211745 on Build GRCh38
rs753351238 Chr.3: 186211841-186211841 on Build GRCh38
rs147898553 Chr.3: 186211812-186211812 on Build GRCh38
rs771188241 Chr.3: 186211893-186211893 on Build GRCh38
rs141678179 Chr.3: 186211847-186211847 on Build GRCh38
rs546785293 Chr.3: 186212179-186212179 on Build GRCh38

View Full Product Details