Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186242418-186242923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKG
Entrez Gene ID
1608
Gene Name
diacylglycerol kinase gamma
Gene Aliases
DAGK3, DGK-GAMMA
UniGene
Hs.683449
Gene Chromosome Location
Chr.3: 186147201-186362234 on Build GRCh38
Transcripts
NM_001080744.1 NM_001080745.1 NM_001346.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
730114 c.1807C>T p.P603S Chr.3: 186242523-186242523 on Build GRCh38
6376448 c.1819A>G p.N607D Chr.3: 186242511-186242511 on Build GRCh38
3992926 c.1795A>T p.R599* Chr.3: 186242535-186242535 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs530375767 Chr.3: 186242759-186242759 on Build GRCh38
rs764617077 Chr.3: 186242565-186242565 on Build GRCh38
rs565129474 Chr.3: 186242739-186242739 on Build GRCh38
rs548234201 Chr.3: 186242887-186242887 on Build GRCh38
rs779852865 Chr.3: 186242479-186242479 on Build GRCh38
rs1009983404 Chr.3: 186242654-186242654 on Build GRCh38
rs112673440 Chr.3: 186242829-186242829 on Build GRCh38
rs115720177 Chr.3: 186242823-186242823 on Build GRCh38
rs746109049 Chr.3: 186242494-186242494 on Build GRCh38
rs986028227 Chr.3: 186242651-186242651 on Build GRCh38
rs1036535657 Chr.3: 186242593-186242593 on Build GRCh38
rs745986075 Chr.3: 186242707-186242707 on Build GRCh38
rs762476788 Chr.3: 186242667-186242667 on Build GRCh38
rs954261878 Chr.3: 186242845-186242845 on Build GRCh38
rs147659054 Chr.3: 186242554-186242554 on Build GRCh38
rs776994759 Chr.3: 186242557-186242557 on Build GRCh38
rs944236017 Chr.3: 186242685-186242685 on Build GRCh38
rs1036649903 Chr.3: 186242808-186242808 on Build GRCh38
rs368971279 Chr.3: 186242516-186242516 on Build GRCh38
rs138600195 Chr.3: 186242863-186242863 on Build GRCh38
rs202135902 Chr.3: 186242585-186242585 on Build GRCh38
rs978465903 Chr.3: 186242690-186242690 on Build GRCh38
rs377731765 Chr.3: 186242558-186242558 on Build GRCh38
rs1022838126 Chr.3: 186242552-186242552 on Build GRCh38
rs375971542 Chr.3: 186242507-186242507 on Build GRCh38
rs956060629 Chr.3: 186242578-186242578 on Build GRCh38
rs907049347 Chr.3: 186242763-186242763 on Build GRCh38
rs764640424 Chr.3: 186242455-186242455 on Build GRCh38
rs1020069547 Chr.3: 186242676-186242679 on Build GRCh38
rs758249775 Chr.3: 186242473-186242473 on Build GRCh38
rs928942638 Chr.3: 186242698-186242698 on Build GRCh38
rs955982116 Chr.3: 186242827-186242827 on Build GRCh38
rs753087425 Chr.3: 186242652-186242652 on Build GRCh38
rs1037289046 Chr.3: 186242686-186242686 on Build GRCh38
rs972950963 Chr.3: 186242892-186242892 on Build GRCh38
rs376608659 Chr.3: 186242624-186242624 on Build GRCh38
rs927084006 Chr.3: 186242703-186242703 on Build GRCh38
rs1040375458 Chr.3: 186242898-186242898 on Build GRCh38
rs753406073 Chr.3: 186242460-186242460 on Build GRCh38
rs6778456 Chr.3: 186242882-186242882 on Build GRCh38
rs149141346 Chr.3: 186242566-186242566 on Build GRCh38
rs373453872 Chr.3: 186242638-186242638 on Build GRCh38
rs375484830 Chr.3: 186242637-186242637 on Build GRCh38
rs774674542 Chr.3: 186242491-186242491 on Build GRCh38
rs754762756 Chr.3: 186242612-186242612 on Build GRCh38
rs765956264 Chr.3: 186242602-186242602 on Build GRCh38
rs748607165 Chr.3: 186242586-186242586 on Build GRCh38
rs370262411 Chr.3: 186242711-186242711 on Build GRCh38
rs1012703914 Chr.3: 186242717-186242717 on Build GRCh38
rs750026740 Chr.3: 186242471-186242471 on Build GRCh38
rs371642085 Chr.3: 186242506-186242506 on Build GRCh38
rs756720940 Chr.3: 186242469-186242469 on Build GRCh38
rs115846554 Chr.3: 186242489-186242489 on Build GRCh38
rs866324848 Chr.3: 186242889-186242889 on Build GRCh38
rs371929344 Chr.3: 186242454-186242454 on Build GRCh38
rs748013473 Chr.3: 186242618-186242618 on Build GRCh38
rs754921068 Chr.3: 186242483-186242483 on Build GRCh38
rs899370697 Chr.3: 186242605-186242605 on Build GRCh38
rs958588552 Chr.3: 186242712-186242712 on Build GRCh38
rs746755811 Chr.3: 186242482-186242482 on Build GRCh38
rs992515775 Chr.3: 186242732-186242732 on Build GRCh38
rs12489063 Chr.3: 186242625-186242625 on Build GRCh38
rs568891773 Chr.3: 186242748-186242748 on Build GRCh38
rs1021664984 Chr.3: 186242834-186242834 on Build GRCh38
rs781357060 Chr.3: 186242688-186242688 on Build GRCh38
rs993206654 Chr.3: 186242487-186242487 on Build GRCh38
rs200822478 Chr.3: 186242488-186242488 on Build GRCh38
rs781164029 Chr.3: 186242616-186242616 on Build GRCh38
rs765001794 Chr.3: 186242470-186242470 on Build GRCh38
rs1002724076 Chr.3: 186242766-186242766 on Build GRCh38
rs761056028 Chr.3: 186242537-186242537 on Build GRCh38
rs201290278 Chr.3: 186242611-186242611 on Build GRCh38
rs552156637 Chr.3: 186242532-186242532 on Build GRCh38
rs373057140 Chr.3: 186242542-186242542 on Build GRCh38

View Full Product Details