Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 190309986-190310533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN1
Entrez Gene ID
9076
Gene Name
claudin 1
Gene Aliases
CLD1, ILVASC, SEMP1
UniGene
Hs.439060
Gene Chromosome Location
Chr.3: 190305701-190322446 on Build GRCh38
Transcripts
NM_021101.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5647395 c.436C>T p.Q146* Chr.3: 190310206-190310206 on Build GRCh38
3333178 c.428G>T p.R143I Chr.3: 190310214-190310214 on Build GRCh38
1042517 c.470C>T p.A157V Chr.3: 190310172-190310172 on Build GRCh38
3333177 c.432C>T p.I144I Chr.3: 190310210-190310210 on Build GRCh38
1670779 c.439G>T p.E147* Chr.3: 190310203-190310203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963586466 Chr.3: 190310313-190310313 on Build GRCh38
rs923677415 Chr.3: 190310386-190310386 on Build GRCh38
rs955005107 Chr.3: 190310376-190310376 on Build GRCh38
rs924768475 Chr.3: 190310426-190310426 on Build GRCh38
rs368329961 Chr.3: 190310168-190310168 on Build GRCh38
rs773376637 Chr.3: 190310239-190310239 on Build GRCh38
rs531415931 Chr.3: 190310381-190310381 on Build GRCh38
rs769700524 Chr.3: 190310228-190310228 on Build GRCh38
rs1045359990 Chr.3: 190310470-190310470 on Build GRCh38
rs973311008 Chr.3: 190310352-190310352 on Build GRCh38
rs374333339 Chr.3: 190310126-190310126 on Build GRCh38
rs1036141994 Chr.3: 190310279-190310279 on Build GRCh38
rs781556791 Chr.3: 190310210-190310210 on Build GRCh38
rs1027977444 Chr.3: 190310355-190310355 on Build GRCh38
rs774898897 Chr.3: 190310275-190310275 on Build GRCh38
rs1051064965 Chr.3: 190310469-190310469 on Build GRCh38
rs377184401 Chr.3: 190310164-190310164 on Build GRCh38
rs753668517 Chr.3: 190310181-190310181 on Build GRCh38
rs536268074 Chr.3: 190310421-190310421 on Build GRCh38
rs932668013 Chr.3: 190310447-190310447 on Build GRCh38
rs749371682 Chr.3: 190310272-190310272 on Build GRCh38
rs367552459 Chr.3: 190310152-190310152 on Build GRCh38
rs748402994 Chr.3: 190310211-190310211 on Build GRCh38
rs753356703 Chr.3: 190310297-190310297 on Build GRCh38
rs114644300 Chr.3: 190310303-190310303 on Build GRCh38
rs766303808 Chr.3: 190310270-190310271 on Build GRCh38
rs150740353 Chr.3: 190310432-190310432 on Build GRCh38
rs766693390 Chr.3: 190310140-190310140 on Build GRCh38
rs891202999 Chr.3: 190310227-190310227 on Build GRCh38
rs745998030 Chr.3: 190310301-190310301 on Build GRCh38
rs755513283 Chr.3: 190310173-190310173 on Build GRCh38
rs939176742 Chr.3: 190310491-190310491 on Build GRCh38
rs774948865 Chr.3: 190310290-190310290 on Build GRCh38
rs373750572 Chr.3: 190310361-190310361 on Build GRCh38
rs571302105 Chr.3: 190310399-190310399 on Build GRCh38
rs763674205 Chr.3: 190310178-190310178 on Build GRCh38
rs199727929 Chr.3: 190310153-190310153 on Build GRCh38
rs372251880 Chr.3: 190310130-190310130 on Build GRCh38
rs747336173 Chr.3: 190310028-190310028 on Build GRCh38
rs150983709 Chr.3: 190310209-190310209 on Build GRCh38
rs3774028 Chr.3: 190310483-190310483 on Build GRCh38
rs185356359 Chr.3: 190310433-190310433 on Build GRCh38
rs917027615 Chr.3: 190310394-190310394 on Build GRCh38
rs763472206 Chr.3: 190310293-190310293 on Build GRCh38
rs181213282 Chr.3: 190310191-190310191 on Build GRCh38
rs139053137 Chr.3: 190310434-190310434 on Build GRCh38
rs1014735218 Chr.3: 190310277-190310277 on Build GRCh38
rs528905129 Chr.3: 190310495-190310495 on Build GRCh38
rs757249673 Chr.3: 190310184-190310184 on Build GRCh38
rs573951377 Chr.3: 190310310-190310310 on Build GRCh38
rs551409503 Chr.3: 190310388-190310388 on Build GRCh38
rs745633918 Chr.3: 190310206-190310206 on Build GRCh38
rs72466473 Chr.3: 190310274-190310274 on Build GRCh38
rs763525135 Chr.3: 190310136-190310136 on Build GRCh38
rs906328206 Chr.3: 190310217-190310217 on Build GRCh38
rs1004839456 Chr.3: 190310304-190310304 on Build GRCh38
rs947757093 Chr.3: 190310436-190310437 on Build GRCh38
rs762559668 Chr.3: 190310106-190310106 on Build GRCh38
rs1041857328 Chr.3: 190310150-190310150 on Build GRCh38
rs111918856 Chr.3: 190310288-190310288 on Build GRCh38
rs759891453 Chr.3: 190310283-190310283 on Build GRCh38
rs142778400 Chr.3: 190310163-190310163 on Build GRCh38
rs760398568 Chr.3: 190310292-190310292 on Build GRCh38

View Full Product Details