Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 190857591-190858090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMNC
Entrez Gene ID
647309
Gene Name
geminin coiled-coil domain containing
Gene Aliases
GEMC1
UniGene
Hs.518490
Gene Chromosome Location
Chr.3: 190843271-190864200 on Build GRCh38
Transcripts
NM_001146686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1042545 c.380C>T p.A127V Chr.3: 190857787-190857787 on Build GRCh38
3945277 c.384+9G>C p.? Chr.3: 190857774-190857774 on Build GRCh38
5369263 c.368T>A p.L123H Chr.3: 190857799-190857799 on Build GRCh38
6256644 c.381C>A p.A127A Chr.3: 190857786-190857786 on Build GRCh38
3781787 c.268-10C>T p.? Chr.3: 190857909-190857909 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184563768 Chr.3: 190857825-190857825 on Build GRCh38
rs1025167042 Chr.3: 190857933-190857933 on Build GRCh38
rs772589916 Chr.3: 190857872-190857872 on Build GRCh38
rs750478690 Chr.3: 190857688-190857688 on Build GRCh38
rs752546753 Chr.3: 190857673-190857673 on Build GRCh38
rs998818351 Chr.3: 190858031-190858031 on Build GRCh38
rs918234794 Chr.3: 190857718-190857718 on Build GRCh38
rs760403119 Chr.3: 190857876-190857876 on Build GRCh38
rs986029836 Chr.3: 190857620-190857620 on Build GRCh38
rs939245826 Chr.3: 190857842-190857842 on Build GRCh38
rs773211032 Chr.3: 190857827-190857827 on Build GRCh38
rs193103620 Chr.3: 190857643-190857643 on Build GRCh38
rs1040511332 Chr.3: 190857723-190857723 on Build GRCh38
rs550614499 Chr.3: 190857622-190857623 on Build GRCh38
rs765084522 Chr.3: 190857640-190857640 on Build GRCh38
rs564469369 Chr.3: 190857849-190857849 on Build GRCh38
rs1007005980 Chr.3: 190857909-190857909 on Build GRCh38
rs762998597 Chr.3: 190857683-190857683 on Build GRCh38
rs373957419 Chr.3: 190857848-190857848 on Build GRCh38
rs878985668 Chr.3: 190857659-190857659 on Build GRCh38
rs983782552 Chr.3: 190858063-190858063 on Build GRCh38
rs943344412 Chr.3: 190857716-190857716 on Build GRCh38
rs763962502 Chr.3: 190857685-190857685 on Build GRCh38
rs972863821 Chr.3: 190858054-190858054 on Build GRCh38
rs753338032 Chr.3: 190857917-190857917 on Build GRCh38
rs146677614 Chr.3: 190858055-190858055 on Build GRCh38
rs73059981 Chr.3: 190857976-190857976 on Build GRCh38
rs754175319 Chr.3: 190857813-190857813 on Build GRCh38
rs1043079134 Chr.3: 190857883-190857883 on Build GRCh38
rs909633245 Chr.3: 190857790-190857790 on Build GRCh38
rs879115795 Chr.3: 190857717-190857717 on Build GRCh38
rs983836423 Chr.3: 190857766-190857766 on Build GRCh38
rs775355789 Chr.3: 190857903-190857903 on Build GRCh38
rs771421241 Chr.3: 190857850-190857850 on Build GRCh38
rs755660834 Chr.3: 190858018-190858018 on Build GRCh38
rs1018843086 Chr.3: 190857924-190857924 on Build GRCh38
rs201964563 Chr.3: 190857871-190857871 on Build GRCh38
rs760132964 Chr.3: 190857859-190857859 on Build GRCh38
rs778146127 Chr.3: 190857978-190857978 on Build GRCh38
rs562036553 Chr.3: 190857644-190857644 on Build GRCh38
rs753697898 Chr.3: 190857964-190857964 on Build GRCh38
rs75848819 Chr.3: 190857801-190857801 on Build GRCh38
rs756270821 Chr.3: 190857749-190857749 on Build GRCh38
rs201915885 Chr.3: 190858059-190858059 on Build GRCh38
rs532472313 Chr.3: 190857951-190857951 on Build GRCh38
rs370294362 Chr.3: 190857863-190857863 on Build GRCh38
rs73888733 Chr.3: 190858066-190858066 on Build GRCh38
rs569642057 Chr.3: 190857889-190857889 on Build GRCh38
rs774752647 Chr.3: 190857852-190857852 on Build GRCh38
rs901909735 Chr.3: 190857730-190857730 on Build GRCh38

View Full Product Details