Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 193410854-193411383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A4
Entrez Gene ID
84239
Gene Name
ATPase 13A4
Gene Aliases
-
UniGene
Hs.674423
Gene Chromosome Location
Chr.3: 193398967-193581725 on Build GRCh38
Transcripts
NM_032279.3 XM_017007318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4931310 c.3272T>C p.I1091T Chr.3: 193411007-193411007 on Build GRCh38
479859 c.3290G>A p.R1097H Chr.3: 193410989-193410989 on Build GRCh38
5016047 c.3234_3235insT p.Q1079fs*12 Chr.3: 193411044-193411045 on Build GRCh38
6207844 c.3295G>A p.D1099N Chr.3: 193410984-193410984 on Build GRCh38
7333471 c.3255C>T p.F1085F Chr.3: 193411024-193411024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008088887 Chr.3: 193411160-193411160 on Build GRCh38
rs1003253395 Chr.3: 193411020-193411020 on Build GRCh38
rs546555437 Chr.3: 193410902-193410902 on Build GRCh38
rs200545168 Chr.3: 193411099-193411099 on Build GRCh38
rs1051682799 Chr.3: 193411139-193411139 on Build GRCh38
rs1004132303 Chr.3: 193411317-193411317 on Build GRCh38
rs557454927 Chr.3: 193411341-193411341 on Build GRCh38
rs959040143 Chr.3: 193411056-193411056 on Build GRCh38
rs1037943385 Chr.3: 193411110-193411110 on Build GRCh38
rs980959218 Chr.3: 193411342-193411342 on Build GRCh38
rs923809840 Chr.3: 193410900-193410900 on Build GRCh38
rs368313622 Chr.3: 193411323-193411323 on Build GRCh38
rs142523415 Chr.3: 193411057-193411057 on Build GRCh38
rs762820976 Chr.3: 193411354-193411354 on Build GRCh38
rs753749300 Chr.3: 193410986-193410986 on Build GRCh38
rs772589811 Chr.3: 193411093-193411093 on Build GRCh38
rs771160175 Chr.3: 193411100-193411100 on Build GRCh38
rs777732175 Chr.3: 193411048-193411048 on Build GRCh38
rs771892085 Chr.3: 193410941-193410941 on Build GRCh38
rs867160724 Chr.3: 193411014-193411014 on Build GRCh38
rs571132381 Chr.3: 193411283-193411283 on Build GRCh38
rs763054132 Chr.3: 193410995-193410996 on Build GRCh38
rs763870482 Chr.3: 193411358-193411358 on Build GRCh38
rs796787099 Chr.3: 193411171-193411172 on Build GRCh38
rs550961390 Chr.3: 193411136-193411136 on Build GRCh38
rs763699813 Chr.3: 193410964-193410964 on Build GRCh38
rs139421235 Chr.3: 193410891-193410891 on Build GRCh38
rs970413291 Chr.3: 193411318-193411318 on Build GRCh38
rs113007405 Chr.3: 193411159-193411159 on Build GRCh38
rs890456781 Chr.3: 193411143-193411143 on Build GRCh38
rs755166699 Chr.3: 193410994-193410994 on Build GRCh38
rs771412732 Chr.3: 193411075-193411075 on Build GRCh38
rs1009239567 Chr.3: 193411276-193411276 on Build GRCh38
rs372740653 Chr.3: 193410929-193410929 on Build GRCh38
rs959365258 Chr.3: 193411329-193411329 on Build GRCh38
rs779306515 Chr.3: 193411086-193411086 on Build GRCh38
rs115378623 Chr.3: 193411137-193411137 on Build GRCh38
rs888479394 Chr.3: 193411279-193411279 on Build GRCh38
rs566394741 Chr.3: 193410927-193410927 on Build GRCh38
rs1024686788 Chr.3: 193411308-193411308 on Build GRCh38
rs1005723287 Chr.3: 193411281-193411281 on Build GRCh38
rs756123095 Chr.3: 193411035-193411035 on Build GRCh38
rs917679967 Chr.3: 193410995-193410995 on Build GRCh38
rs888687928 Chr.3: 193410975-193410975 on Build GRCh38
rs746287047 Chr.3: 193411091-193411091 on Build GRCh38
rs932783445 Chr.3: 193411244-193411244 on Build GRCh38
rs537661764 Chr.3: 193411248-193411248 on Build GRCh38
rs899687055 Chr.3: 193411219-193411219 on Build GRCh38
rs760415319 Chr.3: 193411103-193411103 on Build GRCh38
rs772995293 Chr.3: 193410960-193410960 on Build GRCh38
rs997273864 Chr.3: 193410954-193410954 on Build GRCh38
rs372120256 Chr.3: 193411021-193411021 on Build GRCh38
rs1039102671 Chr.3: 193411175-193411175 on Build GRCh38
rs1012038508 Chr.3: 193411344-193411344 on Build GRCh38
rs760215403 Chr.3: 193410956-193410956 on Build GRCh38
rs765075604 Chr.3: 193410990-193410990 on Build GRCh38
rs759222079 Chr.3: 193410933-193410933 on Build GRCh38
rs1037246945 Chr.3: 193411288-193411288 on Build GRCh38
rs151089128 Chr.3: 193410997-193410997 on Build GRCh38
rs765169494 Chr.3: 193410948-193410948 on Build GRCh38
rs61401139 Chr.3: 193411111-193411111 on Build GRCh38
rs1042226689 Chr.3: 193410894-193410894 on Build GRCh38
rs761713521 Chr.3: 193410989-193410989 on Build GRCh38
rs923736408 Chr.3: 193411162-193411162 on Build GRCh38
rs926818774 Chr.3: 193411038-193411038 on Build GRCh38
rs775119090 Chr.3: 193410951-193410951 on Build GRCh38
rs922694052 Chr.3: 193411205-193411205 on Build GRCh38
rs1032274564 Chr.3: 193411259-193411259 on Build GRCh38
rs752421953 Chr.3: 193411273-193411273 on Build GRCh38
rs944526934 Chr.3: 193411173-193411173 on Build GRCh38
rs978036976 Chr.3: 193411157-193411157 on Build GRCh38
rs113214329 Chr.3: 193410885-193410885 on Build GRCh38
rs901135955 Chr.3: 193410947-193410947 on Build GRCh38
rs765564010 Chr.3: 193411190-193411190 on Build GRCh38

View Full Product Details