Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 194403710-194404226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A3
Entrez Gene ID
79572
Gene Name
ATPase 13A3
Gene Aliases
AFURS1
UniGene
Hs.529609
Gene Chromosome Location
Chr.3: 194402674-194487006 on Build GRCh38
Transcripts
NM_024524.3 XM_005269357.3 XM_011513120.2 XM_011513121.1 XM_011513122.1 XM_011513123.2

SNP Detail

SNP ID Chromosome Location
rs552469274 Chr.3: 194403981-194403981 on Build GRCh38
rs1035929756 Chr.3: 194404158-194404158 on Build GRCh38
rs896150381 Chr.3: 194403918-194403918 on Build GRCh38
rs1023517389 Chr.3: 194403926-194403926 on Build GRCh38
rs546823356 Chr.3: 194403756-194403756 on Build GRCh38
rs960992608 Chr.3: 194403738-194403738 on Build GRCh38
rs979225444 Chr.3: 194403925-194403925 on Build GRCh38
rs529912773 Chr.3: 194403741-194403741 on Build GRCh38
rs947512143 Chr.3: 194403916-194403916 on Build GRCh38
rs570737469 Chr.3: 194403985-194403985 on Build GRCh38
rs972791225 Chr.3: 194404043-194404043 on Build GRCh38
rs201096434 Chr.3: 194404086-194404086 on Build GRCh38
rs955370321 Chr.3: 194404087-194404087 on Build GRCh38
rs1005311420 Chr.3: 194404061-194404061 on Build GRCh38
rs377047250 Chr.3: 194403742-194403742 on Build GRCh38
rs200011258 Chr.3: 194404062-194404062 on Build GRCh38
rs14414 Chr.3: 194403895-194403895 on Build GRCh38
rs758719986 Chr.3: 194404071-194404071 on Build GRCh38
rs190436920 Chr.3: 194404088-194404088 on Build GRCh38
rs60210039 Chr.3: 194403927-194403928 on Build GRCh38
rs879687908 Chr.3: 194404055-194404055 on Build GRCh38
rs746580802 Chr.3: 194404144-194404144 on Build GRCh38
rs972289788 Chr.3: 194403775-194403775 on Build GRCh38
rs185581435 Chr.3: 194403955-194403955 on Build GRCh38
rs747642175 Chr.3: 194404185-194404185 on Build GRCh38
rs9878016 Chr.3: 194403760-194403760 on Build GRCh38
rs780629086 Chr.3: 194404176-194404176 on Build GRCh38
rs933101684 Chr.3: 194403917-194403917 on Build GRCh38
rs935054044 Chr.3: 194403889-194403889 on Build GRCh38
rs527557368 Chr.3: 194403965-194403965 on Build GRCh38
rs916003416 Chr.3: 194403909-194403910 on Build GRCh38
rs753947007 Chr.3: 194403998-194403998 on Build GRCh38
rs768354583 Chr.3: 194404151-194404151 on Build GRCh38
rs767285736 Chr.3: 194403918-194403919 on Build GRCh38
rs761147700 Chr.3: 194403975-194403975 on Build GRCh38
rs987222016 Chr.3: 194404096-194404096 on Build GRCh38
rs779776111 Chr.3: 194404116-194404116 on Build GRCh38
rs911452123 Chr.3: 194404119-194404119 on Build GRCh38
rs758861949 Chr.3: 194403928-194403928 on Build GRCh38
rs1049601965 Chr.3: 194404041-194404041 on Build GRCh38
rs1006118986 Chr.3: 194403959-194403959 on Build GRCh38
rs374156178 Chr.3: 194403928-194403928 on Build GRCh38
rs757818952 Chr.3: 194404032-194404032 on Build GRCh38
rs932498085 Chr.3: 194404001-194404001 on Build GRCh38
rs1041018798 Chr.3: 194403942-194403942 on Build GRCh38
rs538274525 Chr.3: 194403994-194403994 on Build GRCh38
rs961887164 Chr.3: 194403966-194403966 on Build GRCh38
rs904043484 Chr.3: 194403780-194403780 on Build GRCh38
rs942930345 Chr.3: 194404129-194404129 on Build GRCh38
rs3836225 Chr.3: 194403918-194403918 on Build GRCh38
rs779286915 Chr.3: 194404059-194404059 on Build GRCh38
rs796767353 Chr.3: 194403917-194403918 on Build GRCh38
rs1013671385 Chr.3: 194403920-194403920 on Build GRCh38
rs769429344 Chr.3: 194403979-194403979 on Build GRCh38
rs556231966 Chr.3: 194404057-194404057 on Build GRCh38
rs960230001 Chr.3: 194403842-194403842 on Build GRCh38
rs925435493 Chr.3: 194404197-194404197 on Build GRCh38
rs397876388 Chr.3: 194403927-194403927 on Build GRCh38

View Full Product Details