Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 5729229-5729719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVC
Entrez Gene ID
2121
Gene Name
EvC ciliary complex subunit 1
Gene Aliases
DWF-1, EVC1, EVCL
UniGene
Hs.646899
Gene Chromosome Location
Chr.4: 5711197-5829043 on Build GRCh38
Transcripts
NM_001306090.1 NM_001306092.1 NM_153717.2 XM_006713865.3 XM_006713866.3 XM_011513419.2 XM_017007883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5457129 c.310C>T p.P104S Chr.4: 5729316-5729316 on Build GRCh38
3781173 c.311C>T p.P104L Chr.4: 5729317-5729317 on Build GRCh38
6469626 c.378G>A p.K126K Chr.4: 5729384-5729384 on Build GRCh38
4125291 c.346A>G p.K116E Chr.4: 5729352-5729352 on Build GRCh38
6535276 c.333A>G p.T111T Chr.4: 5729339-5729339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764925850 Chr.4: 5729300-5729300 on Build GRCh38
rs557081941 Chr.4: 5729687-5729687 on Build GRCh38
rs779379855 Chr.4: 5729355-5729355 on Build GRCh38
rs955125916 Chr.4: 5729604-5729604 on Build GRCh38
rs866940441 Chr.4: 5729316-5729316 on Build GRCh38
rs192800323 Chr.4: 5729598-5729598 on Build GRCh38
rs780155770 Chr.4: 5729270-5729270 on Build GRCh38
rs886512159 Chr.4: 5729568-5729568 on Build GRCh38
rs750365856 Chr.4: 5729421-5729421 on Build GRCh38
rs913433865 Chr.4: 5729496-5729496 on Build GRCh38
rs1001609281 Chr.4: 5729309-5729309 on Build GRCh38
rs775193215 Chr.4: 5729308-5729308 on Build GRCh38
rs542798399 Chr.4: 5729460-5729460 on Build GRCh38
rs1017287863 Chr.4: 5729573-5729573 on Build GRCh38
rs551724650 Chr.4: 5729542-5729542 on Build GRCh38
rs769018634 Chr.4: 5729457-5729457 on Build GRCh38
rs889977681 Chr.4: 5729538-5729538 on Build GRCh38
rs368345423 Chr.4: 5729284-5729284 on Build GRCh38
rs199748764 Chr.4: 5729277-5729277 on Build GRCh38
rs779287656 Chr.4: 5729434-5729434 on Build GRCh38
rs1055349346 Chr.4: 5729465-5729465 on Build GRCh38
rs763380800 Chr.4: 5729315-5729315 on Build GRCh38
rs149350067 Chr.4: 5729622-5729622 on Build GRCh38
rs541331576 Chr.4: 5729282-5729282 on Build GRCh38
rs763768630 Chr.4: 5729411-5729411 on Build GRCh38
rs981843041 Chr.4: 5729628-5729629 on Build GRCh38
rs761529346 Chr.4: 5729419-5729419 on Build GRCh38
rs895870813 Chr.4: 5729649-5729649 on Build GRCh38
rs370301492 Chr.4: 5729514-5729514 on Build GRCh38
rs745382419 Chr.4: 5729391-5729391 on Build GRCh38
rs775105402 Chr.4: 5729407-5729407 on Build GRCh38
rs373751797 Chr.4: 5729519-5729519 on Build GRCh38
rs755648574 Chr.4: 5729607-5729607 on Build GRCh38
rs774611632 Chr.4: 5729688-5729688 on Build GRCh38
rs1001566646 Chr.4: 5729528-5729528 on Build GRCh38
rs913390171 Chr.4: 5729608-5729608 on Build GRCh38
rs900974818 Chr.4: 5729581-5729581 on Build GRCh38
rs1048461292 Chr.4: 5729673-5729673 on Build GRCh38
rs1034459378 Chr.4: 5729325-5729325 on Build GRCh38
rs373552911 Chr.4: 5729432-5729432 on Build GRCh38
rs200668643 Chr.4: 5729323-5729323 on Build GRCh38
rs937302861 Chr.4: 5729641-5729641 on Build GRCh38
rs772569904 Chr.4: 5729373-5729373 on Build GRCh38
rs1055861659 Chr.4: 5729544-5729544 on Build GRCh38
rs975113723 Chr.4: 5729454-5729454 on Build GRCh38
rs957971912 Chr.4: 5729600-5729600 on Build GRCh38
rs769229885 Chr.4: 5729405-5729405 on Build GRCh38
rs559598572 Chr.4: 5729508-5729508 on Build GRCh38
rs113870218 Chr.4: 5729431-5729431 on Build GRCh38
rs899450901 Chr.4: 5729513-5729513 on Build GRCh38
rs1055808580 Chr.4: 5729642-5729642 on Build GRCh38
rs920830845 Chr.4: 5729669-5729669 on Build GRCh38
rs951334006 Chr.4: 5729381-5729381 on Build GRCh38
rs571424276 Chr.4: 5729582-5729582 on Build GRCh38
rs113320243 Chr.4: 5729501-5729501 on Build GRCh38
rs926213739 Chr.4: 5729630-5729630 on Build GRCh38
rs753090751 Chr.4: 5729436-5729436 on Build GRCh38
rs1047629571 Chr.4: 5729666-5729666 on Build GRCh38
rs150366183 Chr.4: 5729420-5729420 on Build GRCh38
rs946326522 Chr.4: 5729500-5729500 on Build GRCh38
rs1014058008 Chr.4: 5729559-5729559 on Build GRCh38
rs530512638 Chr.4: 5729570-5729570 on Build GRCh38
rs200666993 Chr.4: 5729409-5729409 on Build GRCh38
rs1014341013 Chr.4: 5729659-5729659 on Build GRCh38
rs760784095 Chr.4: 5729425-5729425 on Build GRCh38
rs1051899695 Chr.4: 5729493-5729493 on Build GRCh38
rs1025485105 Chr.4: 5729565-5729565 on Build GRCh38
rs772483071 Chr.4: 5729286-5729286 on Build GRCh38
rs774065578 Chr.4: 5729412-5729412 on Build GRCh38
rs116362495 Chr.4: 5729672-5729672 on Build GRCh38
rs188229024 Chr.4: 5729273-5729273 on Build GRCh38
rs138747947 Chr.4: 5729572-5729572 on Build GRCh38
rs144748530 Chr.4: 5729653-5729653 on Build GRCh38
rs767361058 Chr.4: 5729336-5729336 on Build GRCh38
rs201640469 Chr.4: 5729317-5729317 on Build GRCh38
rs751381591 Chr.4: 5729320-5729320 on Build GRCh38
rs895924307 Chr.4: 5729550-5729550 on Build GRCh38
rs904517610 Chr.4: 5729523-5729523 on Build GRCh38
rs755273705 Chr.4: 5729349-5729349 on Build GRCh38
rs756208272 Chr.4: 5729346-5729346 on Build GRCh38
rs567280707 Chr.4: 5729340-5729340 on Build GRCh38
rs763930196 Chr.4: 5729318-5729318 on Build GRCh38
rs773647675 Chr.4: 5729389-5729389 on Build GRCh38
rs766517088 Chr.4: 5729430-5729430 on Build GRCh38
rs187379961 Chr.4: 5729629-5729629 on Build GRCh38
rs964089562 Chr.4: 5729444-5729444 on Build GRCh38
rs112592757 Chr.4: 5729400-5729400 on Build GRCh38
rs578211088 Chr.4: 5729663-5729663 on Build GRCh38
rs182685672 Chr.4: 5729617-5729617 on Build GRCh38
rs758978764 Chr.4: 5729299-5729299 on Build GRCh38
rs757180182 Chr.4: 5729327-5729327 on Build GRCh38
rs565063766 Chr.4: 5729546-5729546 on Build GRCh38
rs528360373 Chr.4: 5729509-5729509 on Build GRCh38
rs922332110 Chr.4: 5729599-5729599 on Build GRCh38
rs750461424 Chr.4: 5729345-5729345 on Build GRCh38
rs754656867 Chr.4: 5729461-5729461 on Build GRCh38
rs16837598 Chr.4: 5729347-5729347 on Build GRCh38
rs748523193 Chr.4: 5729369-5729369 on Build GRCh38

View Full Product Details