Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 5745063-5745611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVC
Entrez Gene ID
2121
Gene Name
EvC ciliary complex subunit 1
Gene Aliases
DWF-1, EVC1, EVCL
UniGene
Hs.646899
Gene Chromosome Location
Chr.4: 5711197-5829043 on Build GRCh38
Transcripts
NM_001306090.1 NM_001306092.1 NM_153717.2 XM_006713865.3 XM_006713866.3 XM_011513419.2 XM_017007883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5959588 c.922G>C p.E308Q Chr.4: 5745324-5745324 on Build GRCh38
4805791 c.939+1G>T p.? Chr.4: 5745342-5745342 on Build GRCh38
280882 c.900A>C p.E300D Chr.4: 5745302-5745302 on Build GRCh38
5918391 c.808G>A p.E270K Chr.4: 5745210-5745210 on Build GRCh38
4779590 c.845A>G p.N282S Chr.4: 5745247-5745247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368231289 Chr.4: 5745308-5745308 on Build GRCh38
rs121908426 Chr.4: 5745321-5745321 on Build GRCh38
rs762732311 Chr.4: 5745401-5745401 on Build GRCh38
rs753109171 Chr.4: 5745172-5745172 on Build GRCh38
rs753313712 Chr.4: 5745390-5745391 on Build GRCh38
rs780133664 Chr.4: 5745540-5745540 on Build GRCh38
rs771897142 Chr.4: 5745213-5745213 on Build GRCh38
rs2286343 Chr.4: 5745345-5745345 on Build GRCh38
rs752988350 Chr.4: 5745260-5745260 on Build GRCh38
rs796069712 Chr.4: 5745189-5745192 on Build GRCh38
rs376749826 Chr.4: 5745392-5745392 on Build GRCh38
rs796629702 Chr.4: 5745381-5745381 on Build GRCh38
rs374155542 Chr.4: 5745182-5745182 on Build GRCh38
rs1044754513 Chr.4: 5745152-5745152 on Build GRCh38
rs939844212 Chr.4: 5745105-5745105 on Build GRCh38
rs1009790386 Chr.4: 5745441-5745442 on Build GRCh38
rs918964471 Chr.4: 5745584-5745584 on Build GRCh38
rs773954366 Chr.4: 5745198-5745198 on Build GRCh38
rs4689313 Chr.4: 5745189-5745189 on Build GRCh38
rs554199549 Chr.4: 5745188-5745189 on Build GRCh38
rs956149770 Chr.4: 5745431-5745431 on Build GRCh38
rs200363737 Chr.4: 5745385-5745385 on Build GRCh38
rs753304898 Chr.4: 5745191-5745195 on Build GRCh38
rs575589201 Chr.4: 5745236-5745236 on Build GRCh38
rs796964056 Chr.4: 5745090-5745090 on Build GRCh38
rs144142557 Chr.4: 5745391-5745391 on Build GRCh38
rs756222363 Chr.4: 5745535-5745535 on Build GRCh38
rs745906646 Chr.4: 5745376-5745376 on Build GRCh38
rs756133108 Chr.4: 5745392-5745393 on Build GRCh38
rs59741480 Chr.4: 5745541-5745541 on Build GRCh38
rs963076215 Chr.4: 5745566-5745566 on Build GRCh38
rs145300726 Chr.4: 5745330-5745330 on Build GRCh38
rs199743160 Chr.4: 5745386-5745386 on Build GRCh38
rs971036648 Chr.4: 5745456-5745456 on Build GRCh38
rs772037385 Chr.4: 5745315-5745315 on Build GRCh38
rs772113119 Chr.4: 5745187-5745192 on Build GRCh38
rs748894265 Chr.4: 5745190-5745190 on Build GRCh38
rs386671011 Chr.4: 5745189-5745192 on Build GRCh38
rs980704516 Chr.4: 5745457-5745457 on Build GRCh38
rs754158651 Chr.4: 5745159-5745159 on Build GRCh38
rs527255616 Chr.4: 5745274-5745275 on Build GRCh38
rs201282320 Chr.4: 5745350-5745350 on Build GRCh38
rs1032117694 Chr.4: 5745325-5745325 on Build GRCh38
rs779502889 Chr.4: 5745282-5745282 on Build GRCh38
rs372546602 Chr.4: 5745191-5745192 on Build GRCh38
rs150814290 Chr.4: 5745203-5745203 on Build GRCh38
rs886038741 Chr.4: 5745199-5745201 on Build GRCh38
rs759718572 Chr.4: 5745335-5745335 on Build GRCh38
rs777643436 Chr.4: 5745367-5745367 on Build GRCh38
rs28609792 Chr.4: 5745455-5745455 on Build GRCh38
rs770139825 Chr.4: 5745357-5745357 on Build GRCh38
rs115275195 Chr.4: 5745336-5745336 on Build GRCh38
rs369153323 Chr.4: 5745189-5745189 on Build GRCh38
rs778311533 Chr.4: 5745180-5745180 on Build GRCh38
rs758866277 Chr.4: 5745269-5745269 on Build GRCh38
rs370315662 Chr.4: 5745187-5745189 on Build GRCh38
rs764394644 Chr.4: 5745341-5745341 on Build GRCh38
rs375562419 Chr.4: 5745164-5745164 on Build GRCh38
rs902304992 Chr.4: 5745278-5745278 on Build GRCh38
rs55955937 Chr.4: 5745352-5745352 on Build GRCh38
rs553620673 Chr.4: 5745153-5745153 on Build GRCh38
rs919854123 Chr.4: 5745155-5745155 on Build GRCh38
rs764555992 Chr.4: 5745271-5745271 on Build GRCh38
rs533849635 Chr.4: 5745400-5745400 on Build GRCh38
rs754532508 Chr.4: 5745286-5745286 on Build GRCh38
rs755381180 Chr.4: 5745303-5745305 on Build GRCh38
rs999259787 Chr.4: 5745426-5745426 on Build GRCh38
rs758772538 Chr.4: 5745173-5745173 on Build GRCh38
rs199729910 Chr.4: 5745369-5745369 on Build GRCh38
rs757788315 Chr.4: 5745274-5745274 on Build GRCh38
rs16837617 Chr.4: 5745425-5745425 on Build GRCh38
rs13145958 Chr.4: 5745404-5745404 on Build GRCh38
rs1001180 Chr.4: 5745565-5745565 on Build GRCh38
rs574100025 Chr.4: 5745568-5745568 on Build GRCh38
rs771069449 Chr.4: 5745182-5745183 on Build GRCh38
rs1053263004 Chr.4: 5745441-5745441 on Build GRCh38
rs372548610 Chr.4: 5745322-5745322 on Build GRCh38
rs755662595 Chr.4: 5745186-5745186 on Build GRCh38
rs981543181 Chr.4: 5745087-5745087 on Build GRCh38
rs375414448 Chr.4: 5745284-5745284 on Build GRCh38
rs575855063 Chr.4: 5745536-5745536 on Build GRCh38
rs998975965 Chr.4: 5745290-5745290 on Build GRCh38
rs370643731 Chr.4: 5745252-5745252 on Build GRCh38
rs915103359 Chr.4: 5745204-5745204 on Build GRCh38
rs373136313 Chr.4: 5745127-5745127 on Build GRCh38
rs559155235 Chr.4: 5745524-5745524 on Build GRCh38
rs565481068 Chr.4: 5745226-5745226 on Build GRCh38
rs749581634 Chr.4: 5745166-5745169 on Build GRCh38
rs2286342 Chr.4: 5745351-5745351 on Build GRCh38
rs139263450 Chr.4: 5745259-5745259 on Build GRCh38
rs890722382 Chr.4: 5745279-5745279 on Build GRCh38
rs747991962 Chr.4: 5745205-5745205 on Build GRCh38
rs776792609 Chr.4: 5745332-5745332 on Build GRCh38
rs769968051 Chr.4: 5745377-5745377 on Build GRCh38
rs746814100 Chr.4: 5745368-5745368 on Build GRCh38
rs768028302 Chr.4: 5745348-5745348 on Build GRCh38
rs536802925 Chr.4: 5745123-5745123 on Build GRCh38
rs3214258 Chr.4: 5745380-5745380 on Build GRCh38
rs367702379 Chr.4: 5745249-5745249 on Build GRCh38
rs752110410 Chr.4: 5745273-5745273 on Build GRCh38
rs751870400 Chr.4: 5745295-5745301 on Build GRCh38
rs149529742 Chr.4: 5745301-5745301 on Build GRCh38
rs761463883 Chr.4: 5745237-5745239 on Build GRCh38
rs781255824 Chr.4: 5745375-5745375 on Build GRCh38
rs760812070 Chr.4: 5745246-5745246 on Build GRCh38
rs759012138 Chr.4: 5745454-5745454 on Build GRCh38
rs1049373754 Chr.4: 5745168-5745168 on Build GRCh38
rs1037859021 Chr.4: 5745223-5745223 on Build GRCh38
rs61514191 Chr.4: 5745192-5745192 on Build GRCh38
rs573504737 Chr.4: 5745160-5745160 on Build GRCh38

View Full Product Details