Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116397810-116398307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A1
Entrez Gene ID
476
Gene Name
ATPase Na+/K+ transporting subunit alpha 1
Gene Aliases
-
UniGene
Hs.371889
Gene Chromosome Location
Chr.1: 116373173-116404774 on Build GRCh38
Transcripts
NM_000701.7 NM_001160233.1 NM_001160234.1 XM_017001360.1 XM_017001361.1 NR_027646.1

Gene Symbol
ATP1A1-AS1
Entrez Gene ID
84852
Gene Name
ATP1A1 antisense RNA 1
Gene Aliases
ATP1A1OS, C1orf203
UniGene
Hs.664752
Gene Chromosome Location
Chr.1: 116392865-116418622 on Build GRCh38
Transcripts
NM_000701.7 NM_001160233.1 NM_001160234.1 XM_017001360.1 XM_017001361.1 NR_027646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6607362 c.2026G>A p.E676K Chr.1: 116397940-116397940 on Build GRCh38
4475909 c.2024C>T p.S675F Chr.1: 116397938-116397938 on Build GRCh38
4664267 c.2000G>A p.G667D Chr.1: 116397914-116397914 on Build GRCh38
219185 c.2089C>T p.Q697* Chr.1: 116398003-116398003 on Build GRCh38
159268 c.1989C>T p.C663C Chr.1: 116397903-116397903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770790655 Chr.1: 116397874-116397875 on Build GRCh38
rs755662541 Chr.1: 116398267-116398267 on Build GRCh38
rs764612155 Chr.1: 116398054-116398054 on Build GRCh38
rs779993188 Chr.1: 116397936-116397936 on Build GRCh38
rs546359303 Chr.1: 116398241-116398241 on Build GRCh38
rs768375815 Chr.1: 116397940-116397940 on Build GRCh38
rs563446405 Chr.1: 116397860-116397860 on Build GRCh38
rs754577439 Chr.1: 116397860-116397861 on Build GRCh38
rs758792227 Chr.1: 116397883-116397883 on Build GRCh38
rs376365122 Chr.1: 116398059-116398059 on Build GRCh38
rs981441627 Chr.1: 116398249-116398249 on Build GRCh38
rs1002688907 Chr.1: 116398091-116398091 on Build GRCh38
rs752121779 Chr.1: 116397859-116397860 on Build GRCh38
rs77990511 Chr.1: 116397872-116397874 on Build GRCh38
rs750495844 Chr.1: 116398073-116398073 on Build GRCh38
rs1027840023 Chr.1: 116398199-116398199 on Build GRCh38
rs947047439 Chr.1: 116397867-116397867 on Build GRCh38
rs200287013 Chr.1: 116397875-116397875 on Build GRCh38
rs140713179 Chr.1: 116398268-116398268 on Build GRCh38
rs779862831 Chr.1: 116398089-116398089 on Build GRCh38
rs200766300 Chr.1: 116398057-116398057 on Build GRCh38
rs369360789 Chr.1: 116397875-116397875 on Build GRCh38
rs756054875 Chr.1: 116397921-116397921 on Build GRCh38
rs757594670 Chr.1: 116398063-116398063 on Build GRCh38
rs745853855 Chr.1: 116397840-116397840 on Build GRCh38
rs972177024 Chr.1: 116398192-116398192 on Build GRCh38
rs560327937 Chr.1: 116397839-116397839 on Build GRCh38
rs766110446 Chr.1: 116398068-116398068 on Build GRCh38
rs745743585 Chr.1: 116397877-116397881 on Build GRCh38
rs775995585 Chr.1: 116398048-116398048 on Build GRCh38
rs1008557955 Chr.1: 116398136-116398136 on Build GRCh38
rs768320809 Chr.1: 116397853-116397853 on Build GRCh38
rs140135222 Chr.1: 116397913-116397913 on Build GRCh38
rs200285355 Chr.1: 116397904-116397904 on Build GRCh38
rs751169483 Chr.1: 116397855-116397856 on Build GRCh38
rs760179843 Chr.1: 116398032-116398032 on Build GRCh38
rs550805372 Chr.1: 116398125-116398125 on Build GRCh38
rs924663683 Chr.1: 116398272-116398272 on Build GRCh38
rs751776150 Chr.1: 116397885-116397885 on Build GRCh38
rs375599371 Chr.1: 116398075-116398075 on Build GRCh38
rs377724625 Chr.1: 116397912-116397912 on Build GRCh38
rs151323380 Chr.1: 116397939-116397939 on Build GRCh38
rs147280556 Chr.1: 116398235-116398235 on Build GRCh38
rs753022756 Chr.1: 116397881-116397881 on Build GRCh38
rs572513531 Chr.1: 116398115-116398115 on Build GRCh38
rs774116749 Chr.1: 116397954-116397954 on Build GRCh38
rs765691491 Chr.1: 116398045-116398045 on Build GRCh38
rs767071147 Chr.1: 116397857-116397857 on Build GRCh38
rs181070201 Chr.1: 116398018-116398018 on Build GRCh38
rs754375453 Chr.1: 116398187-116398187 on Build GRCh38
rs755705051 Chr.1: 116397860-116397862 on Build GRCh38
rs1018609791 Chr.1: 116398161-116398161 on Build GRCh38
rs1006654437 Chr.1: 116398051-116398051 on Build GRCh38
rs569086956 Chr.1: 116398232-116398232 on Build GRCh38
rs373912666 Chr.1: 116397855-116397855 on Build GRCh38
rs149660732 Chr.1: 116398029-116398029 on Build GRCh38
rs143047843 Chr.1: 116398282-116398282 on Build GRCh38
rs371087257 Chr.1: 116398017-116398017 on Build GRCh38
rs747862363 Chr.1: 116397969-116397969 on Build GRCh38
rs765748374 Chr.1: 116397877-116397877 on Build GRCh38
rs754913955 Chr.1: 116398138-116398138 on Build GRCh38
rs764413614 Chr.1: 116397884-116397884 on Build GRCh38
rs775189876 Chr.1: 116397848-116397848 on Build GRCh38
rs902103517 Chr.1: 116398038-116398038 on Build GRCh38
rs1021962291 Chr.1: 116398144-116398144 on Build GRCh38
rs769770038 Chr.1: 116397846-116397846 on Build GRCh38
rs762785789 Chr.1: 116398050-116398050 on Build GRCh38
rs749088596 Chr.1: 116397852-116397852 on Build GRCh38
rs952335580 Chr.1: 116398204-116398204 on Build GRCh38
rs527773306 Chr.1: 116397874-116397874 on Build GRCh38
rs370072127 Chr.1: 116397854-116397854 on Build GRCh38
rs777385008 Chr.1: 116398273-116398273 on Build GRCh38

View Full Product Details