Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116399247-116399755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A1
Entrez Gene ID
476
Gene Name
ATPase Na+/K+ transporting subunit alpha 1
Gene Aliases
-
UniGene
Hs.371889
Gene Chromosome Location
Chr.1: 116373173-116404774 on Build GRCh38
Transcripts
NM_000701.7 NM_001160233.1 NM_001160234.1 XM_017001360.1 XM_017001361.1 NR_027645.1 NR_027646.1

Gene Symbol
ATP1A1-AS1
Entrez Gene ID
84852
Gene Name
ATP1A1 antisense RNA 1
Gene Aliases
ATP1A1OS, C1orf203
UniGene
Hs.664752
Gene Chromosome Location
Chr.1: 116392865-116418622 on Build GRCh38
Transcripts
NM_000701.7 NM_001160233.1 NM_001160234.1 XM_017001360.1 XM_017001361.1 NR_027645.1 NR_027646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2178198 c.2456C>A p.A819D Chr.1: 116399427-116399427 on Build GRCh38
7093857 c.2531T>G p.L844R Chr.1: 116399502-116399502 on Build GRCh38
4777885 c.2517_2518insC p.K840fs*8 Chr.1: 116399488-116399489 on Build GRCh38
3801399 c.2450T>C p.V817A Chr.1: 116399421-116399421 on Build GRCh38
5735596 c.2468C>T p.A823V Chr.1: 116399439-116399439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778357280 Chr.1: 116399521-116399521 on Build GRCh38
rs773162224 Chr.1: 116399591-116399591 on Build GRCh38
rs749391695 Chr.1: 116399578-116399578 on Build GRCh38
rs571336821 Chr.1: 116399677-116399677 on Build GRCh38
rs879054066 Chr.1: 116399418-116399418 on Build GRCh38
rs1019689855 Chr.1: 116399309-116399309 on Build GRCh38
rs752064129 Chr.1: 116399555-116399555 on Build GRCh38
rs12134762 Chr.1: 116399715-116399715 on Build GRCh38
rs374360169 Chr.1: 116399589-116399589 on Build GRCh38
rs766077481 Chr.1: 116399458-116399458 on Build GRCh38
rs879180654 Chr.1: 116399417-116399417 on Build GRCh38
rs878999201 Chr.1: 116399551-116399551 on Build GRCh38
rs145603535 Chr.1: 116399446-116399446 on Build GRCh38
rs748686358 Chr.1: 116399673-116399673 on Build GRCh38
rs762893078 Chr.1: 116399398-116399398 on Build GRCh38
rs886736060 Chr.1: 116399731-116399731 on Build GRCh38
rs563915526 Chr.1: 116399367-116399367 on Build GRCh38
rs954094705 Chr.1: 116399297-116399297 on Build GRCh38
rs1021906246 Chr.1: 116399651-116399651 on Build GRCh38
rs146093017 Chr.1: 116399506-116399506 on Build GRCh38
rs566961905 Chr.1: 116399581-116399581 on Build GRCh38
rs770027849 Chr.1: 116399571-116399571 on Build GRCh38
rs975888839 Chr.1: 116399316-116399316 on Build GRCh38
rs780098925 Chr.1: 116399572-116399572 on Build GRCh38
rs760132449 Chr.1: 116399371-116399371 on Build GRCh38
rs963515301 Chr.1: 116399517-116399517 on Build GRCh38
rs140757717 Chr.1: 116399396-116399396 on Build GRCh38
rs143621813 Chr.1: 116399689-116399689 on Build GRCh38
rs752682423 Chr.1: 116399366-116399366 on Build GRCh38
rs879032202 Chr.1: 116399545-116399545 on Build GRCh38
rs547257447 Chr.1: 116399374-116399374 on Build GRCh38
rs564674909 Chr.1: 116399614-116399614 on Build GRCh38
rs540824543 Chr.1: 116399672-116399672 on Build GRCh38
rs768565294 Chr.1: 116399580-116399580 on Build GRCh38
rs377754449 Chr.1: 116399556-116399556 on Build GRCh38
rs879223959 Chr.1: 116399549-116399549 on Build GRCh38
rs533104598 Chr.1: 116399476-116399476 on Build GRCh38
rs572774640 Chr.1: 116399702-116399702 on Build GRCh38
rs756876559 Chr.1: 116399451-116399451 on Build GRCh38
rs975857496 Chr.1: 116399713-116399713 on Build GRCh38
rs960318565 Chr.1: 116399603-116399603 on Build GRCh38
rs865875870 Chr.1: 116399486-116399486 on Build GRCh38
rs746210378 Chr.1: 116399568-116399568 on Build GRCh38
rs750241957 Chr.1: 116399435-116399435 on Build GRCh38
rs746153260 Chr.1: 116399372-116399372 on Build GRCh38
rs879132863 Chr.1: 116399414-116399414 on Build GRCh38
rs112014183 Chr.1: 116399654-116399654 on Build GRCh38
rs761686227 Chr.1: 116399409-116399409 on Build GRCh38
rs554348104 Chr.1: 116399701-116399701 on Build GRCh38
rs767480768 Chr.1: 116399413-116399413 on Build GRCh38
rs149820510 Chr.1: 116399512-116399512 on Build GRCh38
rs781575872 Chr.1: 116399559-116399559 on Build GRCh38
rs371191515 Chr.1: 116399582-116399582 on Build GRCh38
rs148907350 Chr.1: 116399644-116399644 on Build GRCh38
rs753467237 Chr.1: 116399450-116399450 on Build GRCh38
rs554987249 Chr.1: 116399717-116399717 on Build GRCh38
rs878953125 Chr.1: 116399547-116399547 on Build GRCh38
rs12724229 Chr.1: 116399492-116399492 on Build GRCh38
rs913190368 Chr.1: 116399612-116399612 on Build GRCh38
rs144606788 Chr.1: 116399402-116399402 on Build GRCh38
rs992835463 Chr.1: 116399607-116399607 on Build GRCh38
rs186049733 Chr.1: 116399310-116399310 on Build GRCh38
rs755431362 Chr.1: 116399665-116399665 on Build GRCh38
rs985933981 Chr.1: 116399306-116399306 on Build GRCh38
rs769974823 Chr.1: 116399375-116399375 on Build GRCh38
rs892024166 Chr.1: 116399641-116399641 on Build GRCh38
rs879006856 Chr.1: 116399415-116399415 on Build GRCh38
rs774093509 Chr.1: 116399407-116399407 on Build GRCh38

View Full Product Details