Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 117865331-117865875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDAP2
Entrez Gene ID
54834
Gene Name
ganglioside induced differentiation associated protein 2
Gene Aliases
MACROD3
UniGene
Hs.310809
Gene Chromosome Location
Chr.1: 117860227-117930042 on Build GRCh38
Transcripts
NM_017686.3 XM_005270959.2 XM_011541623.2 XM_017001531.1 XM_017001532.1

SNP Detail

SNP ID Chromosome Location
rs182491737 Chr.1: 117865702-117865702 on Build GRCh38
rs944973255 Chr.1: 117865497-117865497 on Build GRCh38
rs938090041 Chr.1: 117865659-117865659 on Build GRCh38
rs186867100 Chr.1: 117865400-117865400 on Build GRCh38
rs1055129457 Chr.1: 117865437-117865437 on Build GRCh38
rs200524703 Chr.1: 117865732-117865732 on Build GRCh38
rs535181671 Chr.1: 117865599-117865599 on Build GRCh38
rs907711479 Chr.1: 117865615-117865615 on Build GRCh38
rs893244111 Chr.1: 117865587-117865587 on Build GRCh38
rs565924898 Chr.1: 117865743-117865743 on Build GRCh38
rs1002647749 Chr.1: 117865433-117865433 on Build GRCh38
rs1000578563 Chr.1: 117865578-117865578 on Build GRCh38
rs552844422 Chr.1: 117865591-117865591 on Build GRCh38
rs539641688 Chr.1: 117865827-117865827 on Build GRCh38
rs190037886 Chr.1: 117865614-117865614 on Build GRCh38
rs941806184 Chr.1: 117865621-117865621 on Build GRCh38
rs764350941 Chr.1: 117865480-117865480 on Build GRCh38
rs574007462 Chr.1: 117865571-117865571 on Build GRCh38
rs920394225 Chr.1: 117865454-117865454 on Build GRCh38
rs79357772 Chr.1: 117865606-117865606 on Build GRCh38
rs966813226 Chr.1: 117865577-117865577 on Build GRCh38
rs990530755 Chr.1: 117865695-117865695 on Build GRCh38
rs576638417 Chr.1: 117865803-117865803 on Build GRCh38
rs547851192 Chr.1: 117865471-117865471 on Build GRCh38
rs1047910044 Chr.1: 117865495-117865495 on Build GRCh38
rs930795756 Chr.1: 117865470-117865470 on Build GRCh38
rs893179532 Chr.1: 117865846-117865846 on Build GRCh38
rs1032086508 Chr.1: 117865579-117865579 on Build GRCh38
rs775917905 Chr.1: 117865362-117865362 on Build GRCh38
rs1037530204 Chr.1: 117865650-117865650 on Build GRCh38
rs908058743 Chr.1: 117865742-117865742 on Build GRCh38
rs990975246 Chr.1: 117865435-117865435 on Build GRCh38
rs1055550099 Chr.1: 117865675-117865675 on Build GRCh38
rs920323211 Chr.1: 117865730-117865730 on Build GRCh38
rs535862869 Chr.1: 117865665-117865665 on Build GRCh38
rs868655437 Chr.1: 117865713-117865713 on Build GRCh38
rs1014258712 Chr.1: 117865745-117865745 on Build GRCh38
rs1001029405 Chr.1: 117865848-117865848 on Build GRCh38
rs17037690 Chr.1: 117865653-117865653 on Build GRCh38
rs944921065 Chr.1: 117865764-117865764 on Build GRCh38
rs907680286 Chr.1: 117865364-117865364 on Build GRCh38
rs17037692 Chr.1: 117865835-117865835 on Build GRCh38
rs188897326 Chr.1: 117865427-117865427 on Build GRCh38
rs757580831 Chr.1: 117865541-117865541 on Build GRCh38
rs981742822 Chr.1: 117865391-117865391 on Build GRCh38
rs558375855 Chr.1: 117865797-117865797 on Build GRCh38
rs1010721167 Chr.1: 117865596-117865597 on Build GRCh38
rs181307388 Chr.1: 117865584-117865584 on Build GRCh38
rs954799866 Chr.1: 117865598-117865598 on Build GRCh38
rs559145873 Chr.1: 117865413-117865413 on Build GRCh38
rs543301234 Chr.1: 117865741-117865741 on Build GRCh38
rs185668664 Chr.1: 117865597-117865597 on Build GRCh38
rs886669422 Chr.1: 117865496-117865496 on Build GRCh38
rs74113066 Chr.1: 117865736-117865736 on Build GRCh38
rs896761255 Chr.1: 117865453-117865453 on Build GRCh38

View Full Product Details