Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 117866402-117866925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDAP2
Entrez Gene ID
54834
Gene Name
ganglioside induced differentiation associated protein 2
Gene Aliases
MACROD3
UniGene
Hs.310809
Gene Chromosome Location
Chr.1: 117860227-117930042 on Build GRCh38
Transcripts
NM_017686.3 XM_005270959.2 XM_011541623.2 XM_017001531.1 XM_017001532.1

SNP Detail

SNP ID Chromosome Location
rs912017368 Chr.1: 117866682-117866682 on Build GRCh38
rs901565136 Chr.1: 117866460-117866460 on Build GRCh38
rs901514441 Chr.1: 117866657-117866657 on Build GRCh38
rs551342407 Chr.1: 117866875-117866875 on Build GRCh38
rs1024159007 Chr.1: 117866809-117866809 on Build GRCh38
rs989877602 Chr.1: 117866651-117866651 on Build GRCh38
rs932200896 Chr.1: 117866450-117866450 on Build GRCh38
rs931400855 Chr.1: 117866875-117866876 on Build GRCh38
rs1028727919 Chr.1: 117866686-117866686 on Build GRCh38
rs563327263 Chr.1: 117866461-117866461 on Build GRCh38
rs1052359722 Chr.1: 117866454-117866454 on Build GRCh38
rs776861034 Chr.1: 117866481-117866481 on Build GRCh38
rs561687334 Chr.1: 117866803-117866803 on Build GRCh38
rs4659349 Chr.1: 117866624-117866624 on Build GRCh38
rs1051965426 Chr.1: 117866680-117866680 on Build GRCh38
rs1014617582 Chr.1: 117866502-117866502 on Build GRCh38
rs1035986563 Chr.1: 117866842-117866842 on Build GRCh38
rs1012534419 Chr.1: 117866445-117866445 on Build GRCh38
rs748347908 Chr.1: 117866836-117866837 on Build GRCh38
rs911684702 Chr.1: 117866853-117866853 on Build GRCh38
rs928628262 Chr.1: 117866561-117866561 on Build GRCh38
rs762029455 Chr.1: 117866591-117866591 on Build GRCh38
rs572348959 Chr.1: 117866526-117866526 on Build GRCh38
rs1029197952 Chr.1: 117866505-117866505 on Build GRCh38
rs574516271 Chr.1: 117866448-117866448 on Build GRCh38
rs1011724358 Chr.1: 117866790-117866790 on Build GRCh38
rs940567155 Chr.1: 117866892-117866892 on Build GRCh38
rs978122614 Chr.1: 117866598-117866598 on Build GRCh38
rs909050720 Chr.1: 117866877-117866877 on Build GRCh38
rs894423556 Chr.1: 117866802-117866802 on Build GRCh38
rs940425573 Chr.1: 117866692-117866692 on Build GRCh38
rs191905406 Chr.1: 117866522-117866522 on Build GRCh38
rs528875358 Chr.1: 117866898-117866901 on Build GRCh38
rs12090075 Chr.1: 117866768-117866768 on Build GRCh38
rs977675572 Chr.1: 117866824-117866825 on Build GRCh38
rs150202760 Chr.1: 117866683-117866683 on Build GRCh38
rs922247982 Chr.1: 117866660-117866660 on Build GRCh38
rs967142867 Chr.1: 117866815-117866815 on Build GRCh38
rs1004479246 Chr.1: 117866452-117866452 on Build GRCh38
rs763073651 Chr.1: 117866731-117866731 on Build GRCh38
rs953149788 Chr.1: 117866821-117866821 on Build GRCh38
rs1036080474 Chr.1: 117866697-117866697 on Build GRCh38
rs898795475 Chr.1: 117866509-117866509 on Build GRCh38
rs539471079 Chr.1: 117866826-117866826 on Build GRCh38
rs576051716 Chr.1: 117866714-117866714 on Build GRCh38
rs543055593 Chr.1: 117866750-117866750 on Build GRCh38
rs529432951 Chr.1: 117866628-117866628 on Build GRCh38
rs958869486 Chr.1: 117866523-117866523 on Build GRCh38
rs970800469 Chr.1: 117866587-117866587 on Build GRCh38
rs997626107 Chr.1: 117866664-117866664 on Build GRCh38
rs960062516 Chr.1: 117866845-117866845 on Build GRCh38
rs898908366 Chr.1: 117866726-117866726 on Build GRCh38
rs991929606 Chr.1: 117866861-117866861 on Build GRCh38
rs867068661 Chr.1: 117866671-117866671 on Build GRCh38
rs1048897815 Chr.1: 117866619-117866619 on Build GRCh38
rs915972071 Chr.1: 117866869-117866869 on Build GRCh38
rs77334515 Chr.1: 117866433-117866433 on Build GRCh38
rs113493258 Chr.1: 117866616-117866616 on Build GRCh38
rs971925124 Chr.1: 117866887-117866887 on Build GRCh38
rs1029106307 Chr.1: 117866819-117866819 on Build GRCh38
rs150683842 Chr.1: 117866438-117866438 on Build GRCh38
rs547206665 Chr.1: 117866621-117866621 on Build GRCh38
rs940617864 Chr.1: 117866633-117866633 on Build GRCh38
rs7547388 Chr.1: 117866881-117866881 on Build GRCh38
rs987668729 Chr.1: 117866832-117866832 on Build GRCh38
rs80285278 Chr.1: 117866456-117866456 on Build GRCh38
rs146464120 Chr.1: 117866881-117866881 on Build GRCh38
rs1055852403 Chr.1: 117866798-117866798 on Build GRCh38
rs994544479 Chr.1: 117866733-117866733 on Build GRCh38
rs575323402 Chr.1: 117866517-117866517 on Build GRCh38
rs869074143 Chr.1: 117866875-117866881 on Build GRCh38
rs75759330 Chr.1: 117866541-117866541 on Build GRCh38

View Full Product Details