Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 117868130-117868665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDAP2
Entrez Gene ID
54834
Gene Name
ganglioside induced differentiation associated protein 2
Gene Aliases
MACROD3
UniGene
Hs.310809
Gene Chromosome Location
Chr.1: 117860227-117930042 on Build GRCh38
Transcripts
NM_017686.3 XM_005270959.2 XM_011541623.2 XM_017001531.1 XM_017001532.1

SNP Detail

SNP ID Chromosome Location
rs955678682 Chr.1: 117868239-117868239 on Build GRCh38
rs993535334 Chr.1: 117868193-117868193 on Build GRCh38
rs1023980414 Chr.1: 117868497-117868497 on Build GRCh38
rs929361147 Chr.1: 117868427-117868427 on Build GRCh38
rs545164983 Chr.1: 117868613-117868613 on Build GRCh38
rs1049037896 Chr.1: 117868199-117868199 on Build GRCh38
rs367610582 Chr.1: 117868398-117868398 on Build GRCh38
rs919362096 Chr.1: 117868419-117868419 on Build GRCh38
rs912393629 Chr.1: 117868393-117868393 on Build GRCh38
rs916960973 Chr.1: 117868254-117868254 on Build GRCh38
rs985525026 Chr.1: 117868564-117868564 on Build GRCh38
rs972900791 Chr.1: 117868579-117868579 on Build GRCh38
rs762227932 Chr.1: 117868181-117868181 on Build GRCh38
rs918870882 Chr.1: 117868362-117868362 on Build GRCh38
rs1051122345 Chr.1: 117868395-117868395 on Build GRCh38
rs769777160 Chr.1: 117868463-117868463 on Build GRCh38
rs1006976427 Chr.1: 117868403-117868403 on Build GRCh38
rs578040553 Chr.1: 117868553-117868553 on Build GRCh38
rs887761787 Chr.1: 117868436-117868436 on Build GRCh38
rs770593662 Chr.1: 117868257-117868257 on Build GRCh38
rs1011304319 Chr.1: 117868528-117868528 on Build GRCh38
rs576067624 Chr.1: 117868637-117868637 on Build GRCh38
rs948987285 Chr.1: 117868479-117868479 on Build GRCh38
rs914215127 Chr.1: 117868341-117868341 on Build GRCh38
rs980643545 Chr.1: 117868265-117868265 on Build GRCh38
rs953704807 Chr.1: 117868541-117868541 on Build GRCh38
rs1023572561 Chr.1: 117868225-117868225 on Build GRCh38
rs556060949 Chr.1: 117868363-117868363 on Build GRCh38
rs955772906 Chr.1: 117868280-117868280 on Build GRCh38
rs888052868 Chr.1: 117868207-117868207 on Build GRCh38
rs1031574334 Chr.1: 117868442-117868442 on Build GRCh38
rs954119649 Chr.1: 117868359-117868359 on Build GRCh38
rs971498015 Chr.1: 117868243-117868243 on Build GRCh38
rs1044617926 Chr.1: 117868480-117868480 on Build GRCh38
rs56072360 Chr.1: 117868435-117868435 on Build GRCh38
rs752527629 Chr.1: 117868565-117868565 on Build GRCh38
rs1003020305 Chr.1: 117868508-117868508 on Build GRCh38
rs776978108 Chr.1: 117868157-117868157 on Build GRCh38
rs17037698 Chr.1: 117868416-117868416 on Build GRCh38
rs539594143 Chr.1: 117868267-117868267 on Build GRCh38
rs142502287 Chr.1: 117868513-117868513 on Build GRCh38
rs6677641 Chr.1: 117868525-117868525 on Build GRCh38
rs979820101 Chr.1: 117868515-117868515 on Build GRCh38
rs1013139927 Chr.1: 117868223-117868223 on Build GRCh38
rs769985069 Chr.1: 117868248-117868248 on Build GRCh38
rs989803424 Chr.1: 117868318-117868318 on Build GRCh38
rs1031625313 Chr.1: 117868196-117868196 on Build GRCh38
rs372365061 Chr.1: 117868384-117868384 on Build GRCh38
rs965269177 Chr.1: 117868585-117868585 on Build GRCh38
rs557576150 Chr.1: 117868333-117868333 on Build GRCh38
rs139229461 Chr.1: 117868494-117868494 on Build GRCh38
rs1043870809 Chr.1: 117868409-117868409 on Build GRCh38
rs77169235 Chr.1: 117868537-117868537 on Build GRCh38

View Full Product Details