Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7758522-7759025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFAP1
Entrez Gene ID
60312
Gene Name
actin filament associated protein 1
Gene Aliases
AFAP, AFAP-110, AFAP110
UniGene
Hs.529369
Gene Chromosome Location
Chr.4: 7758713-7939926 on Build GRCh38
Transcripts
NM_001134647.1 NM_198595.2 XM_006713909.3 XM_011513544.2 XM_017008535.1 NR_026892.1

Gene Symbol
AFAP1-AS1
Entrez Gene ID
84740
Gene Name
AFAP1 antisense RNA 1
Gene Aliases
AFAP1-AS, AFAP1AS
UniGene
Hs.663029
Gene Chromosome Location
Chr.4: 7754090-7778928 on Build GRCh38
Transcripts
NM_001134647.1 NM_198595.2 XM_006713909.3 XM_011513544.2 XM_017008535.1 NR_026892.1

SNP Detail

SNP ID Chromosome Location
rs190109880 Chr.4: 7758931-7758931 on Build GRCh38
rs574668927 Chr.4: 7758588-7758589 on Build GRCh38
rs994017711 Chr.4: 7758611-7758611 on Build GRCh38
rs566057561 Chr.4: 7758598-7758598 on Build GRCh38
rs776589585 Chr.4: 7758916-7758916 on Build GRCh38
rs4689161 Chr.4: 7758584-7758584 on Build GRCh38
rs942993091 Chr.4: 7758985-7758985 on Build GRCh38
rs780596263 Chr.4: 7758560-7758560 on Build GRCh38
rs182365165 Chr.4: 7758636-7758636 on Build GRCh38
rs570892388 Chr.4: 7758620-7758620 on Build GRCh38
rs938568187 Chr.4: 7758949-7758949 on Build GRCh38
rs990791675 Chr.4: 7758644-7758644 on Build GRCh38
rs924138189 Chr.4: 7758690-7758690 on Build GRCh38
rs986895634 Chr.4: 7758967-7758967 on Build GRCh38
rs534871275 Chr.4: 7758709-7758709 on Build GRCh38
rs1034279266 Chr.4: 7758763-7758763 on Build GRCh38
rs574542719 Chr.4: 7758801-7758801 on Build GRCh38
rs1022346663 Chr.4: 7758833-7758833 on Build GRCh38
rs866214736 Chr.4: 7758791-7758791 on Build GRCh38
rs760638851 Chr.4: 7758813-7758813 on Build GRCh38
rs1031067362 Chr.4: 7758934-7758934 on Build GRCh38
rs764770165 Chr.4: 7758562-7758562 on Build GRCh38
rs35175103 Chr.4: 7758754-7758755 on Build GRCh38
rs1046795872 Chr.4: 7758749-7758749 on Build GRCh38
rs764185435 Chr.4: 7758844-7758844 on Build GRCh38
rs368479799 Chr.4: 7758797-7758797 on Build GRCh38
rs1012258326 Chr.4: 7758832-7758832 on Build GRCh38
rs1015838280 Chr.4: 7758743-7758743 on Build GRCh38
rs575368208 Chr.4: 7758864-7758864 on Build GRCh38
rs555883125 Chr.4: 7758614-7758614 on Build GRCh38
rs1037961364 Chr.4: 7758723-7758723 on Build GRCh38
rs971244899 Chr.4: 7758619-7758619 on Build GRCh38
rs756318485 Chr.4: 7758859-7758859 on Build GRCh38
rs911452407 Chr.4: 7758708-7758708 on Build GRCh38
rs934162954 Chr.4: 7758699-7758699 on Build GRCh38
rs1034248092 Chr.4: 7758633-7758633 on Build GRCh38
rs1013099052 Chr.4: 7758559-7758559 on Build GRCh38
rs2285772 Chr.4: 7758677-7758677 on Build GRCh38
rs959377422 Chr.4: 7758635-7758635 on Build GRCh38
rs977332400 Chr.4: 7758921-7758921 on Build GRCh38
rs907052642 Chr.4: 7758753-7758753 on Build GRCh38
rs968071163 Chr.4: 7758882-7758882 on Build GRCh38
rs978200984 Chr.4: 7758678-7758678 on Build GRCh38
rs915338078 Chr.4: 7758672-7758672 on Build GRCh38
rs1002684454 Chr.4: 7758759-7758759 on Build GRCh38
rs763892046 Chr.4: 7758668-7758668 on Build GRCh38
rs947194571 Chr.4: 7758904-7758904 on Build GRCh38
rs907575964 Chr.4: 7758571-7758571 on Build GRCh38
rs537026553 Chr.4: 7758651-7758651 on Build GRCh38
rs765225770 Chr.4: 7758939-7758939 on Build GRCh38
rs186654141 Chr.4: 7758812-7758812 on Build GRCh38
rs557405611 Chr.4: 7758851-7758851 on Build GRCh38
rs775827995 Chr.4: 7758802-7758802 on Build GRCh38
rs1013378958 Chr.4: 7758712-7758712 on Build GRCh38
rs572264579 Chr.4: 7758935-7758935 on Build GRCh38
rs565667156 Chr.4: 7758994-7758994 on Build GRCh38
rs750208926 Chr.4: 7758984-7758984 on Build GRCh38
rs888607259 Chr.4: 7758735-7758735 on Build GRCh38
rs79380309 Chr.4: 7758591-7758591 on Build GRCh38
rs889479680 Chr.4: 7758587-7758587 on Build GRCh38
rs955748805 Chr.4: 7758945-7758945 on Build GRCh38
rs139293785 Chr.4: 7758657-7758657 on Build GRCh38
rs553003330 Chr.4: 7758742-7758742 on Build GRCh38
rs986278798 Chr.4: 7758886-7758886 on Build GRCh38
rs975756771 Chr.4: 7758779-7758779 on Build GRCh38
rs914444332 Chr.4: 7758887-7758887 on Build GRCh38
rs774786104 Chr.4: 7758588-7758588 on Build GRCh38
rs752629300 Chr.4: 7758567-7758567 on Build GRCh38
rs879258877 Chr.4: 7758565-7758565 on Build GRCh38
rs994357372 Chr.4: 7758744-7758744 on Build GRCh38
rs914349754 Chr.4: 7758599-7758599 on Build GRCh38
rs1051312294 Chr.4: 7758703-7758703 on Build GRCh38
rs894465222 Chr.4: 7758970-7758974 on Build GRCh38
rs983437226 Chr.4: 7758563-7758563 on Build GRCh38

View Full Product Details