Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7760056-7760549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFAP1
Entrez Gene ID
60312
Gene Name
actin filament associated protein 1
Gene Aliases
AFAP, AFAP-110, AFAP110
UniGene
Hs.529369
Gene Chromosome Location
Chr.4: 7758713-7939926 on Build GRCh38
Transcripts
NM_001134647.1 NM_198595.2 XM_006713909.3 XM_011513544.2 XM_017008535.1 NR_026892.1

Gene Symbol
AFAP1-AS1
Entrez Gene ID
84740
Gene Name
AFAP1 antisense RNA 1
Gene Aliases
AFAP1-AS, AFAP1AS
UniGene
Hs.663029
Gene Chromosome Location
Chr.4: 7754090-7778928 on Build GRCh38
Transcripts
NM_001134647.1 NM_198595.2 XM_006713909.3 XM_011513544.2 XM_017008535.1 NR_026892.1

SNP Detail

SNP ID Chromosome Location
rs879724321 Chr.4: 7760382-7760382 on Build GRCh38
rs1005230316 Chr.4: 7760525-7760525 on Build GRCh38
rs926913449 Chr.4: 7760318-7760318 on Build GRCh38
rs892529505 Chr.4: 7760383-7760383 on Build GRCh38
rs192755165 Chr.4: 7760119-7760119 on Build GRCh38
rs1050986735 Chr.4: 7760171-7760171 on Build GRCh38
rs188692065 Chr.4: 7760341-7760341 on Build GRCh38
rs983387672 Chr.4: 7760319-7760319 on Build GRCh38
rs1000900707 Chr.4: 7760228-7760228 on Build GRCh38
rs1014818264 Chr.4: 7760527-7760527 on Build GRCh38
rs184158957 Chr.4: 7760260-7760260 on Build GRCh38
rs952018753 Chr.4: 7760267-7760267 on Build GRCh38
rs373804136 Chr.4: 7760377-7760377 on Build GRCh38
rs755306542 Chr.4: 7760394-7760395 on Build GRCh38
rs547086285 Chr.4: 7760090-7760090 on Build GRCh38
rs572604449 Chr.4: 7760244-7760244 on Build GRCh38
rs973710356 Chr.4: 7760443-7760443 on Build GRCh38
rs184509713 Chr.4: 7760436-7760436 on Build GRCh38
rs1027942965 Chr.4: 7760485-7760485 on Build GRCh38
rs138472026 Chr.4: 7760468-7760468 on Build GRCh38
rs891051409 Chr.4: 7760364-7760364 on Build GRCh38
rs999866862 Chr.4: 7760159-7760159 on Build GRCh38
rs545609499 Chr.4: 7760177-7760177 on Build GRCh38
rs896988141 Chr.4: 7760135-7760135 on Build GRCh38
rs964532654 Chr.4: 7760258-7760258 on Build GRCh38
rs929675958 Chr.4: 7760465-7760465 on Build GRCh38
rs1008631659 Chr.4: 7760231-7760231 on Build GRCh38
rs1057223081 Chr.4: 7760108-7760108 on Build GRCh38
rs936284004 Chr.4: 7760402-7760402 on Build GRCh38
rs779075086 Chr.4: 7760418-7760418 on Build GRCh38
rs534008897 Chr.4: 7760406-7760406 on Build GRCh38
rs1006906247 Chr.4: 7760212-7760212 on Build GRCh38
rs112548370 Chr.4: 7760372-7760372 on Build GRCh38
rs1013721632 Chr.4: 7760385-7760385 on Build GRCh38
rs538228218 Chr.4: 7760488-7760488 on Build GRCh38
rs71175428 Chr.4: 7760169-7760170 on Build GRCh38
rs73080598 Chr.4: 7760125-7760125 on Build GRCh38
rs781322920 Chr.4: 7760162-7760162 on Build GRCh38
rs918352545 Chr.4: 7760461-7760461 on Build GRCh38
rs970017140 Chr.4: 7760301-7760301 on Build GRCh38
rs550194585 Chr.4: 7760355-7760355 on Build GRCh38
rs917307001 Chr.4: 7760359-7760359 on Build GRCh38
rs562431598 Chr.4: 7760369-7760369 on Build GRCh38
rs927075884 Chr.4: 7760110-7760110 on Build GRCh38
rs1053361898 Chr.4: 7760403-7760403 on Build GRCh38
rs896009282 Chr.4: 7760109-7760109 on Build GRCh38
rs1027583336 Chr.4: 7760262-7760262 on Build GRCh38
rs578258887 Chr.4: 7760137-7760137 on Build GRCh38
rs114614723 Chr.4: 7760186-7760186 on Build GRCh38
rs979801284 Chr.4: 7760317-7760317 on Build GRCh38
rs931346469 Chr.4: 7760087-7760087 on Build GRCh38
rs556636519 Chr.4: 7760493-7760493 on Build GRCh38
rs916927822 Chr.4: 7760088-7760088 on Build GRCh38
rs762815887 Chr.4: 7760342-7760342 on Build GRCh38
rs191600732 Chr.4: 7760370-7760370 on Build GRCh38
rs796627281 Chr.4: 7760318-7760319 on Build GRCh38
rs891450920 Chr.4: 7760203-7760203 on Build GRCh38
rs1025531117 Chr.4: 7760397-7760397 on Build GRCh38
rs566510010 Chr.4: 7760376-7760376 on Build GRCh38
rs912380372 Chr.4: 7760492-7760492 on Build GRCh38
rs1038112764 Chr.4: 7760340-7760340 on Build GRCh38
rs540950290 Chr.4: 7760405-7760405 on Build GRCh38
rs968160709 Chr.4: 7760219-7760219 on Build GRCh38
rs918195282 Chr.4: 7760289-7760289 on Build GRCh38
rs900229222 Chr.4: 7760442-7760442 on Build GRCh38
rs992928882 Chr.4: 7760357-7760357 on Build GRCh38
rs773305152 Chr.4: 7760325-7760325 on Build GRCh38
rs115339438 Chr.4: 7760094-7760094 on Build GRCh38
rs565919987 Chr.4: 7760249-7760249 on Build GRCh38
rs200519161 Chr.4: 7760435-7760436 on Build GRCh38
rs561842819 Chr.4: 7760268-7760268 on Build GRCh38
rs908628859 Chr.4: 7760093-7760093 on Build GRCh38
rs926139338 Chr.4: 7760373-7760373 on Build GRCh38
rs748199292 Chr.4: 7760218-7760218 on Build GRCh38

View Full Product Details