Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7983147-7983636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABLIM2
Entrez Gene ID
84448
Gene Name
actin binding LIM protein family member 2
Gene Aliases
-
UniGene
Hs.233404
Gene Chromosome Location
Chr.4: 7965310-8158832 on Build GRCh38
Transcripts
NM_001130083.1 NM_001130084.1 NM_001130085.1 NM_001130086.1 NM_001130087.1 NM_001286688.1 NM_032432.4 XM_005248014.4 XM_005248017.4 XM_005248018.4 XM_005248019.4 XM_005248020.4 XM_005248021.4 XM_005248022.4 XM_005248023.4 XM_005248024.4 XM_005248027.4 XM_005248028.4 XM_005248029.4 XM_005248030.4 XM_006713923.3 XM_006713924.3 XM_006713925.3 XM_006713927.3 XM_011513586.2 XM_017008706.1 XM_017008707.1 XM_017008708.1 XM_017008709.1 XM_017008710.1 XM_017008711.1 XM_017008712.1 XM_017008713.1 XM_017008714.1 XM_017008715.1 XM_017008716.1 XM_017008717.1 XM_017008718.1 XM_017008719.1 XM_017008720.1 XM_017008721.1 XM_017008722.1 XM_017008724.1 XM_017008726.1 XM_017008727.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4472823 c.1467C>T p.D489D Chr.4: 7983279-7983279 on Build GRCh38
5060499 c.1433A>G p.N478S Chr.4: 7983313-7983313 on Build GRCh38
6222791 c.1409C>T p.P470L Chr.4: 7983337-7983337 on Build GRCh38
7259956 c.1394-6_1394-5insCT p.? Chr.4: 7983559-7983560 on Build GRCh38
7259955 c.1736-6_1736-5insCT p.? Chr.4: 7983559-7983560 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778095749 Chr.4: 7983323-7983323 on Build GRCh38
rs566338028 Chr.4: 7983165-7983165 on Build GRCh38
rs371079889 Chr.4: 7983438-7983438 on Build GRCh38
rs746566402 Chr.4: 7983573-7983573 on Build GRCh38
rs75115054 Chr.4: 7983363-7983363 on Build GRCh38
rs758817750 Chr.4: 7983196-7983196 on Build GRCh38
rs907919600 Chr.4: 7983213-7983213 on Build GRCh38
rs866670061 Chr.4: 7983518-7983518 on Build GRCh38
rs752650225 Chr.4: 7983383-7983383 on Build GRCh38
rs370012949 Chr.4: 7983569-7983569 on Build GRCh38
rs757435517 Chr.4: 7983396-7983396 on Build GRCh38
rs369888216 Chr.4: 7983476-7983476 on Build GRCh38
rs770773180 Chr.4: 7983581-7983581 on Build GRCh38
rs764888243 Chr.4: 7983182-7983182 on Build GRCh38
rs199606489 Chr.4: 7983517-7983517 on Build GRCh38
rs759444010 Chr.4: 7983516-7983516 on Build GRCh38
rs145815093 Chr.4: 7983439-7983439 on Build GRCh38
rs750016618 Chr.4: 7983304-7983304 on Build GRCh38
rs200961918 Chr.4: 7983559-7983560 on Build GRCh38
rs368085466 Chr.4: 7983246-7983246 on Build GRCh38
rs759293120 Chr.4: 7983585-7983585 on Build GRCh38
rs1052551908 Chr.4: 7983459-7983459 on Build GRCh38
rs948019131 Chr.4: 7983418-7983418 on Build GRCh38
rs772599189 Chr.4: 7983512-7983512 on Build GRCh38
rs775974578 Chr.4: 7983349-7983349 on Build GRCh38
rs754967373 Chr.4: 7983315-7983315 on Build GRCh38
rs776472115 Chr.4: 7983582-7983582 on Build GRCh38
rs369413327 Chr.4: 7983389-7983389 on Build GRCh38
rs919757119 Chr.4: 7983193-7983193 on Build GRCh38
rs372895534 Chr.4: 7983309-7983309 on Build GRCh38
rs752913991 Chr.4: 7983560-7983561 on Build GRCh38
rs201156123 Chr.4: 7983277-7983277 on Build GRCh38
rs766059141 Chr.4: 7983373-7983373 on Build GRCh38
rs764007420 Chr.4: 7983603-7983603 on Build GRCh38
rs756409784 Chr.4: 7983560-7983563 on Build GRCh38
rs1028890650 Chr.4: 7983212-7983212 on Build GRCh38
rs749446854 Chr.4: 7983511-7983511 on Build GRCh38
rs200231234 Chr.4: 7983303-7983303 on Build GRCh38
rs532562201 Chr.4: 7983509-7983509 on Build GRCh38
rs991331351 Chr.4: 7983591-7983591 on Build GRCh38
rs150701353 Chr.4: 7983243-7983243 on Build GRCh38
rs925495021 Chr.4: 7983422-7983422 on Build GRCh38
rs761167311 Chr.4: 7983273-7983273 on Build GRCh38
rs780076428 Chr.4: 7983252-7983252 on Build GRCh38
rs1036980550 Chr.4: 7983342-7983342 on Build GRCh38
rs200508979 Chr.4: 7983320-7983320 on Build GRCh38
rs770347292 Chr.4: 7983348-7983348 on Build GRCh38
rs963993798 Chr.4: 7983172-7983172 on Build GRCh38
rs768663886 Chr.4: 7983255-7983255 on Build GRCh38
rs772349719 Chr.4: 7983259-7983259 on Build GRCh38
rs766106415 Chr.4: 7983310-7983310 on Build GRCh38
rs749597966 Chr.4: 7983549-7983549 on Build GRCh38
rs753644689 Chr.4: 7983312-7983312 on Build GRCh38
rs762797806 Chr.4: 7983597-7983597 on Build GRCh38
rs763757729 Chr.4: 7983570-7983571 on Build GRCh38
rs922565060 Chr.4: 7983612-7983612 on Build GRCh38
rs773292428 Chr.4: 7983270-7983270 on Build GRCh38
rs760273214 Chr.4: 7983368-7983368 on Build GRCh38
rs780653832 Chr.4: 7983242-7983242 on Build GRCh38
rs779162927 Chr.4: 7983257-7983257 on Build GRCh38
rs948310217 Chr.4: 7983235-7983235 on Build GRCh38
rs780423693 Chr.4: 7983541-7983541 on Build GRCh38
rs746175441 Chr.4: 7983338-7983338 on Build GRCh38
rs891319584 Chr.4: 7983464-7983464 on Build GRCh38
rs1044395799 Chr.4: 7983238-7983238 on Build GRCh38
rs747046089 Chr.4: 7983390-7983390 on Build GRCh38
rs375999116 Chr.4: 7983377-7983377 on Build GRCh38
rs560054422 Chr.4: 7983321-7983321 on Build GRCh38
rs998548162 Chr.4: 7983495-7983495 on Build GRCh38
rs747471823 Chr.4: 7983284-7983284 on Build GRCh38
rs776417134 Chr.4: 7983515-7983515 on Build GRCh38
rs977077502 Chr.4: 7983247-7983247 on Build GRCh38
rs760531559 Chr.4: 7983514-7983514 on Build GRCh38
rs758403367 Chr.4: 7983322-7983322 on Build GRCh38
rs754295363 Chr.4: 7983221-7983221 on Build GRCh38
rs766967949 Chr.4: 7983216-7983216 on Build GRCh38
rs1021419398 Chr.4: 7983449-7983449 on Build GRCh38
rs376436867 Chr.4: 7983540-7983540 on Build GRCh38
rs775163452 Chr.4: 7983592-7983592 on Build GRCh38
rs901424739 Chr.4: 7983207-7983207 on Build GRCh38
rs748171294 Chr.4: 7983258-7983258 on Build GRCh38
rs1043749952 Chr.4: 7983420-7983420 on Build GRCh38
rs367611964 Chr.4: 7983336-7983336 on Build GRCh38
rs758307287 Chr.4: 7983384-7983384 on Build GRCh38
rs747218662 Chr.4: 7983331-7983331 on Build GRCh38
rs749212199 Chr.4: 7983254-7983254 on Build GRCh38
rs769506006 Chr.4: 7983588-7983588 on Build GRCh38
rs779460847 Chr.4: 7983572-7983572 on Build GRCh38
rs371140553 Chr.4: 7983274-7983274 on Build GRCh38
rs369579061 Chr.4: 7983526-7983526 on Build GRCh38
rs765667522 Chr.4: 7983285-7983285 on Build GRCh38
rs760889081 Chr.4: 7983215-7983215 on Build GRCh38
rs370214035 Chr.4: 7983230-7983230 on Build GRCh38
rs187487439 Chr.4: 7983183-7983183 on Build GRCh38
rs746302043 Chr.4: 7983506-7983506 on Build GRCh38
rs35503576 Chr.4: 7983290-7983291 on Build GRCh38
rs149817377 Chr.4: 7983398-7983398 on Build GRCh38
rs982630913 Chr.4: 7983197-7983197 on Build GRCh38
rs6833003 Chr.4: 7983354-7983354 on Build GRCh38
rs776029845 Chr.4: 7983510-7983510 on Build GRCh38
rs373344302 Chr.4: 7983535-7983535 on Build GRCh38
rs772725032 Chr.4: 7983364-7983364 on Build GRCh38
rs755982683 Chr.4: 7983305-7983305 on Build GRCh38
rs998393892 Chr.4: 7983208-7983208 on Build GRCh38
rs17179948 Chr.4: 7983191-7983191 on Build GRCh38
rs757326290 Chr.4: 7983534-7983534 on Build GRCh38
rs375619424 Chr.4: 7983222-7983222 on Build GRCh38
rs773867605 Chr.4: 7983286-7983286 on Build GRCh38
rs961423903 Chr.4: 7983593-7983593 on Build GRCh38
rs201190788 Chr.4: 7983609-7983609 on Build GRCh38
rs751031972 Chr.4: 7983226-7983226 on Build GRCh38
rs374407006 Chr.4: 7983301-7983301 on Build GRCh38
rs751650705 Chr.4: 7983533-7983533 on Build GRCh38
rs752497933 Chr.4: 7983525-7983525 on Build GRCh38
rs771145414 Chr.4: 7983333-7983333 on Build GRCh38

View Full Product Details