Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7992679-7993174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABLIM2
Entrez Gene ID
84448
Gene Name
actin binding LIM protein family member 2
Gene Aliases
-
UniGene
Hs.233404
Gene Chromosome Location
Chr.4: 7965310-8158832 on Build GRCh38
Transcripts
NM_001130083.1 NM_001130084.1 NM_001130085.1 NM_001130086.1 NM_001130087.1 NM_001286688.1 NM_032432.4 XM_005248014.4 XM_005248017.4 XM_005248018.4 XM_005248019.4 XM_005248020.4 XM_005248021.4 XM_005248022.4 XM_005248023.4 XM_005248024.4 XM_005248027.4 XM_005248028.4 XM_005248029.4 XM_005248030.4 XM_006713923.3 XM_006713924.3 XM_006713925.3 XM_006713927.3 XM_011513586.2 XM_017008706.1 XM_017008707.1 XM_017008708.1 XM_017008709.1 XM_017008710.1 XM_017008711.1 XM_017008712.1 XM_017008713.1 XM_017008714.1 XM_017008715.1 XM_017008716.1 XM_017008717.1 XM_017008718.1 XM_017008719.1 XM_017008720.1 XM_017008721.1 XM_017008722.1 XM_017008724.1 XM_017008726.1 XM_017008727.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1057987 c.1664A>C p.N555T Chr.4: 7992882-7992882 on Build GRCh38
3826303 c.1662G>T p.K554N Chr.4: 7992884-7992884 on Build GRCh38
361258 c.1623C>T p.F541F Chr.4: 7992923-7992923 on Build GRCh38
3606320 c.1657G>A p.G553R Chr.4: 7992889-7992889 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1049577695 Chr.4: 7992845-7992845 on Build GRCh38
rs200683603 Chr.4: 7992951-7992951 on Build GRCh38
rs1048264630 Chr.4: 7992993-7992993 on Build GRCh38
rs1035996375 Chr.4: 7992762-7992762 on Build GRCh38
rs780986806 Chr.4: 7992954-7992954 on Build GRCh38
rs138524467 Chr.4: 7993137-7993137 on Build GRCh38
rs969597105 Chr.4: 7992755-7992755 on Build GRCh38
rs377566520 Chr.4: 7992834-7992834 on Build GRCh38
rs1032539894 Chr.4: 7993121-7993121 on Build GRCh38
rs537474866 Chr.4: 7993134-7993134 on Build GRCh38
rs1042830503 Chr.4: 7993017-7993017 on Build GRCh38
rs1052110551 Chr.4: 7992805-7992805 on Build GRCh38
rs754668427 Chr.4: 7992866-7992866 on Build GRCh38
rs776785180 Chr.4: 7992883-7992883 on Build GRCh38
rs767965422 Chr.4: 7993031-7993031 on Build GRCh38
rs1023909718 Chr.4: 7992904-7992904 on Build GRCh38
rs770119507 Chr.4: 7992892-7992892 on Build GRCh38
rs201636882 Chr.4: 7992968-7992968 on Build GRCh38
rs753448837 Chr.4: 7992924-7992924 on Build GRCh38
rs764740616 Chr.4: 7992851-7992851 on Build GRCh38
rs1014647268 Chr.4: 7993118-7993118 on Build GRCh38
rs200114935 Chr.4: 7992960-7992960 on Build GRCh38
rs548037920 Chr.4: 7993012-7993012 on Build GRCh38
rs912465171 Chr.4: 7992792-7992792 on Build GRCh38
rs998927001 Chr.4: 7993039-7993039 on Build GRCh38
rs752092614 Chr.4: 7992855-7992855 on Build GRCh38
rs932969998 Chr.4: 7993001-7993001 on Build GRCh38
rs997483036 Chr.4: 7992778-7992778 on Build GRCh38
rs761161566 Chr.4: 7992829-7992829 on Build GRCh38
rs567018094 Chr.4: 7993103-7993103 on Build GRCh38
rs556466250 Chr.4: 7992868-7992868 on Build GRCh38
rs943964322 Chr.4: 7992800-7992800 on Build GRCh38
rs776199866 Chr.4: 7992785-7992785 on Build GRCh38
rs969922302 Chr.4: 7992941-7992941 on Build GRCh38
rs561324407 Chr.4: 7992961-7992961 on Build GRCh38
rs1051711800 Chr.4: 7993005-7993005 on Build GRCh38
rs754226986 Chr.4: 7993041-7993041 on Build GRCh38
rs759832745 Chr.4: 7992825-7992825 on Build GRCh38
rs572241651 Chr.4: 7992959-7992959 on Build GRCh38
rs201721300 Chr.4: 7992946-7992946 on Build GRCh38
rs934975707 Chr.4: 7992768-7992768 on Build GRCh38
rs913964343 Chr.4: 7992748-7992748 on Build GRCh38
rs1004839307 Chr.4: 7992857-7992857 on Build GRCh38
rs1045690537 Chr.4: 7993086-7993086 on Build GRCh38
rs916112726 Chr.4: 7992719-7992719 on Build GRCh38
rs777576520 Chr.4: 7992945-7992945 on Build GRCh38
rs745998768 Chr.4: 7992956-7992956 on Build GRCh38
rs979569586 Chr.4: 7992757-7992757 on Build GRCh38
rs984292831 Chr.4: 7993146-7993146 on Build GRCh38
rs909285409 Chr.4: 7992994-7992994 on Build GRCh38
rs908936863 Chr.4: 7992756-7992756 on Build GRCh38
rs201611635 Chr.4: 7992867-7992867 on Build GRCh38
rs371373214 Chr.4: 7992958-7992958 on Build GRCh38
rs375843410 Chr.4: 7992914-7992914 on Build GRCh38
rs35219850 Chr.4: 7992733-7992733 on Build GRCh38
rs1015123737 Chr.4: 7992717-7992717 on Build GRCh38
rs548679711 Chr.4: 7993095-7993095 on Build GRCh38
rs530187100 Chr.4: 7993046-7993046 on Build GRCh38
rs750867017 Chr.4: 7992971-7992971 on Build GRCh38
rs970783546 Chr.4: 7992760-7992760 on Build GRCh38
rs116559239 Chr.4: 7993151-7993151 on Build GRCh38
rs757658947 Chr.4: 7992859-7992859 on Build GRCh38
rs553699602 Chr.4: 7992957-7992957 on Build GRCh38
rs748684051 Chr.4: 7992861-7992861 on Build GRCh38
rs113235836 Chr.4: 7993142-7993142 on Build GRCh38
rs987843192 Chr.4: 7992775-7992775 on Build GRCh38
rs1007362252 Chr.4: 7993010-7993010 on Build GRCh38
rs763457214 Chr.4: 7992903-7992903 on Build GRCh38
rs41266509 Chr.4: 7992999-7992999 on Build GRCh38
rs566598676 Chr.4: 7992705-7992705 on Build GRCh38
rs921197509 Chr.4: 7992973-7992973 on Build GRCh38
rs548642564 Chr.4: 7992764-7992764 on Build GRCh38
rs887904895 Chr.4: 7992854-7992854 on Build GRCh38
rs142057971 Chr.4: 7992716-7992717 on Build GRCh38
rs1024252645 Chr.4: 7992753-7992753 on Build GRCh38
rs1044859777 Chr.4: 7992754-7992754 on Build GRCh38
rs776524188 Chr.4: 7992824-7992824 on Build GRCh38
rs1013592012 Chr.4: 7993048-7993048 on Build GRCh38
rs762382809 Chr.4: 7992912-7992912 on Build GRCh38
rs372788097 Chr.4: 7992828-7992828 on Build GRCh38
rs182893687 Chr.4: 7992821-7992821 on Build GRCh38
rs374293632 Chr.4: 7993038-7993038 on Build GRCh38
rs113135992 Chr.4: 7992928-7992928 on Build GRCh38
rs775822582 Chr.4: 7992897-7992897 on Build GRCh38
rs931133599 Chr.4: 7992987-7992987 on Build GRCh38
rs757998895 Chr.4: 7992944-7992944 on Build GRCh38
rs751928912 Chr.4: 7992907-7992907 on Build GRCh38
rs752326759 Chr.4: 7992940-7992940 on Build GRCh38
rs941723236 Chr.4: 7992982-7992982 on Build GRCh38
rs770733990 Chr.4: 7992878-7992878 on Build GRCh38
rs1039635221 Chr.4: 7992808-7992808 on Build GRCh38
rs998813949 Chr.4: 7992873-7992873 on Build GRCh38
rs754444694 Chr.4: 7992929-7992929 on Build GRCh38
rs897747885 Chr.4: 7992772-7992774 on Build GRCh38
rs532679318 Chr.4: 7993009-7993009 on Build GRCh38
rs767858259 Chr.4: 7992920-7992920 on Build GRCh38
rs778579147 Chr.4: 7992934-7992934 on Build GRCh38
rs759520777 Chr.4: 7992884-7992884 on Build GRCh38
rs538578923 Chr.4: 7993150-7993150 on Build GRCh38
rs960303126 Chr.4: 7992951-7992952 on Build GRCh38
rs375569090 Chr.4: 7992964-7992964 on Build GRCh38

View Full Product Details