Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8007341-8007834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABLIM2
Entrez Gene ID
84448
Gene Name
actin binding LIM protein family member 2
Gene Aliases
-
UniGene
Hs.233404
Gene Chromosome Location
Chr.4: 7965310-8158832 on Build GRCh38
Transcripts
NM_001130083.1 NM_001130084.1 NM_001130085.1 NM_001130086.1 NM_001130087.1 NM_001130088.1 NM_001286688.1 NM_032432.4 XM_005248014.4 XM_005248017.4 XM_005248018.4 XM_005248019.4 XM_005248020.4 XM_005248021.4 XM_005248022.4 XM_005248023.4 XM_005248024.4 XM_005248027.4 XM_005248028.4 XM_005248029.4 XM_005248030.4 XM_005248031.4 XM_006713923.3 XM_006713924.3 XM_006713925.3 XM_006713927.3 XM_011513586.2 XM_017008706.1 XM_017008707.1 XM_017008708.1 XM_017008709.1 XM_017008710.1 XM_017008711.1 XM_017008712.1 XM_017008713.1 XM_017008714.1 XM_017008715.1 XM_017008716.1 XM_017008717.1 XM_017008718.1 XM_017008719.1 XM_017008720.1 XM_017008721.1 XM_017008722.1 XM_017008723.1 XM_017008724.1 XM_017008725.1 XM_017008726.1 XM_017008727.1

SNP Detail

SNP ID Chromosome Location
rs141451130 Chr.4: 8007399-8007399 on Build GRCh38
rs905031118 Chr.4: 8007624-8007624 on Build GRCh38
rs542924453 Chr.4: 8007741-8007741 on Build GRCh38
rs111347589 Chr.4: 8007414-8007414 on Build GRCh38
rs749871298 Chr.4: 8007712-8007712 on Build GRCh38
rs1054035940 Chr.4: 8007638-8007638 on Build GRCh38
rs893196804 Chr.4: 8007639-8007639 on Build GRCh38
rs963490160 Chr.4: 8007409-8007409 on Build GRCh38
rs1044985037 Chr.4: 8007620-8007620 on Build GRCh38
rs896361599 Chr.4: 8007378-8007378 on Build GRCh38
rs112685505 Chr.4: 8007469-8007469 on Build GRCh38
rs6817309 Chr.4: 8007487-8007487 on Build GRCh38
rs905062333 Chr.4: 8007400-8007400 on Build GRCh38
rs1029284920 Chr.4: 8007709-8007709 on Build GRCh38
rs1016793030 Chr.4: 8007408-8007408 on Build GRCh38
rs987731683 Chr.4: 8007659-8007659 on Build GRCh38
rs892106523 Chr.4: 8007529-8007529 on Build GRCh38
rs764821171 Chr.4: 8007656-8007656 on Build GRCh38
rs538356608 Chr.4: 8007563-8007563 on Build GRCh38
rs752122035 Chr.4: 8007676-8007676 on Build GRCh38
rs572704752 Chr.4: 8007740-8007740 on Build GRCh38
rs926507971 Chr.4: 8007779-8007779 on Build GRCh38
rs143754025 Chr.4: 8007586-8007586 on Build GRCh38
rs997766858 Chr.4: 8007704-8007704 on Build GRCh38
rs535666844 Chr.4: 8007625-8007625 on Build GRCh38
rs936420692 Chr.4: 8007525-8007525 on Build GRCh38
rs775460437 Chr.4: 8007447-8007447 on Build GRCh38
rs987700673 Chr.4: 8007455-8007455 on Build GRCh38
rs578106297 Chr.4: 8007655-8007655 on Build GRCh38
rs553705553 Chr.4: 8007753-8007753 on Build GRCh38
rs146625899 Chr.4: 8007374-8007374 on Build GRCh38
rs949093318 Chr.4: 8007752-8007752 on Build GRCh38
rs908930794 Chr.4: 8007527-8007527 on Build GRCh38
rs78613406 Chr.4: 8007436-8007436 on Build GRCh38
rs183159766 Chr.4: 8007515-8007515 on Build GRCh38
rs759558544 Chr.4: 8007424-8007424 on Build GRCh38
rs562180527 Chr.4: 8007362-8007362 on Build GRCh38
rs1013937012 Chr.4: 8007388-8007388 on Build GRCh38
rs535526045 Chr.4: 8007749-8007749 on Build GRCh38
rs1020722698 Chr.4: 8007437-8007437 on Build GRCh38
rs910842162 Chr.4: 8007687-8007687 on Build GRCh38
rs980647558 Chr.4: 8007776-8007776 on Build GRCh38
rs953567264 Chr.4: 8007494-8007494 on Build GRCh38
rs1013458180 Chr.4: 8007565-8007565 on Build GRCh38
rs957592925 Chr.4: 8007416-8007416 on Build GRCh38
rs984533187 Chr.4: 8007498-8007498 on Build GRCh38
rs966465906 Chr.4: 8007700-8007700 on Build GRCh38
rs949088209 Chr.4: 8007761-8007761 on Build GRCh38
rs922078013 Chr.4: 8007493-8007493 on Build GRCh38
rs75669199 Chr.4: 8007654-8007654 on Build GRCh38
rs940435914 Chr.4: 8007530-8007530 on Build GRCh38
rs949290094 Chr.4: 8007597-8007597 on Build GRCh38
rs892139103 Chr.4: 8007757-8007757 on Build GRCh38
rs561161891 Chr.4: 8007748-8007748 on Build GRCh38
rs1036002774 Chr.4: 8007567-8007567 on Build GRCh38
rs1016078379 Chr.4: 8007621-8007621 on Build GRCh38
rs553842723 Chr.4: 8007738-8007738 on Build GRCh38
rs368446178 Chr.4: 8007406-8007406 on Build GRCh38
rs984915117 Chr.4: 8007736-8007736 on Build GRCh38
rs374685971 Chr.4: 8007691-8007691 on Build GRCh38

View Full Product Details